Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 157332733-157333241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA2
Entrez Gene ID
2891
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Gene Aliases
GLUR2, GLURB, GluA2, GluR-K2, HBGR2
UniGene
Hs.32763
Gene Chromosome Location
Chr.4: 157205099-157370583 on Build GRCh38
Transcripts
NM_000826.3 NM_001083619.1 NM_001083620.1 XM_017008113.1 XM_017008114.1 XM_017008115.1 XM_017008116.1 XM_017008117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029866 c.896T>A p.L299Q Chr.4: 157332832-157332832 on Build GRCh38
1052595 c.958G>C p.E320Q Chr.4: 157332894-157332894 on Build GRCh38
381546 c.972G>T p.R324S Chr.4: 157332908-157332908 on Build GRCh38
4123096 c.1002A>G p.P334P Chr.4: 157332938-157332938 on Build GRCh38
1052599 c.997A>C p.N333H Chr.4: 157332933-157332933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919170590 Chr.4: 157332983-157332983 on Build GRCh38
rs150963106 Chr.4: 157332947-157332947 on Build GRCh38
rs142738440 Chr.4: 157332755-157332755 on Build GRCh38
rs777296512 Chr.4: 157332871-157332871 on Build GRCh38
rs773054719 Chr.4: 157332819-157332819 on Build GRCh38
rs112635183 Chr.4: 157333139-157333139 on Build GRCh38
rs113378576 Chr.4: 157333217-157333217 on Build GRCh38
rs767317250 Chr.4: 157332845-157332845 on Build GRCh38
rs759450137 Chr.4: 157332990-157332990 on Build GRCh38
rs761788910 Chr.4: 157332953-157332953 on Build GRCh38
rs375083724 Chr.4: 157332846-157332846 on Build GRCh38
rs1024084113 Chr.4: 157333033-157333033 on Build GRCh38
rs971901720 Chr.4: 157332968-157332968 on Build GRCh38
rs771056190 Chr.4: 157332961-157332961 on Build GRCh38
rs763523654 Chr.4: 157333007-157333007 on Build GRCh38
rs771228618 Chr.4: 157332787-157332787 on Build GRCh38
rs775036710 Chr.4: 157333006-157333006 on Build GRCh38
rs368699030 Chr.4: 157333025-157333025 on Build GRCh38
rs368332843 Chr.4: 157332880-157332880 on Build GRCh38
rs143505003 Chr.4: 157332903-157332903 on Build GRCh38
rs749750444 Chr.4: 157332775-157332775 on Build GRCh38
rs953443263 Chr.4: 157333194-157333194 on Build GRCh38
rs142538282 Chr.4: 157332904-157332904 on Build GRCh38
rs527751151 Chr.4: 157333086-157333086 on Build GRCh38
rs758369126 Chr.4: 157333029-157333029 on Build GRCh38
rs773563113 Chr.4: 157333047-157333047 on Build GRCh38
rs1014964953 Chr.4: 157333085-157333085 on Build GRCh38
rs34460606 Chr.4: 157332827-157332827 on Build GRCh38
rs386681114 Chr.4: 157333092-157333093 on Build GRCh38
rs768626790 Chr.4: 157332941-157332941 on Build GRCh38
rs531398207 Chr.4: 157332797-157332797 on Build GRCh38
rs908973504 Chr.4: 157332944-157332944 on Build GRCh38
rs763348848 Chr.4: 157333119-157333119 on Build GRCh38
rs371644615 Chr.4: 157332788-157332788 on Build GRCh38
rs746101408 Chr.4: 157332794-157332794 on Build GRCh38
rs760720089 Chr.4: 157332805-157332805 on Build GRCh38
rs914258259 Chr.4: 157333218-157333218 on Build GRCh38
rs372589339 Chr.4: 157333210-157333210 on Build GRCh38
rs1006300699 Chr.4: 157333070-157333070 on Build GRCh38
rs559286454 Chr.4: 157333099-157333099 on Build GRCh38
rs755601111 Chr.4: 157332848-157332848 on Build GRCh38
rs544927349 Chr.4: 157333148-157333148 on Build GRCh38
rs746273489 Chr.4: 157332909-157332909 on Build GRCh38
rs762882361 Chr.4: 157332821-157332821 on Build GRCh38
rs775661808 Chr.4: 157332798-157332798 on Build GRCh38
rs763795535 Chr.4: 157332899-157332899 on Build GRCh38
rs549518190 Chr.4: 157332809-157332809 on Build GRCh38
rs747218196 Chr.4: 157332920-157332920 on Build GRCh38
rs759399187 Chr.4: 157332831-157332831 on Build GRCh38
rs772405015 Chr.4: 157332915-157332915 on Build GRCh38
rs767368130 Chr.4: 157332994-157332994 on Build GRCh38
rs1043512006 Chr.4: 157333064-157333064 on Build GRCh38
rs200712576 Chr.4: 157332767-157332767 on Build GRCh38
rs375489479 Chr.4: 157332998-157332998 on Build GRCh38
rs766888300 Chr.4: 157333193-157333193 on Build GRCh38
rs774397311 Chr.4: 157332984-157332984 on Build GRCh38
rs577660460 Chr.4: 157333145-157333145 on Build GRCh38
rs780276830 Chr.4: 157332916-157332916 on Build GRCh38
rs866921546 Chr.4: 157333010-157333010 on Build GRCh38
rs1030225527 Chr.4: 157333178-157333178 on Build GRCh38
rs78067001 Chr.4: 157333065-157333065 on Build GRCh38
rs753444287 Chr.4: 157333012-157333012 on Build GRCh38
rs766292841 Chr.4: 157332825-157332825 on Build GRCh38
rs914311607 Chr.4: 157332795-157332795 on Build GRCh38
rs199965016 Chr.4: 157332875-157332875 on Build GRCh38

View Full Product Details