Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168216318-168216862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5

SNP Detail

SNP ID Chromosome Location
rs571259748 Chr.4: 168216593-168216593 on Build GRCh38
rs950454534 Chr.4: 168216505-168216505 on Build GRCh38
rs942681156 Chr.4: 168216758-168216758 on Build GRCh38
rs1015883814 Chr.4: 168216634-168216634 on Build GRCh38
rs560285388 Chr.4: 168216419-168216419 on Build GRCh38
rs527991512 Chr.4: 168216430-168216430 on Build GRCh38
rs375430839 Chr.4: 168216383-168216383 on Build GRCh38
rs996906454 Chr.4: 168216556-168216556 on Build GRCh38
rs935812639 Chr.4: 168216718-168216718 on Build GRCh38
rs1051133952 Chr.4: 168216725-168216725 on Build GRCh38
rs891409855 Chr.4: 168216365-168216365 on Build GRCh38
rs1007549564 Chr.4: 168216399-168216399 on Build GRCh38
rs549711813 Chr.4: 168216516-168216516 on Build GRCh38
rs901151718 Chr.4: 168216823-168216823 on Build GRCh38
rs979796991 Chr.4: 168216674-168216674 on Build GRCh38
rs979241258 Chr.4: 168216660-168216660 on Build GRCh38
rs775044813 Chr.4: 168216646-168216646 on Build GRCh38
rs183989326 Chr.4: 168216722-168216722 on Build GRCh38
rs948457028 Chr.4: 168216386-168216386 on Build GRCh38
rs376045750 Chr.4: 168216606-168216606 on Build GRCh38
rs992478656 Chr.4: 168216645-168216645 on Build GRCh38
rs554565775 Chr.4: 168216743-168216743 on Build GRCh38
rs545199670 Chr.4: 168216401-168216401 on Build GRCh38
rs917037386 Chr.4: 168216374-168216374 on Build GRCh38
rs1040375156 Chr.4: 168216772-168216772 on Build GRCh38
rs991146947 Chr.4: 168216582-168216582 on Build GRCh38
rs746292970 Chr.4: 168216587-168216587 on Build GRCh38
rs768013717 Chr.4: 168216818-168216818 on Build GRCh38
rs1023913967 Chr.4: 168216655-168216655 on Build GRCh38
rs1028928454 Chr.4: 168216558-168216558 on Build GRCh38
rs748807705 Chr.4: 168216802-168216802 on Build GRCh38
rs901280838 Chr.4: 168216553-168216553 on Build GRCh38
rs547632307 Chr.4: 168216694-168216694 on Build GRCh38
rs1009604435 Chr.4: 168216473-168216473 on Build GRCh38
rs992447652 Chr.4: 168216359-168216359 on Build GRCh38
rs924050349 Chr.4: 168216693-168216693 on Build GRCh38
rs139771533 Chr.4: 168216435-168216435 on Build GRCh38
rs1055037122 Chr.4: 168216817-168216817 on Build GRCh38
rs10975 Chr.4: 168216436-168216436 on Build GRCh38
rs369983691 Chr.4: 168216561-168216561 on Build GRCh38
rs916467699 Chr.4: 168216605-168216605 on Build GRCh38
rs191861580 Chr.4: 168216621-168216621 on Build GRCh38
rs769565826 Chr.4: 168216641-168216641 on Build GRCh38
rs777928174 Chr.4: 168216403-168216403 on Build GRCh38
rs939389179 Chr.4: 168216767-168216767 on Build GRCh38
rs565866299 Chr.4: 168216713-168216713 on Build GRCh38
rs533423277 Chr.4: 168216342-168216342 on Build GRCh38
rs780212065 Chr.4: 168216801-168216801 on Build GRCh38
rs1035090861 Chr.4: 168216531-168216531 on Build GRCh38
rs531204657 Chr.4: 168216470-168216470 on Build GRCh38
rs373418028 Chr.4: 168216809-168216809 on Build GRCh38
rs952871679 Chr.4: 168216596-168216596 on Build GRCh38
rs925778503 Chr.4: 168216404-168216404 on Build GRCh38

View Full Product Details