Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 173529688-173530182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAND2
Entrez Gene ID
9464
Gene Name
heart and neural crest derivatives expressed 2
Gene Aliases
DHAND2, Hed, Thing2, bHLHa26, dHand
UniGene
Hs.388245
Gene Chromosome Location
Chr.4: 173526501-173530227 on Build GRCh38
Transcripts
NM_021973.2 NR_136197.1

Gene Symbol
HAND2-AS1
Entrez Gene ID
79804
Gene Name
HAND2 antisense RNA 1 (head to head)
Gene Aliases
DEIN, NBLA00301
UniGene
Hs.61435
Gene Chromosome Location
Chr.4: 173528600-173591255 on Build GRCh38
Transcripts
NM_021973.2 NR_136197.1

SNP Detail

SNP ID Chromosome Location
rs939163136 Chr.4: 173529835-173529835 on Build GRCh38
rs1037705790 Chr.4: 173530093-173530093 on Build GRCh38
rs959977689 Chr.4: 173529901-173529901 on Build GRCh38
rs1051601393 Chr.4: 173530019-173530035 on Build GRCh38
rs992727959 Chr.4: 173529993-173529993 on Build GRCh38
rs916281573 Chr.4: 173529850-173529850 on Build GRCh38
rs918727765 Chr.4: 173530143-173530143 on Build GRCh38
rs747827768 Chr.4: 173529999-173529999 on Build GRCh38
rs931476960 Chr.4: 173530153-173530153 on Build GRCh38
rs527432210 Chr.4: 173530096-173530096 on Build GRCh38
rs542922265 Chr.4: 173529984-173529984 on Build GRCh38
rs1054883865 Chr.4: 173529846-173529846 on Build GRCh38
rs1010599457 Chr.4: 173529902-173529902 on Build GRCh38
rs1050399263 Chr.4: 173529756-173529756 on Build GRCh38
rs927812073 Chr.4: 173529814-173529814 on Build GRCh38
rs1041559266 Chr.4: 173530019-173530019 on Build GRCh38
rs968540757 Chr.4: 173529981-173529981 on Build GRCh38
rs958349947 Chr.4: 173529990-173529990 on Build GRCh38
rs904597607 Chr.4: 173529781-173529781 on Build GRCh38
rs917161634 Chr.4: 173529994-173529994 on Build GRCh38
rs944283011 Chr.4: 173530085-173530085 on Build GRCh38
rs895679429 Chr.4: 173529746-173529746 on Build GRCh38
rs558366301 Chr.4: 173529883-173529883 on Build GRCh38
rs1037188261 Chr.4: 173529803-173529803 on Build GRCh38
rs905829035 Chr.4: 173529873-173529873 on Build GRCh38
rs926973132 Chr.4: 173529967-173529967 on Build GRCh38
rs995565389 Chr.4: 173529853-173529853 on Build GRCh38
rs1009167280 Chr.4: 173529810-173529810 on Build GRCh38
rs926920558 Chr.4: 173529717-173529717 on Build GRCh38
rs1036138596 Chr.4: 173529886-173529886 on Build GRCh38
rs754803301 Chr.4: 173529931-173529931 on Build GRCh38
rs554546012 Chr.4: 173529851-173529851 on Build GRCh38
rs199527389 Chr.4: 173529942-173529942 on Build GRCh38
rs978454578 Chr.4: 173529992-173529992 on Build GRCh38
rs936119873 Chr.4: 173530071-173530072 on Build GRCh38
rs1056946929 Chr.4: 173529733-173529733 on Build GRCh38
rs564963647 Chr.4: 173530082-173530082 on Build GRCh38
rs931664950 Chr.4: 173529731-173529731 on Build GRCh38
rs944321241 Chr.4: 173529785-173529785 on Build GRCh38
rs544909428 Chr.4: 173530079-173530079 on Build GRCh38
rs1008886245 Chr.4: 173530110-173530110 on Build GRCh38
rs573811608 Chr.4: 173529947-173529947 on Build GRCh38
rs1028997923 Chr.4: 173529786-173529786 on Build GRCh38
rs781028561 Chr.4: 173529934-173529934 on Build GRCh38
rs921696206 Chr.4: 173529998-173529998 on Build GRCh38
rs1001544287 Chr.4: 173529885-173529885 on Build GRCh38
rs576165683 Chr.4: 173530044-173530044 on Build GRCh38
rs534693591 Chr.4: 173529805-173529805 on Build GRCh38
rs747924752 Chr.4: 173529966-173529966 on Build GRCh38
rs889150710 Chr.4: 173530104-173530104 on Build GRCh38
rs190755061 Chr.4: 173529986-173529986 on Build GRCh38
rs62341031 Chr.4: 173529865-173529865 on Build GRCh38
rs1040691453 Chr.4: 173530152-173530152 on Build GRCh38
rs912819269 Chr.4: 173529762-173529762 on Build GRCh38
rs1048523155 Chr.4: 173529857-173529857 on Build GRCh38

View Full Product Details