Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174658940-174659436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA3
Entrez Gene ID
8001
Gene Name
glycine receptor alpha 3
Gene Aliases
-
UniGene
Hs.413099
Gene Chromosome Location
Chr.4: 174642011-174829568 on Build GRCh38
Transcripts
NM_001042543.2 NM_006529.3 XM_011532268.2 XM_017008630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123483 c.942A>C p.K314N Chr.4: 174659183-174659183 on Build GRCh38
4609054 c.1003G>T p.A335S Chr.4: 174659122-174659122 on Build GRCh38
7171641 c.997G>A p.E333K Chr.4: 174659128-174659128 on Build GRCh38
350988 c.949G>C p.D317H Chr.4: 174659176-174659176 on Build GRCh38
6671733 c.1020A>G p.V340V Chr.4: 174659105-174659105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868737753 Chr.4: 174659094-174659094 on Build GRCh38
rs560093755 Chr.4: 174659191-174659191 on Build GRCh38
rs1019096579 Chr.4: 174658995-174658995 on Build GRCh38
rs903253340 Chr.4: 174659268-174659268 on Build GRCh38
rs201919777 Chr.4: 174659032-174659032 on Build GRCh38
rs759857592 Chr.4: 174659186-174659186 on Build GRCh38
rs780128023 Chr.4: 174659347-174659350 on Build GRCh38
rs992405387 Chr.4: 174659377-174659377 on Build GRCh38
rs770557670 Chr.4: 174659093-174659093 on Build GRCh38
rs748608600 Chr.4: 174659020-174659020 on Build GRCh38
rs1017304763 Chr.4: 174659301-174659302 on Build GRCh38
rs779878360 Chr.4: 174659225-174659225 on Build GRCh38
rs774601822 Chr.4: 174659245-174659245 on Build GRCh38
rs111484273 Chr.4: 174659395-174659395 on Build GRCh38
rs950406456 Chr.4: 174659384-174659386 on Build GRCh38
rs369790717 Chr.4: 174659007-174659007 on Build GRCh38
rs965687656 Chr.4: 174659308-174659308 on Build GRCh38
rs371578056 Chr.4: 174659008-174659008 on Build GRCh38
rs573685163 Chr.4: 174659264-174659264 on Build GRCh38
rs566685256 Chr.4: 174659301-174659301 on Build GRCh38
rs769288436 Chr.4: 174659026-174659026 on Build GRCh38
rs751995690 Chr.4: 174659004-174659004 on Build GRCh38
rs867171370 Chr.4: 174659133-174659133 on Build GRCh38
rs896561488 Chr.4: 174659098-174659098 on Build GRCh38
rs937937888 Chr.4: 174659233-174659233 on Build GRCh38
rs565071741 Chr.4: 174659345-174659345 on Build GRCh38
rs748414872 Chr.4: 174659101-174659102 on Build GRCh38
rs775181879 Chr.4: 174659131-174659131 on Build GRCh38
rs200427979 Chr.4: 174659144-174659144 on Build GRCh38
rs758671619 Chr.4: 174659070-174659070 on Build GRCh38
rs372935265 Chr.4: 174659032-174659032 on Build GRCh38
rs760554975 Chr.4: 174659056-174659056 on Build GRCh38
rs778405533 Chr.4: 174659221-174659221 on Build GRCh38
rs972135436 Chr.4: 174659021-174659021 on Build GRCh38
rs774106199 Chr.4: 174659039-174659039 on Build GRCh38
rs773991227 Chr.4: 174659230-174659231 on Build GRCh38
rs749174715 Chr.4: 174659223-174659224 on Build GRCh38
rs771088809 Chr.4: 174659238-174659238 on Build GRCh38
rs753341272 Chr.4: 174659212-174659212 on Build GRCh38
rs750510260 Chr.4: 174659065-174659065 on Build GRCh38
rs937815101 Chr.4: 174659063-174659063 on Build GRCh38
rs748841046 Chr.4: 174659086-174659086 on Build GRCh38
rs745484429 Chr.4: 174659223-174659223 on Build GRCh38
rs954186944 Chr.4: 174658972-174658972 on Build GRCh38
rs893413315 Chr.4: 174659287-174659287 on Build GRCh38
rs915896237 Chr.4: 174659239-174659239 on Build GRCh38
rs539891118 Chr.4: 174659111-174659111 on Build GRCh38
rs781425532 Chr.4: 174659074-174659074 on Build GRCh38
rs191819635 Chr.4: 174659286-174659286 on Build GRCh38
rs547163003 Chr.4: 174659060-174659060 on Build GRCh38
rs778055117 Chr.4: 174659221-174659221 on Build GRCh38
rs768638635 Chr.4: 174659231-174659231 on Build GRCh38
rs775429142 Chr.4: 174659047-174659047 on Build GRCh38
rs149371787 Chr.4: 174659290-174659290 on Build GRCh38
rs370497572 Chr.4: 174659033-174659033 on Build GRCh38
rs748637883 Chr.4: 174659077-174659077 on Build GRCh38
rs771794578 Chr.4: 174659116-174659116 on Build GRCh38
rs774055425 Chr.4: 174659103-174659103 on Build GRCh38
rs756687547 Chr.4: 174659213-174659213 on Build GRCh38
rs1018735130 Chr.4: 174659241-174659241 on Build GRCh38
rs985865640 Chr.4: 174658994-174658994 on Build GRCh38
rs781756935 Chr.4: 174659017-174659017 on Build GRCh38
rs4365704 Chr.4: 174659040-174659040 on Build GRCh38
rs945442193 Chr.4: 174659107-174659107 on Build GRCh38
rs144798393 Chr.4: 174659389-174659389 on Build GRCh38
rs147848250 Chr.4: 174659192-174659192 on Build GRCh38
rs546808702 Chr.4: 174659073-174659073 on Build GRCh38
rs561400457 Chr.4: 174659027-174659027 on Build GRCh38
rs746838333 Chr.4: 174659228-174659228 on Build GRCh38
rs756607195 Chr.4: 174659084-174659084 on Build GRCh38
rs75754580 Chr.4: 174659324-174659324 on Build GRCh38
rs777302621 Chr.4: 174659085-174659085 on Build GRCh38
rs373674993 Chr.4: 174659031-174659031 on Build GRCh38
rs6553802 Chr.4: 174658968-174658968 on Build GRCh38
rs773329348 Chr.4: 174659157-174659157 on Build GRCh38
rs542301650 Chr.4: 174659284-174659284 on Build GRCh38
rs556030557 Chr.4: 174658967-174658967 on Build GRCh38
rs144463868 Chr.4: 174659179-174659179 on Build GRCh38
rs751971798 Chr.4: 174659072-174659072 on Build GRCh38
rs1049948729 Chr.4: 174659219-174659219 on Build GRCh38
rs552241327 Chr.4: 174659346-174659349 on Build GRCh38
rs766770260 Chr.4: 174659071-174659071 on Build GRCh38
rs1026086013 Chr.4: 174659328-174659329 on Build GRCh38
rs761859085 Chr.4: 174659237-174659237 on Build GRCh38
rs1045808494 Chr.4: 174659268-174659268 on Build GRCh38
rs751776274 Chr.4: 174659182-174659182 on Build GRCh38
rs532125507 Chr.4: 174659027-174659032 on Build GRCh38
rs917725107 Chr.4: 174659379-174659379 on Build GRCh38
rs372446190 Chr.4: 174659188-174659188 on Build GRCh38
rs756199121 Chr.4: 174659204-174659205 on Build GRCh38
rs767435123 Chr.4: 174659041-174659041 on Build GRCh38
rs776647849 Chr.4: 174659234-174659234 on Build GRCh38
rs778603395 Chr.4: 174659411-174659411 on Build GRCh38
rs763195630 Chr.4: 174659176-174659176 on Build GRCh38
rs752945115 Chr.4: 174659224-174659224 on Build GRCh38
rs926520600 Chr.4: 174659339-174659339 on Build GRCh38
rs770937308 Chr.4: 174659227-174659228 on Build GRCh38
rs951676825 Chr.4: 174659323-174659323 on Build GRCh38
rs760631561 Chr.4: 174659136-174659136 on Build GRCh38

View Full Product Details