Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176330149-176330691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPCS3
Entrez Gene ID
60559
Gene Name
signal peptidase complex subunit 3
Gene Aliases
PRO3567, SPC22/23, SPC3, YLR066W
UniGene
Hs.42194
Gene Chromosome Location
Chr.4: 176319939-176332245 on Build GRCh38
Transcripts
NM_021928.3

SNP Detail

SNP ID Chromosome Location
rs961499914 Chr.4: 176330365-176330365 on Build GRCh38
rs919762948 Chr.4: 176330385-176330385 on Build GRCh38
rs1044048590 Chr.4: 176330178-176330178 on Build GRCh38
rs1042493384 Chr.4: 176330549-176330549 on Build GRCh38
rs889762941 Chr.4: 176330601-176330601 on Build GRCh38
rs750452956 Chr.4: 176330300-176330300 on Build GRCh38
rs1036258114 Chr.4: 176330186-176330186 on Build GRCh38
rs930574001 Chr.4: 176330202-176330202 on Build GRCh38
rs757429158 Chr.4: 176330341-176330341 on Build GRCh38
rs868679797 Chr.4: 176330221-176330221 on Build GRCh38
rs182097521 Chr.4: 176330475-176330475 on Build GRCh38
rs549719182 Chr.4: 176330605-176330605 on Build GRCh38
rs539021038 Chr.4: 176330526-176330526 on Build GRCh38
rs896634360 Chr.4: 176330265-176330265 on Build GRCh38
rs966168300 Chr.4: 176330664-176330664 on Build GRCh38
rs972127106 Chr.4: 176330378-176330378 on Build GRCh38
rs754890543 Chr.4: 176330465-176330465 on Build GRCh38
rs1009995455 Chr.4: 176330529-176330529 on Build GRCh38
rs761150316 Chr.4: 176330236-176330236 on Build GRCh38
rs954076554 Chr.4: 176330344-176330344 on Build GRCh38
rs1001037646 Chr.4: 176330594-176330594 on Build GRCh38
rs35030858 Chr.4: 176330177-176330177 on Build GRCh38
rs1001063750 Chr.4: 176330583-176330583 on Build GRCh38
rs187142786 Chr.4: 176330660-176330660 on Build GRCh38
rs764103473 Chr.4: 176330252-176330252 on Build GRCh38
rs903904123 Chr.4: 176330594-176330600 on Build GRCh38
rs984740507 Chr.4: 176330459-176330459 on Build GRCh38
rs1001073879 Chr.4: 176330340-176330340 on Build GRCh38
rs968260559 Chr.4: 176330301-176330301 on Build GRCh38
rs748116242 Chr.4: 176330479-176330479 on Build GRCh38
rs571868647 Chr.4: 176330412-176330412 on Build GRCh38
rs1019621594 Chr.4: 176330627-176330627 on Build GRCh38
rs1021180834 Chr.4: 176330297-176330297 on Build GRCh38
rs865850503 Chr.4: 176330315-176330315 on Build GRCh38
rs919186946 Chr.4: 176330187-176330187 on Build GRCh38
rs557614198 Chr.4: 176330397-176330397 on Build GRCh38
rs146606212 Chr.4: 176330233-176330233 on Build GRCh38
rs143387131 Chr.4: 176330370-176330370 on Build GRCh38
rs968237196 Chr.4: 176330319-176330319 on Build GRCh38
rs1008619112 Chr.4: 176330619-176330619 on Build GRCh38
rs559122526 Chr.4: 176330606-176330606 on Build GRCh38
rs750663190 Chr.4: 176330454-176330454 on Build GRCh38
rs912658766 Chr.4: 176330358-176330358 on Build GRCh38
rs937845284 Chr.4: 176330466-176330466 on Build GRCh38
rs559000193 Chr.4: 176330349-176330349 on Build GRCh38
rs533103585 Chr.4: 176330205-176330205 on Build GRCh38
rs753915625 Chr.4: 176330257-176330257 on Build GRCh38
rs111465107 Chr.4: 176330381-176330381 on Build GRCh38
rs554711797 Chr.4: 176330661-176330661 on Build GRCh38
rs73871985 Chr.4: 176330354-176330354 on Build GRCh38
rs972883330 Chr.4: 176330651-176330651 on Build GRCh38
rs926934033 Chr.4: 176330436-176330436 on Build GRCh38
rs961723370 Chr.4: 176330630-176330630 on Build GRCh38
rs758469765 Chr.4: 176330561-176330561 on Build GRCh38
rs1057313845 Chr.4: 176330251-176330251 on Build GRCh38
rs1010000788 Chr.4: 176330270-176330270 on Build GRCh38
rs532100270 Chr.4: 176330188-176330188 on Build GRCh38
rs1008823373 Chr.4: 176330618-176330618 on Build GRCh38
rs781107746 Chr.4: 176330236-176330237 on Build GRCh38
rs547958896 Chr.4: 176330369-176330369 on Build GRCh38
rs148201775 Chr.4: 176330659-176330659 on Build GRCh38
rs896448180 Chr.4: 176330263-176330263 on Build GRCh38
rs780597373 Chr.4: 176330614-176330614 on Build GRCh38
rs781109055 Chr.4: 176330467-176330467 on Build GRCh38
rs7688500 Chr.4: 176330393-176330393 on Build GRCh38
rs1056818769 Chr.4: 176330457-176330457 on Build GRCh38
rs1028560406 Chr.4: 176330586-176330586 on Build GRCh38
rs1010126274 Chr.4: 176330279-176330279 on Build GRCh38
rs891510085 Chr.4: 176330483-176330483 on Build GRCh38

View Full Product Details