Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183318913-183319422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN22
Entrez Gene ID
53842
Gene Name
claudin 22
Gene Aliases
CLDN21
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183318067-183320774 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183098677-183320777 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5

SNP Detail

SNP ID Chromosome Location
rs1049093228 Chr.4: 183319069-183319069 on Build GRCh38
rs1001042453 Chr.4: 183319114-183319114 on Build GRCh38
rs192797190 Chr.4: 183319236-183319236 on Build GRCh38
rs4862156 Chr.4: 183319210-183319210 on Build GRCh38
rs915805926 Chr.4: 183318951-183318951 on Build GRCh38
rs878855498 Chr.4: 183319314-183319314 on Build GRCh38
rs766693039 Chr.4: 183319105-183319105 on Build GRCh38
rs971701779 Chr.4: 183318958-183318958 on Build GRCh38
rs905134022 Chr.4: 183319003-183319003 on Build GRCh38
rs187009320 Chr.4: 183319195-183319195 on Build GRCh38
rs901875164 Chr.4: 183319093-183319093 on Build GRCh38
rs1046258074 Chr.4: 183318944-183318944 on Build GRCh38
rs190344127 Chr.4: 183318960-183318960 on Build GRCh38
rs937930518 Chr.4: 183319392-183319392 on Build GRCh38
rs919182407 Chr.4: 183319279-183319279 on Build GRCh38
rs537913375 Chr.4: 183319072-183319072 on Build GRCh38
rs532557060 Chr.4: 183319051-183319051 on Build GRCh38
rs1006298490 Chr.4: 183319067-183319067 on Build GRCh38
rs557294662 Chr.4: 183319165-183319165 on Build GRCh38
rs1038994855 Chr.4: 183319092-183319092 on Build GRCh38
rs1046203986 Chr.4: 183319320-183319320 on Build GRCh38
rs919076678 Chr.4: 183318992-183318992 on Build GRCh38
rs556367577 Chr.4: 183319384-183319384 on Build GRCh38
rs953596855 Chr.4: 183319154-183319154 on Build GRCh38
rs1030558646 Chr.4: 183319138-183319138 on Build GRCh38
rs527609785 Chr.4: 183319289-183319289 on Build GRCh38
rs552493930 Chr.4: 183319297-183319297 on Build GRCh38
rs750925890 Chr.4: 183319048-183319048 on Build GRCh38
rs538043173 Chr.4: 183319337-183319337 on Build GRCh38
rs887817513 Chr.4: 183319067-183319069 on Build GRCh38
rs543185073 Chr.4: 183319267-183319267 on Build GRCh38
rs1017084771 Chr.4: 183319032-183319040 on Build GRCh38
rs981410909 Chr.4: 183318975-183318975 on Build GRCh38
rs950569781 Chr.4: 183319305-183319305 on Build GRCh38
rs987873198 Chr.4: 183319261-183319261 on Build GRCh38
rs943351349 Chr.4: 183319077-183319077 on Build GRCh38
rs182541578 Chr.4: 183319100-183319100 on Build GRCh38
rs943445966 Chr.4: 183319375-183319375 on Build GRCh38
rs762748060 Chr.4: 183319094-183319094 on Build GRCh38
rs751805036 Chr.4: 183319110-183319110 on Build GRCh38
rs913641884 Chr.4: 183319302-183319302 on Build GRCh38
rs971120747 Chr.4: 183319254-183319254 on Build GRCh38
rs781254341 Chr.4: 183319183-183319183 on Build GRCh38
rs570720606 Chr.4: 183319328-183319328 on Build GRCh38
rs755139832 Chr.4: 183319123-183319123 on Build GRCh38
rs909075364 Chr.4: 183319332-183319332 on Build GRCh38
rs566734721 Chr.4: 183319237-183319237 on Build GRCh38
rs749217071 Chr.4: 183319304-183319304 on Build GRCh38

View Full Product Details