Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183678937-183679485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC11
Entrez Gene ID
60684
Gene Name
trafficking protein particle complex 11
Gene Aliases
C4orf41, FOIGR, GRY, LGMD2S
UniGene
Hs.443240
Gene Chromosome Location
Chr.4: 183659267-183713594 on Build GRCh38
Transcripts
NM_021942.5 NM_199053.2 XM_017008537.1 XM_017008538.1 XM_017008539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908454 c.850C>T p.Q284* Chr.4: 183679371-183679371 on Build GRCh38
3760503 c.897C>A p.I299I Chr.4: 183679418-183679418 on Build GRCh38
297555 c.846T>C p.C282C Chr.4: 183679367-183679367 on Build GRCh38
3760504 c.927A>G p.A309A Chr.4: 183679448-183679448 on Build GRCh38
1198856 c.915G>T p.K305N Chr.4: 183679436-183679436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568608770 Chr.4: 183679379-183679379 on Build GRCh38
rs768253464 Chr.4: 183679420-183679420 on Build GRCh38
rs529512303 Chr.4: 183679278-183679278 on Build GRCh38
rs979992626 Chr.4: 183679252-183679252 on Build GRCh38
rs1049816779 Chr.4: 183679209-183679209 on Build GRCh38
rs139419771 Chr.4: 183679376-183679376 on Build GRCh38
rs183478450 Chr.4: 183679206-183679206 on Build GRCh38
rs907759415 Chr.4: 183679165-183679165 on Build GRCh38
rs778314763 Chr.4: 183679383-183679383 on Build GRCh38
rs1040149065 Chr.4: 183679174-183679174 on Build GRCh38
rs932193012 Chr.4: 183679091-183679091 on Build GRCh38
rs771578631 Chr.4: 183679408-183679408 on Build GRCh38
rs1035570999 Chr.4: 183679257-183679257 on Build GRCh38
rs890743174 Chr.4: 183679104-183679104 on Build GRCh38
rs979841800 Chr.4: 183679109-183679109 on Build GRCh38
rs60017398 Chr.4: 183678986-183678986 on Build GRCh38
rs1052446026 Chr.4: 183679097-183679097 on Build GRCh38
rs773173688 Chr.4: 183679437-183679437 on Build GRCh38
rs996003842 Chr.4: 183679208-183679208 on Build GRCh38
rs191687588 Chr.4: 183679167-183679167 on Build GRCh38
rs569030465 Chr.4: 183679018-183679018 on Build GRCh38
rs970291569 Chr.4: 183679234-183679234 on Build GRCh38
rs113457212 Chr.4: 183679164-183679164 on Build GRCh38
rs746590830 Chr.4: 183679023-183679023 on Build GRCh38
rs563886118 Chr.4: 183679216-183679216 on Build GRCh38
rs374498633 Chr.4: 183679407-183679407 on Build GRCh38
rs755084302 Chr.4: 183679375-183679375 on Build GRCh38
rs763852465 Chr.4: 183679340-183679340 on Build GRCh38
rs959947581 Chr.4: 183679266-183679266 on Build GRCh38
rs776410109 Chr.4: 183679426-183679426 on Build GRCh38
rs765105196 Chr.4: 183679355-183679355 on Build GRCh38
rs941793989 Chr.4: 183679032-183679032 on Build GRCh38
rs62357990 Chr.4: 183679418-183679418 on Build GRCh38
rs367938699 Chr.4: 183679438-183679438 on Build GRCh38
rs749597402 Chr.4: 183679395-183679395 on Build GRCh38
rs371401679 Chr.4: 183679406-183679406 on Build GRCh38
rs889898147 Chr.4: 183679245-183679245 on Build GRCh38
rs1011393120 Chr.4: 183679179-183679179 on Build GRCh38
rs187249092 Chr.4: 183679112-183679112 on Build GRCh38
rs13115321 Chr.4: 183679085-183679085 on Build GRCh38
rs971555083 Chr.4: 183679394-183679394 on Build GRCh38
rs1037845018 Chr.4: 183679078-183679078 on Build GRCh38
rs533435477 Chr.4: 183679124-183679124 on Build GRCh38
rs938318596 Chr.4: 183679012-183679012 on Build GRCh38
rs114748616 Chr.4: 183679344-183679344 on Build GRCh38
rs999451066 Chr.4: 183679110-183679110 on Build GRCh38
rs756356792 Chr.4: 183679382-183679382 on Build GRCh38
rs749837144 Chr.4: 183679317-183679317 on Build GRCh38
rs764354419 Chr.4: 183679462-183679462 on Build GRCh38
rs535328166 Chr.4: 183679449-183679449 on Build GRCh38
rs1023741544 Chr.4: 183679207-183679207 on Build GRCh38
rs200646990 Chr.4: 183679359-183679359 on Build GRCh38
rs772500441 Chr.4: 183679378-183679378 on Build GRCh38
rs932135252 Chr.4: 183679416-183679416 on Build GRCh38
rs745325177 Chr.4: 183679320-183679320 on Build GRCh38
rs761587688 Chr.4: 183679436-183679436 on Build GRCh38
rs771893992 Chr.4: 183678978-183678978 on Build GRCh38
rs771526031 Chr.4: 183679319-183679319 on Build GRCh38
rs771678650 Chr.4: 183679325-183679325 on Build GRCh38
rs768223521 Chr.4: 183679069-183679069 on Build GRCh38
rs34767735 Chr.4: 183679409-183679409 on Build GRCh38
rs540983493 Chr.4: 183679090-183679090 on Build GRCh38
rs113851701 Chr.4: 183679331-183679331 on Build GRCh38
rs549692677 Chr.4: 183679291-183679291 on Build GRCh38
rs73872659 Chr.4: 183679329-183679329 on Build GRCh38
rs956289312 Chr.4: 183679048-183679048 on Build GRCh38
rs138722849 Chr.4: 183679419-183679419 on Build GRCh38
rs751652729 Chr.4: 183679373-183679373 on Build GRCh38
rs148833310 Chr.4: 183679452-183679452 on Build GRCh38
rs907584219 Chr.4: 183679030-183679030 on Build GRCh38
rs901676544 Chr.4: 183679176-183679176 on Build GRCh38
rs4241779 Chr.4: 183679448-183679448 on Build GRCh38
rs368922437 Chr.4: 183679350-183679350 on Build GRCh38

View Full Product Details