Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185266878-185267386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX25
Entrez Gene ID
83891
Gene Name
sorting nexin 25
Gene Aliases
MSTP043, SBBI31
UniGene
Hs.369091
Gene Chromosome Location
Chr.4: 185204237-185363971 on Build GRCh38
Transcripts
NM_001317781.1 XM_005263267.4 XM_011532302.2 XM_011532303.2 XM_011532304.2 XM_011532305.2 XM_011532306.2 XM_011532307.2 XM_011532308.2 XM_017008680.1 XM_017008681.1 XM_017008682.1 XM_017008683.1 XM_017008684.1 XM_017008686.1 XM_017008687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
323509 c.430G>C p.E144Q Chr.4: 185266986-185266986 on Build GRCh38
1053764 c.520T>A p.Y174N Chr.4: 185267076-185267076 on Build GRCh38
447669 c.574G>A p.D192N Chr.4: 185267130-185267130 on Build GRCh38
1428878 c.525C>A p.T175T Chr.4: 185267081-185267081 on Build GRCh38
6283613 c.599+7G>A p.? Chr.4: 185267162-185267162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185911188 Chr.4: 185267105-185267105 on Build GRCh38
rs749922208 Chr.4: 185267146-185267146 on Build GRCh38
rs376254288 Chr.4: 185267112-185267112 on Build GRCh38
rs905217915 Chr.4: 185267118-185267118 on Build GRCh38
rs763058153 Chr.4: 185266971-185266971 on Build GRCh38
rs1003626043 Chr.4: 185267176-185267176 on Build GRCh38
rs1052638039 Chr.4: 185267225-185267226 on Build GRCh38
rs890668509 Chr.4: 185266935-185266935 on Build GRCh38
rs367660208 Chr.4: 185267174-185267174 on Build GRCh38
rs747490471 Chr.4: 185267205-185267205 on Build GRCh38
rs112410475 Chr.4: 185267115-185267115 on Build GRCh38
rs780866204 Chr.4: 185267182-185267182 on Build GRCh38
rs377693046 Chr.4: 185266957-185266957 on Build GRCh38
rs201057461 Chr.4: 185267010-185267010 on Build GRCh38
rs774951680 Chr.4: 185266979-185266979 on Build GRCh38
rs959961030 Chr.4: 185267337-185267337 on Build GRCh38
rs764660311 Chr.4: 185267140-185267140 on Build GRCh38
rs757953951 Chr.4: 185267168-185267168 on Build GRCh38
rs527841478 Chr.4: 185267054-185267054 on Build GRCh38
rs942144343 Chr.4: 185267110-185267110 on Build GRCh38
rs57930723 Chr.4: 185267233-185267233 on Build GRCh38
rs183326029 Chr.4: 185267307-185267307 on Build GRCh38
rs764396183 Chr.4: 185267108-185267108 on Build GRCh38
rs761067201 Chr.4: 185267106-185267106 on Build GRCh38
rs151109091 Chr.4: 185267120-185267120 on Build GRCh38
rs753097198 Chr.4: 185266994-185266994 on Build GRCh38
rs746798245 Chr.4: 185267055-185267055 on Build GRCh38
rs561963388 Chr.4: 185267039-185267039 on Build GRCh38
rs868184988 Chr.4: 185267123-185267123 on Build GRCh38
rs753470605 Chr.4: 185267027-185267027 on Build GRCh38
rs773357578 Chr.4: 185266965-185266965 on Build GRCh38
rs1051926912 Chr.4: 185266934-185266934 on Build GRCh38
rs146830652 Chr.4: 185266983-185266983 on Build GRCh38
rs745834682 Chr.4: 185266927-185266927 on Build GRCh38
rs190773276 Chr.4: 185267202-185267202 on Build GRCh38
rs756715701 Chr.4: 185267029-185267029 on Build GRCh38
rs760998326 Chr.4: 185267021-185267021 on Build GRCh38
rs745917823 Chr.4: 185267189-185267189 on Build GRCh38
rs569930827 Chr.4: 185267188-185267188 on Build GRCh38
rs754756289 Chr.4: 185267175-185267175 on Build GRCh38
rs776731780 Chr.4: 185266971-185266972 on Build GRCh38
rs200300992 Chr.4: 185267091-185267091 on Build GRCh38
rs920942767 Chr.4: 185267013-185267013 on Build GRCh38
rs924316428 Chr.4: 185267347-185267347 on Build GRCh38
rs774169423 Chr.4: 185267316-185267316 on Build GRCh38
rs775906138 Chr.4: 185267097-185267097 on Build GRCh38
rs766310152 Chr.4: 185267170-185267170 on Build GRCh38
rs1051329287 Chr.4: 185267047-185267047 on Build GRCh38
rs774581601 Chr.4: 185267073-185267073 on Build GRCh38
rs920910219 Chr.4: 185267261-185267261 on Build GRCh38
rs749458238 Chr.4: 185267184-185267184 on Build GRCh38
rs3112865 Chr.4: 185267306-185267306 on Build GRCh38
rs1035482980 Chr.4: 185267090-185267090 on Build GRCh38
rs778017947 Chr.4: 185267066-185267066 on Build GRCh38
rs779914033 Chr.4: 185267046-185267046 on Build GRCh38
rs143685015 Chr.4: 185266978-185266978 on Build GRCh38
rs142932010 Chr.4: 185267060-185267060 on Build GRCh38
rs145186704 Chr.4: 185266907-185266907 on Build GRCh38
rs374744866 Chr.4: 185267198-185267198 on Build GRCh38
rs1035597519 Chr.4: 185267334-185267335 on Build GRCh38
rs371336554 Chr.4: 185267038-185267038 on Build GRCh38
rs770044011 Chr.4: 185266959-185266959 on Build GRCh38
rs749498377 Chr.4: 185267069-185267069 on Build GRCh38
rs570857535 Chr.4: 185267078-185267078 on Build GRCh38
rs1011335468 Chr.4: 185267292-185267292 on Build GRCh38
rs77905550 Chr.4: 185267251-185267251 on Build GRCh38
rs771078798 Chr.4: 185267071-185267071 on Build GRCh38
rs147644007 Chr.4: 185267185-185267185 on Build GRCh38
rs374149781 Chr.4: 185266949-185266949 on Build GRCh38
rs890622144 Chr.4: 185267048-185267048 on Build GRCh38
rs756048042 Chr.4: 185267220-185267220 on Build GRCh38
rs771804831 Chr.4: 185266940-185266940 on Build GRCh38
rs1016741424 Chr.4: 185267203-185267203 on Build GRCh38
rs202195524 Chr.4: 185266980-185266980 on Build GRCh38
rs768763998 Chr.4: 185266959-185266962 on Build GRCh38
rs761573277 Chr.4: 185267330-185267330 on Build GRCh38
rs373840026 Chr.4: 185266982-185266982 on Build GRCh38
rs74834700 Chr.4: 185267232-185267233 on Build GRCh38
rs772689880 Chr.4: 185267096-185267096 on Build GRCh38
rs761468524 Chr.4: 185267131-185267131 on Build GRCh38

View Full Product Details