Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186533974-186534485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTNR1A
Entrez Gene ID
4543
Gene Name
melatonin receptor 1A
Gene Aliases
MEL-1A-R, MT1
UniGene
Hs.243467
Gene Chromosome Location
Chr.4: 186532994-186555567 on Build GRCh38
Transcripts
NM_005958.4 XM_011532002.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671468 c.608T>C p.V203A Chr.4: 186534134-186534134 on Build GRCh38
6166863 c.514C>G p.P172A Chr.4: 186534228-186534228 on Build GRCh38
3602635 c.431C>T p.S144F Chr.4: 186534311-186534311 on Build GRCh38
3602633 c.472G>A p.A158T Chr.4: 186534270-186534270 on Build GRCh38
5400963 c.768C>T p.F256F Chr.4: 186533974-186533974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7654853 Chr.4: 186534107-186534107 on Build GRCh38
rs777236387 Chr.4: 186534172-186534172 on Build GRCh38
rs776459084 Chr.4: 186534267-186534267 on Build GRCh38
rs112305851 Chr.4: 186534121-186534121 on Build GRCh38
rs766690045 Chr.4: 186534337-186534337 on Build GRCh38
rs775185938 Chr.4: 186534085-186534085 on Build GRCh38
rs763220216 Chr.4: 186534204-186534204 on Build GRCh38
rs138789502 Chr.4: 186534268-186534268 on Build GRCh38
rs990314428 Chr.4: 186534005-186534005 on Build GRCh38
rs200493837 Chr.4: 186534130-186534130 on Build GRCh38
rs750427308 Chr.4: 186533997-186533997 on Build GRCh38
rs762288381 Chr.4: 186534300-186534300 on Build GRCh38
rs8192549 Chr.4: 186534175-186534175 on Build GRCh38
rs757006715 Chr.4: 186534251-186534251 on Build GRCh38
rs148108300 Chr.4: 186534384-186534384 on Build GRCh38
rs762715969 Chr.4: 186534395-186534395 on Build GRCh38
rs1021054387 Chr.4: 186534369-186534369 on Build GRCh38
rs779237267 Chr.4: 186534055-186534056 on Build GRCh38
rs769374717 Chr.4: 186534056-186534056 on Build GRCh38
rs140524704 Chr.4: 186534350-186534350 on Build GRCh38
rs768872673 Chr.4: 186534454-186534454 on Build GRCh38
rs368437747 Chr.4: 186534405-186534405 on Build GRCh38
rs758565411 Chr.4: 186534384-186534385 on Build GRCh38
rs751040971 Chr.4: 186534406-186534406 on Build GRCh38
rs371828727 Chr.4: 186534331-186534331 on Build GRCh38
rs373893049 Chr.4: 186534414-186534414 on Build GRCh38
rs757400733 Chr.4: 186534000-186534001 on Build GRCh38
rs149110401 Chr.4: 186534261-186534261 on Build GRCh38
rs779787234 Chr.4: 186534258-186534258 on Build GRCh38
rs757937351 Chr.4: 186534256-186534256 on Build GRCh38
rs576228243 Chr.4: 186534080-186534080 on Build GRCh38
rs772292314 Chr.4: 186534443-186534443 on Build GRCh38
rs140300261 Chr.4: 186534016-186534016 on Build GRCh38
rs747815814 Chr.4: 186534047-186534047 on Build GRCh38
rs891240633 Chr.4: 186534460-186534460 on Build GRCh38
rs769593513 Chr.4: 186534344-186534344 on Build GRCh38
rs773150171 Chr.4: 186534232-186534232 on Build GRCh38
rs374634426 Chr.4: 186534271-186534271 on Build GRCh38
rs750086653 Chr.4: 186534388-186534388 on Build GRCh38
rs770581983 Chr.4: 186534069-186534069 on Build GRCh38
rs374232438 Chr.4: 186534133-186534133 on Build GRCh38
rs1036758640 Chr.4: 186534243-186534244 on Build GRCh38
rs766425059 Chr.4: 186534214-186534214 on Build GRCh38
rs966053745 Chr.4: 186534043-186534043 on Build GRCh38
rs755902349 Chr.4: 186534421-186534421 on Build GRCh38
rs754471002 Chr.4: 186534411-186534411 on Build GRCh38
rs370719848 Chr.4: 186534100-186534100 on Build GRCh38
rs371001585 Chr.4: 186534255-186534255 on Build GRCh38
rs772036163 Chr.4: 186534359-186534359 on Build GRCh38
rs267600114 Chr.4: 186534147-186534147 on Build GRCh38
rs201696287 Chr.4: 186534236-186534236 on Build GRCh38
rs149520289 Chr.4: 186534325-186534325 on Build GRCh38
rs781596329 Chr.4: 186534044-186534044 on Build GRCh38
rs533724160 Chr.4: 186534336-186534336 on Build GRCh38
rs371929194 Chr.4: 186534226-186534226 on Build GRCh38
rs561679720 Chr.4: 186534102-186534102 on Build GRCh38
rs745766636 Chr.4: 186534181-186534181 on Build GRCh38
rs201798233 Chr.4: 186534150-186534150 on Build GRCh38
rs765762273 Chr.4: 186534385-186534385 on Build GRCh38
rs750670225 Chr.4: 186534115-186534115 on Build GRCh38
rs866093242 Chr.4: 186534055-186534055 on Build GRCh38
rs752967556 Chr.4: 186534160-186534160 on Build GRCh38
rs980454535 Chr.4: 186534051-186534051 on Build GRCh38
rs367557623 Chr.4: 186534292-186534292 on Build GRCh38
rs767642752 Chr.4: 186534277-186534277 on Build GRCh38
rs746059681 Chr.4: 186534442-186534442 on Build GRCh38
rs140222606 Chr.4: 186534171-186534171 on Build GRCh38
rs113102490 Chr.4: 186534283-186534283 on Build GRCh38
rs146472456 Chr.4: 186534301-186534301 on Build GRCh38
rs748958201 Chr.4: 186534174-186534174 on Build GRCh38
rs769155491 Chr.4: 186534378-186534378 on Build GRCh38
rs773302416 Chr.4: 186534196-186534196 on Build GRCh38
rs371594884 Chr.4: 186534062-186534062 on Build GRCh38
rs746723591 Chr.4: 186534189-186534189 on Build GRCh38
rs755528087 Chr.4: 186534151-186534151 on Build GRCh38
rs774483149 Chr.4: 186534082-186534082 on Build GRCh38
rs1050934349 Chr.4: 186534459-186534459 on Build GRCh38
rs775518993 Chr.4: 186534187-186534187 on Build GRCh38
rs28383653 Chr.4: 186534245-186534245 on Build GRCh38
rs759362137 Chr.4: 186534154-186534154 on Build GRCh38
rs377204813 Chr.4: 186534040-186534040 on Build GRCh38
rs779672200 Chr.4: 186534330-186534330 on Build GRCh38
rs768322406 Chr.4: 186534264-186534264 on Build GRCh38
rs533536924 Chr.4: 186534195-186534195 on Build GRCh38
rs747046359 Chr.4: 186534451-186534451 on Build GRCh38
rs780126144 Chr.4: 186534430-186534430 on Build GRCh38
rs772092531 Chr.4: 186534186-186534186 on Build GRCh38
rs750467563 Chr.4: 186534218-186534219 on Build GRCh38
rs760697035 Chr.4: 186534370-186534370 on Build GRCh38
rs775674720 Chr.4: 186534368-186534368 on Build GRCh38
rs938992473 Chr.4: 186534081-186534081 on Build GRCh38
rs114219953 Chr.4: 186534455-186534455 on Build GRCh38
rs781051514 Chr.4: 186534412-186534412 on Build GRCh38
rs764154841 Chr.4: 186534278-186534278 on Build GRCh38
rs759645807 Chr.4: 186534227-186534227 on Build GRCh38
rs746172727 Chr.4: 186534357-186534357 on Build GRCh38
rs1800884 Chr.4: 186534272-186534272 on Build GRCh38
rs780174305 Chr.4: 186534449-186534449 on Build GRCh38
rs753830313 Chr.4: 186534419-186534419 on Build GRCh38
rs267600115 Chr.4: 186534148-186534148 on Build GRCh38
rs774368784 Chr.4: 186534076-186534076 on Build GRCh38
rs145677888 Chr.4: 186534379-186534379 on Build GRCh38
rs372744863 Chr.4: 186534000-186534000 on Build GRCh38
rs147194584 Chr.4: 186534436-186534436 on Build GRCh38
rs756370086 Chr.4: 186534233-186534233 on Build GRCh38
rs778736102 Chr.4: 186534252-186534252 on Build GRCh38
rs1019516271 Chr.4: 186534279-186534279 on Build GRCh38
rs538090504 Chr.4: 186534289-186534289 on Build GRCh38
rs200759422 Chr.4: 186534235-186534235 on Build GRCh38
rs1018062210 Chr.4: 186534024-186534024 on Build GRCh38
rs752065361 Chr.4: 186534021-186534021 on Build GRCh38
rs979558798 Chr.4: 186534129-186534129 on Build GRCh38
rs150858716 Chr.4: 186534068-186534068 on Build GRCh38
rs568819396 Chr.4: 186534329-186534329 on Build GRCh38
rs754984108 Chr.4: 186534334-186534334 on Build GRCh38

View Full Product Details