Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10441772-10442262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROPN1L
Entrez Gene ID
83853
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
ASP, RSPH11
UniGene
Hs.381089
Gene Chromosome Location
Chr.5: 10441862-10482807 on Build GRCh38
Transcripts
NM_001201466.1 NM_031916.4 XM_006714503.2 XM_006714504.2 XM_017009946.1 XM_017009947.1 NR_102746.1

Gene Symbol
ROPN1L-AS1
Entrez Gene ID
100505845
Gene Name
ROPN1L antisense RNA 1
Gene Aliases
-
UniGene
Hs.599395
Gene Chromosome Location
Chr.5: 10440735-10441851 on Build GRCh38
Transcripts
NM_001201466.1 NM_031916.4 XM_006714503.2 XM_006714504.2 XM_017009946.1 XM_017009947.1 NR_102746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4503440 c.63C>A p.D21E Chr.5: 10442230-10442230 on Build GRCh38
3607454 c.36G>A p.Q12Q Chr.5: 10442203-10442203 on Build GRCh38
4126993 c.51G>A p.P17P Chr.5: 10442218-10442218 on Build GRCh38
6101450 c.47_48CC>TT p.P16L Chr.5: 10442214-10442215 on Build GRCh38
6027057 c.79A>G p.T27A Chr.5: 10442246-10442246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567902794 Chr.5: 10441960-10441960 on Build GRCh38
rs749738041 Chr.5: 10442153-10442153 on Build GRCh38
rs764462958 Chr.5: 10442230-10442230 on Build GRCh38
rs994653603 Chr.5: 10442087-10442087 on Build GRCh38
rs768427973 Chr.5: 10442199-10442199 on Build GRCh38
rs746844249 Chr.5: 10442179-10442179 on Build GRCh38
rs964489014 Chr.5: 10441859-10441859 on Build GRCh38
rs1049955393 Chr.5: 10442040-10442040 on Build GRCh38
rs575028236 Chr.5: 10441825-10441825 on Build GRCh38
rs967755949 Chr.5: 10441802-10441802 on Build GRCh38
rs1011996688 Chr.5: 10441836-10441836 on Build GRCh38
rs546093775 Chr.5: 10441899-10441899 on Build GRCh38
rs772328018 Chr.5: 10442164-10442164 on Build GRCh38
rs934112585 Chr.5: 10441887-10441887 on Build GRCh38
rs896332670 Chr.5: 10441949-10441949 on Build GRCh38
rs754333927 Chr.5: 10442233-10442233 on Build GRCh38
rs745877124 Chr.5: 10442216-10442216 on Build GRCh38
rs756441351 Chr.5: 10442226-10442226 on Build GRCh38
rs776629239 Chr.5: 10442121-10442121 on Build GRCh38
rs760134882 Chr.5: 10442171-10442171 on Build GRCh38
rs752074390 Chr.5: 10441821-10441821 on Build GRCh38
rs566292292 Chr.5: 10442146-10442146 on Build GRCh38
rs775674023 Chr.5: 10442166-10442166 on Build GRCh38
rs1008162347 Chr.5: 10442069-10442069 on Build GRCh38
rs370329506 Chr.5: 10442092-10442092 on Build GRCh38
rs1049987928 Chr.5: 10442220-10442220 on Build GRCh38
rs1020765623 Chr.5: 10441953-10441953 on Build GRCh38
rs893285729 Chr.5: 10442020-10442020 on Build GRCh38
rs772892187 Chr.5: 10442205-10442205 on Build GRCh38
rs1047426 Chr.5: 10441956-10441956 on Build GRCh38
rs1038805413 Chr.5: 10441906-10441906 on Build GRCh38
rs979530412 Chr.5: 10441807-10441807 on Build GRCh38
rs370906334 Chr.5: 10442159-10442159 on Build GRCh38
rs757297462 Chr.5: 10442154-10442154 on Build GRCh38
rs965870462 Chr.5: 10441913-10441913 on Build GRCh38
rs866621700 Chr.5: 10441921-10441921 on Build GRCh38
rs372992553 Chr.5: 10441890-10441890 on Build GRCh38
rs373103334 Chr.5: 10442122-10442122 on Build GRCh38
rs1009328262 Chr.5: 10441952-10441952 on Build GRCh38
rs919176370 Chr.5: 10441942-10441942 on Build GRCh38
rs1056437856 Chr.5: 10441944-10441944 on Build GRCh38
rs778211684 Chr.5: 10442149-10442149 on Build GRCh38
rs143747968 Chr.5: 10442120-10442120 on Build GRCh38
rs922768301 Chr.5: 10441875-10441875 on Build GRCh38
rs531812370 Chr.5: 10441937-10441937 on Build GRCh38
rs753163537 Chr.5: 10442134-10442134 on Build GRCh38
rs922985554 Chr.5: 10442055-10442055 on Build GRCh38
rs779041842 Chr.5: 10442102-10442102 on Build GRCh38
rs1011857022 Chr.5: 10442021-10442021 on Build GRCh38
rs1038617360 Chr.5: 10442082-10442082 on Build GRCh38
rs145970601 Chr.5: 10442219-10442219 on Build GRCh38
rs528290328 Chr.5: 10441917-10441917 on Build GRCh38
rs937981284 Chr.5: 10442094-10442094 on Build GRCh38
rs749361453 Chr.5: 10441892-10441892 on Build GRCh38
rs866529804 Chr.5: 10441908-10441908 on Build GRCh38
rs745916364 Chr.5: 10442161-10442161 on Build GRCh38
rs754917814 Chr.5: 10442132-10442132 on Build GRCh38
rs928373760 Chr.5: 10442152-10442152 on Build GRCh38
rs975826886 Chr.5: 10441866-10441866 on Build GRCh38
rs964333051 Chr.5: 10442023-10442023 on Build GRCh38
rs1053256388 Chr.5: 10442007-10442007 on Build GRCh38
rs184031617 Chr.5: 10441903-10441903 on Build GRCh38
rs905184704 Chr.5: 10441912-10441912 on Build GRCh38
rs374704312 Chr.5: 10442123-10442123 on Build GRCh38
rs563701025 Chr.5: 10441862-10441862 on Build GRCh38
rs1026186879 Chr.5: 10442109-10442109 on Build GRCh38
rs762533473 Chr.5: 10442214-10442214 on Build GRCh38
rs539024871 Chr.5: 10442083-10442083 on Build GRCh38
rs893347308 Chr.5: 10441832-10441832 on Build GRCh38
rs1045520686 Chr.5: 10441973-10441973 on Build GRCh38
rs761766011 Chr.5: 10442200-10442200 on Build GRCh38
rs941642324 Chr.5: 10442080-10442080 on Build GRCh38
rs570130036 Chr.5: 10442041-10442041 on Build GRCh38
rs188816627 Chr.5: 10442031-10442031 on Build GRCh38
rs983380034 Chr.5: 10442068-10442068 on Build GRCh38
rs908696654 Chr.5: 10442071-10442071 on Build GRCh38
rs112014572 Chr.5: 10441994-10441994 on Build GRCh38
rs971884461 Chr.5: 10441932-10441932 on Build GRCh38
rs766101906 Chr.5: 10442133-10442133 on Build GRCh38
rs543918140 Chr.5: 10441849-10441849 on Build GRCh38
rs955542917 Chr.5: 10442060-10442060 on Build GRCh38
rs954029874 Chr.5: 10441878-10441878 on Build GRCh38
rs766194872 Chr.5: 10442217-10442217 on Build GRCh38

View Full Product Details