Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 21750899-21751413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH12
Entrez Gene ID
1010
Gene Name
cadherin 12
Gene Aliases
CDHB
UniGene
Hs.113684
Gene Chromosome Location
Chr.5: 21750191-22853622 on Build GRCh38
Transcripts
NM_001317227.1 NM_001317228.1 NM_004061.4 XM_011513927.2 XM_017008920.1 XM_017008921.1

SNP Detail

SNP ID Chromosome Location
rs530412478 Chr.5: 21751105-21751105 on Build GRCh38
rs533819026 Chr.5: 21751344-21751344 on Build GRCh38
rs183954121 Chr.5: 21751059-21751059 on Build GRCh38
rs936206491 Chr.5: 21751022-21751022 on Build GRCh38
rs962523479 Chr.5: 21751067-21751067 on Build GRCh38
rs1028128522 Chr.5: 21751169-21751169 on Build GRCh38
rs1015294934 Chr.5: 21751332-21751332 on Build GRCh38
rs894341847 Chr.5: 21751273-21751273 on Build GRCh38
rs552500593 Chr.5: 21751213-21751213 on Build GRCh38
rs1015367289 Chr.5: 21751057-21751057 on Build GRCh38
rs945397530 Chr.5: 21751123-21751123 on Build GRCh38
rs1012874845 Chr.5: 21750926-21750926 on Build GRCh38
rs940063068 Chr.5: 21751371-21751371 on Build GRCh38
rs530909247 Chr.5: 21751009-21751009 on Build GRCh38
rs10570044 Chr.5: 21751259-21751260 on Build GRCh38
rs772231516 Chr.5: 21751186-21751188 on Build GRCh38
rs966739193 Chr.5: 21750960-21750960 on Build GRCh38
rs532186587 Chr.5: 21751387-21751387 on Build GRCh38
rs867063987 Chr.5: 21751329-21751329 on Build GRCh38
rs936793230 Chr.5: 21751216-21751216 on Build GRCh38
rs74844861 Chr.5: 21751260-21751260 on Build GRCh38
rs561165189 Chr.5: 21751081-21751081 on Build GRCh38
rs70957058 Chr.5: 21750950-21750951 on Build GRCh38
rs916082427 Chr.5: 21751297-21751297 on Build GRCh38
rs977741585 Chr.5: 21750960-21750960 on Build GRCh38
rs979489184 Chr.5: 21751277-21751277 on Build GRCh38
rs907094371 Chr.5: 21751290-21751290 on Build GRCh38
rs966437858 Chr.5: 21751271-21751271 on Build GRCh38
rs532724993 Chr.5: 21751204-21751204 on Build GRCh38
rs796770777 Chr.5: 21751173-21751173 on Build GRCh38
rs200473163 Chr.5: 21751253-21751253 on Build GRCh38
rs567747702 Chr.5: 21751295-21751295 on Build GRCh38
rs955145056 Chr.5: 21750935-21750935 on Build GRCh38
rs986485511 Chr.5: 21750947-21750947 on Build GRCh38
rs35176274 Chr.5: 21750942-21750943 on Build GRCh38
rs762202205 Chr.5: 21751355-21751355 on Build GRCh38
rs567340200 Chr.5: 21750976-21750976 on Build GRCh38
rs981554850 Chr.5: 21751328-21751328 on Build GRCh38
rs547343995 Chr.5: 21751115-21751115 on Build GRCh38
rs987022132 Chr.5: 21751116-21751116 on Build GRCh38
rs1020585998 Chr.5: 21750952-21750952 on Build GRCh38
rs537585281 Chr.5: 21751287-21751287 on Build GRCh38
rs927410898 Chr.5: 21750963-21750963 on Build GRCh38
rs982403292 Chr.5: 21751006-21751006 on Build GRCh38
rs548167357 Chr.5: 21751288-21751288 on Build GRCh38
rs553613614 Chr.5: 21751357-21751357 on Build GRCh38
rs201384349 Chr.5: 21751262-21751262 on Build GRCh38
rs1024219091 Chr.5: 21750954-21750954 on Build GRCh38
rs188190991 Chr.5: 21751117-21751117 on Build GRCh38
rs898390649 Chr.5: 21751156-21751157 on Build GRCh38
rs1042340426 Chr.5: 21751130-21751130 on Build GRCh38
rs775295594 Chr.5: 21750923-21750923 on Build GRCh38
rs144406736 Chr.5: 21751074-21751074 on Build GRCh38
rs747886374 Chr.5: 21750946-21750947 on Build GRCh38
rs5866521 Chr.5: 21750941-21750942 on Build GRCh38
rs996753828 Chr.5: 21751168-21751168 on Build GRCh38
rs1012719970 Chr.5: 21751281-21751281 on Build GRCh38
rs75187785 Chr.5: 21750951-21750951 on Build GRCh38
rs529740382 Chr.5: 21751010-21751010 on Build GRCh38
rs950193080 Chr.5: 21751300-21751300 on Build GRCh38
rs796850823 Chr.5: 21751260-21751261 on Build GRCh38
rs1038797665 Chr.5: 21751378-21751378 on Build GRCh38
rs774870757 Chr.5: 21751125-21751125 on Build GRCh38
rs111329458 Chr.5: 21750955-21750955 on Build GRCh38

View Full Product Details