Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 24487747-24488259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH10
Entrez Gene ID
1008
Gene Name
cadherin 10
Gene Aliases
-
UniGene
Hs.92489
Gene Chromosome Location
Chr.5: 24487100-24644978 on Build GRCh38
Transcripts
NM_001317222.1 NM_001317224.1 NM_006727.4 XM_011513923.2 XM_017008915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3013915 c.2215G>T p.E739* Chr.5: 24487815-24487815 on Build GRCh38
5975573 c.2179G>T p.A727S Chr.5: 24487851-24487851 on Build GRCh38
70108 c.1982G>A p.G661D Chr.5: 24488048-24488048 on Build GRCh38
6103505 c.1956C>A p.D652E Chr.5: 24488074-24488074 on Build GRCh38
4141763 c.2152C>A p.L718I Chr.5: 24487878-24487878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749221632 Chr.5: 24487816-24487816 on Build GRCh38
rs773208707 Chr.5: 24488194-24488194 on Build GRCh38
rs985566802 Chr.5: 24488071-24488071 on Build GRCh38
rs763423183 Chr.5: 24487862-24487862 on Build GRCh38
rs751881000 Chr.5: 24487879-24487879 on Build GRCh38
rs746847549 Chr.5: 24488079-24488079 on Build GRCh38
rs760760327 Chr.5: 24488008-24488008 on Build GRCh38
rs200154239 Chr.5: 24487986-24487986 on Build GRCh38
rs747555737 Chr.5: 24488092-24488092 on Build GRCh38
rs771711060 Chr.5: 24488147-24488147 on Build GRCh38
rs202220094 Chr.5: 24488103-24488103 on Build GRCh38
rs747903579 Chr.5: 24488189-24488189 on Build GRCh38
rs756185558 Chr.5: 24487917-24487917 on Build GRCh38
rs756200941 Chr.5: 24487805-24487805 on Build GRCh38
rs1036386867 Chr.5: 24487897-24487897 on Build GRCh38
rs757099072 Chr.5: 24487934-24487934 on Build GRCh38
rs767907920 Chr.5: 24487994-24487994 on Build GRCh38
rs201007562 Chr.5: 24488083-24488083 on Build GRCh38
rs752982862 Chr.5: 24487903-24487903 on Build GRCh38
rs763581492 Chr.5: 24487851-24487851 on Build GRCh38
rs985564698 Chr.5: 24487804-24487804 on Build GRCh38
rs761101753 Chr.5: 24488129-24488129 on Build GRCh38
rs771781685 Chr.5: 24487844-24487844 on Build GRCh38
rs747967376 Chr.5: 24488053-24488053 on Build GRCh38
rs373536756 Chr.5: 24488203-24488203 on Build GRCh38
rs760083492 Chr.5: 24488105-24488105 on Build GRCh38
rs141569837 Chr.5: 24488183-24488183 on Build GRCh38
rs780696583 Chr.5: 24488185-24488185 on Build GRCh38
rs543550761 Chr.5: 24487949-24487949 on Build GRCh38
rs982045454 Chr.5: 24488094-24488094 on Build GRCh38
rs927839759 Chr.5: 24488168-24488168 on Build GRCh38
rs771276034 Chr.5: 24487974-24487974 on Build GRCh38
rs779851472 Chr.5: 24487967-24487967 on Build GRCh38
rs775792413 Chr.5: 24488125-24488125 on Build GRCh38
rs759882971 Chr.5: 24487991-24487991 on Build GRCh38
rs765449654 Chr.5: 24488153-24488153 on Build GRCh38
rs748826661 Chr.5: 24488195-24488195 on Build GRCh38
rs200327856 Chr.5: 24488148-24488148 on Build GRCh38
rs963500072 Chr.5: 24487884-24487884 on Build GRCh38
rs780903483 Chr.5: 24488086-24488086 on Build GRCh38
rs201737578 Chr.5: 24488014-24488014 on Build GRCh38
rs769406878 Chr.5: 24488093-24488093 on Build GRCh38
rs746701849 Chr.5: 24487968-24487968 on Build GRCh38
rs141645081 Chr.5: 24488011-24488011 on Build GRCh38
rs946823134 Chr.5: 24488224-24488224 on Build GRCh38
rs201104078 Chr.5: 24487922-24487922 on Build GRCh38
rs777761277 Chr.5: 24487809-24487809 on Build GRCh38
rs781424714 Chr.5: 24487771-24487771 on Build GRCh38
rs775959130 Chr.5: 24487972-24487972 on Build GRCh38
rs146231810 Chr.5: 24488026-24488026 on Build GRCh38
rs542487965 Chr.5: 24487840-24487840 on Build GRCh38
rs1029850703 Chr.5: 24487783-24487783 on Build GRCh38
rs772588728 Chr.5: 24488118-24488118 on Build GRCh38
rs773966996 Chr.5: 24487834-24487834 on Build GRCh38
rs766454620 Chr.5: 24488164-24488164 on Build GRCh38
rs973320967 Chr.5: 24487958-24487958 on Build GRCh38
rs751298070 Chr.5: 24487992-24487993 on Build GRCh38
rs1027042839 Chr.5: 24487969-24487969 on Build GRCh38
rs770736913 Chr.5: 24487820-24487820 on Build GRCh38
rs944055917 Chr.5: 24488130-24488130 on Build GRCh38
rs267600592 Chr.5: 24487860-24487860 on Build GRCh38
rs759899651 Chr.5: 24487880-24487880 on Build GRCh38
rs560951980 Chr.5: 24487902-24487902 on Build GRCh38
rs758238763 Chr.5: 24488030-24488030 on Build GRCh38
rs758365581 Chr.5: 24488155-24488155 on Build GRCh38
rs961564242 Chr.5: 24487918-24487918 on Build GRCh38
rs772047494 Chr.5: 24487962-24487962 on Build GRCh38
rs867119624 Chr.5: 24488157-24488157 on Build GRCh38
rs753973333 Chr.5: 24487924-24487924 on Build GRCh38
rs750366779 Chr.5: 24488154-24488154 on Build GRCh38
rs928939235 Chr.5: 24488013-24488013 on Build GRCh38
rs760251937 Chr.5: 24487846-24487846 on Build GRCh38
rs764161207 Chr.5: 24488132-24488132 on Build GRCh38
rs778995281 Chr.5: 24487948-24487948 on Build GRCh38
rs776967205 Chr.5: 24488135-24488135 on Build GRCh38
rs780047745 Chr.5: 24488034-24488034 on Build GRCh38
rs746736772 Chr.5: 24487876-24487876 on Build GRCh38
rs369660807 Chr.5: 24488177-24488177 on Build GRCh38
rs768194920 Chr.5: 24487971-24487971 on Build GRCh38
rs376656809 Chr.5: 24488172-24488172 on Build GRCh38
rs752893007 Chr.5: 24488005-24488005 on Build GRCh38
rs143036065 Chr.5: 24487856-24487856 on Build GRCh38
rs867597685 Chr.5: 24487891-24487891 on Build GRCh38
rs149466992 Chr.5: 24487800-24487800 on Build GRCh38

View Full Product Details