Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 32711232-32711724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPR3
Entrez Gene ID
4883
Gene Name
natriuretic peptide receptor 3
Gene Aliases
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC
UniGene
Hs.13528
Gene Chromosome Location
Chr.5: 32689076-32791724 on Build GRCh38
Transcripts
NM_000908.3 NM_001204375.1 NM_001204376.1 XM_005248309.1 XM_011514047.1 XM_011514048.2 XM_011514049.2 XM_011514050.2 XM_017009492.1

SNP Detail

SNP ID Chromosome Location
rs887709216 Chr.5: 32711671-32711671 on Build GRCh38
rs897895547 Chr.5: 32711619-32711619 on Build GRCh38
rs368476386 Chr.5: 32711275-32711275 on Build GRCh38
rs990625436 Chr.5: 32711305-32711305 on Build GRCh38
rs1051051890 Chr.5: 32711540-32711540 on Build GRCh38
rs867573249 Chr.5: 32711302-32711302 on Build GRCh38
rs991888406 Chr.5: 32711472-32711472 on Build GRCh38
rs543749560 Chr.5: 32711510-32711510 on Build GRCh38
rs377623742 Chr.5: 32711545-32711545 on Build GRCh38
rs897797815 Chr.5: 32711342-32711342 on Build GRCh38
rs1024802849 Chr.5: 32711566-32711567 on Build GRCh38
rs915979854 Chr.5: 32711311-32711311 on Build GRCh38
rs1049022246 Chr.5: 32711670-32711670 on Build GRCh38
rs1040471351 Chr.5: 32711602-32711602 on Build GRCh38
rs574754280 Chr.5: 32711428-32711428 on Build GRCh38
rs1002459506 Chr.5: 32711393-32711393 on Build GRCh38
rs971445976 Chr.5: 32711580-32711580 on Build GRCh38
rs780845266 Chr.5: 32711315-32711315 on Build GRCh38
rs981846640 Chr.5: 32711321-32711321 on Build GRCh38
rs1042023789 Chr.5: 32711264-32711264 on Build GRCh38
rs1040781419 Chr.5: 32711337-32711337 on Build GRCh38
rs914840618 Chr.5: 32711360-32711360 on Build GRCh38
rs371013970 Chr.5: 32711546-32711546 on Build GRCh38
rs1016151974 Chr.5: 32711514-32711514 on Build GRCh38
rs199815172 Chr.5: 32711389-32711390 on Build GRCh38
rs971475356 Chr.5: 32711314-32711314 on Build GRCh38
rs372556295 Chr.5: 32711381-32711381 on Build GRCh38
rs956038575 Chr.5: 32711623-32711623 on Build GRCh38
rs9716700 Chr.5: 32711527-32711527 on Build GRCh38
rs886067874 Chr.5: 32711501-32711502 on Build GRCh38
rs960737389 Chr.5: 32711453-32711453 on Build GRCh38
rs966407734 Chr.5: 32711500-32711500 on Build GRCh38
rs943505998 Chr.5: 32711366-32711366 on Build GRCh38
rs187841166 Chr.5: 32711423-32711423 on Build GRCh38
rs538733853 Chr.5: 32711252-32711252 on Build GRCh38
rs1054673764 Chr.5: 32711346-32711346 on Build GRCh38
rs565655829 Chr.5: 32711306-32711306 on Build GRCh38
rs981514097 Chr.5: 32711585-32711585 on Build GRCh38
rs1006105208 Chr.5: 32711259-32711259 on Build GRCh38
rs1050606100 Chr.5: 32711291-32711291 on Build GRCh38
rs914809421 Chr.5: 32711651-32711651 on Build GRCh38
rs990585813 Chr.5: 32711642-32711642 on Build GRCh38
rs879019072 Chr.5: 32711567-32711568 on Build GRCh38
rs1006320035 Chr.5: 32711502-32711503 on Build GRCh38
rs1039242471 Chr.5: 32711380-32711380 on Build GRCh38
rs534582853 Chr.5: 32711326-32711326 on Build GRCh38
rs138317732 Chr.5: 32711547-32711548 on Build GRCh38
rs926224578 Chr.5: 32711516-32711516 on Build GRCh38
rs901591690 Chr.5: 32711384-32711384 on Build GRCh38
rs951950802 Chr.5: 32711386-32711386 on Build GRCh38
rs192928389 Chr.5: 32711576-32711576 on Build GRCh38
rs577054152 Chr.5: 32711533-32711533 on Build GRCh38
rs936192376 Chr.5: 32711276-32711276 on Build GRCh38
rs979076034 Chr.5: 32711515-32711515 on Build GRCh38
rs1027551464 Chr.5: 32711356-32711356 on Build GRCh38
rs188884565 Chr.5: 32711572-32711572 on Build GRCh38
rs1002085719 Chr.5: 32711684-32711684 on Build GRCh38
rs929336158 Chr.5: 32711650-32711650 on Build GRCh38
rs1008960196 Chr.5: 32711324-32711324 on Build GRCh38
rs375329153 Chr.5: 32711547-32711547 on Build GRCh38
rs959174444 Chr.5: 32711279-32711279 on Build GRCh38
rs993455844 Chr.5: 32711343-32711343 on Build GRCh38
rs373785425 Chr.5: 32711543-32711543 on Build GRCh38
rs59057102 Chr.5: 32711571-32711572 on Build GRCh38
rs913644338 Chr.5: 32711474-32711474 on Build GRCh38

View Full Product Details