Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33936294-33936788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXFP3
Entrez Gene ID
51289
Gene Name
relaxin/insulin like family peptide receptor 3
Gene Aliases
GPCR135, RLN3R1, RXFPR3, SALPR
UniGene
Hs.170146
Gene Chromosome Location
Chr.5: 33936386-33938918 on Build GRCh38
Transcripts
NM_016568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6546127 c.46G>T p.A16S Chr.5: 33936786-33936786 on Build GRCh38
402590 c.3G>C p.M1I Chr.5: 33936743-33936743 on Build GRCh38
1437090 c.28G>T p.A10S Chr.5: 33936768-33936768 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746493245 Chr.5: 33936731-33936731 on Build GRCh38
rs576452923 Chr.5: 33936730-33936730 on Build GRCh38
rs1022088265 Chr.5: 33936517-33936517 on Build GRCh38
rs915986249 Chr.5: 33936386-33936386 on Build GRCh38
rs1034558166 Chr.5: 33936572-33936572 on Build GRCh38
rs546732529 Chr.5: 33936348-33936348 on Build GRCh38
rs1052771577 Chr.5: 33936507-33936507 on Build GRCh38
rs773760844 Chr.5: 33936735-33936735 on Build GRCh38
rs554570314 Chr.5: 33936402-33936402 on Build GRCh38
rs774576036 Chr.5: 33936748-33936748 on Build GRCh38
rs970090584 Chr.5: 33936683-33936683 on Build GRCh38
rs538709464 Chr.5: 33936441-33936441 on Build GRCh38
rs34504136 Chr.5: 33936460-33936460 on Build GRCh38
rs936458614 Chr.5: 33936624-33936624 on Build GRCh38
rs533035395 Chr.5: 33936707-33936707 on Build GRCh38
rs868694822 Chr.5: 33936751-33936751 on Build GRCh38
rs960315099 Chr.5: 33936332-33936332 on Build GRCh38
rs577223199 Chr.5: 33936580-33936580 on Build GRCh38
rs561701363 Chr.5: 33936622-33936622 on Build GRCh38
rs939165610 Chr.5: 33936711-33936711 on Build GRCh38
rs770258102 Chr.5: 33936734-33936734 on Build GRCh38
rs537415635 Chr.5: 33936498-33936498 on Build GRCh38
rs915997570 Chr.5: 33936657-33936657 on Build GRCh38
rs745462777 Chr.5: 33936714-33936714 on Build GRCh38
rs899550874 Chr.5: 33936526-33936526 on Build GRCh38
rs746756868 Chr.5: 33936415-33936415 on Build GRCh38
rs763230282 Chr.5: 33936743-33936743 on Build GRCh38
rs759713382 Chr.5: 33936749-33936749 on Build GRCh38
rs925053060 Chr.5: 33936359-33936359 on Build GRCh38
rs752664289 Chr.5: 33936690-33936690 on Build GRCh38
rs945047175 Chr.5: 33936354-33936354 on Build GRCh38
rs778438543 Chr.5: 33936713-33936713 on Build GRCh38
rs944008158 Chr.5: 33936420-33936420 on Build GRCh38
rs751809552 Chr.5: 33936669-33936669 on Build GRCh38
rs546091513 Chr.5: 33936588-33936588 on Build GRCh38
rs978185279 Chr.5: 33936610-33936610 on Build GRCh38
rs113835504 Chr.5: 33936706-33936706 on Build GRCh38
rs376505780 Chr.5: 33936747-33936747 on Build GRCh38
rs756087176 Chr.5: 33936696-33936696 on Build GRCh38
rs553597788 Chr.5: 33936593-33936593 on Build GRCh38
rs983200604 Chr.5: 33936329-33936329 on Build GRCh38
rs768590526 Chr.5: 33936494-33936494 on Build GRCh38
rs977841175 Chr.5: 33936356-33936356 on Build GRCh38
rs544148340 Chr.5: 33936651-33936651 on Build GRCh38
rs75108914 Chr.5: 33936598-33936598 on Build GRCh38
rs1010987254 Chr.5: 33936516-33936516 on Build GRCh38
rs79014396 Chr.5: 33936616-33936616 on Build GRCh38
rs897119412 Chr.5: 33936452-33936452 on Build GRCh38
rs181254835 Chr.5: 33936660-33936660 on Build GRCh38
rs186391670 Chr.5: 33936324-33936324 on Build GRCh38
rs980583712 Chr.5: 33936684-33936684 on Build GRCh38
rs116094831 Chr.5: 33936470-33936470 on Build GRCh38
rs777745057 Chr.5: 33936702-33936702 on Build GRCh38
rs535356377 Chr.5: 33936384-33936384 on Build GRCh38
rs892155863 Chr.5: 33936513-33936513 on Build GRCh38
rs767696837 Chr.5: 33936764-33936764 on Build GRCh38
rs997020801 Chr.5: 33936540-33936540 on Build GRCh38
rs780077691 Chr.5: 33936368-33936368 on Build GRCh38
rs771732729 Chr.5: 33936716-33936716 on Build GRCh38
rs564264208 Chr.5: 33936544-33936544 on Build GRCh38
rs532985140 Chr.5: 33936331-33936331 on Build GRCh38
rs189675844 Chr.5: 33936380-33936380 on Build GRCh38
rs1041360798 Chr.5: 33936450-33936450 on Build GRCh38
rs917553827 Chr.5: 33936349-33936349 on Build GRCh38
rs78479587 Chr.5: 33936633-33936633 on Build GRCh38
rs985257408 Chr.5: 33936627-33936627 on Build GRCh38

View Full Product Details