Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33945504-33946004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A2
Entrez Gene ID
51151
Gene Name
solute carrier family 45 member 2
Gene Aliases
1A1, AIM1, MATP, OCA4, SHEP5
UniGene
Hs.278962
Gene Chromosome Location
Chr.5: 33944616-33984675 on Build GRCh38
Transcripts
NM_001012509.3 NM_016180.4

SNP Detail

SNP ID Chromosome Location
rs1354127 Chr.5: 33945696-33945696 on Build GRCh38
rs938349742 Chr.5: 33945897-33945897 on Build GRCh38
rs146922924 Chr.5: 33945797-33945798 on Build GRCh38
rs887763023 Chr.5: 33945726-33945726 on Build GRCh38
rs1055212361 Chr.5: 33945754-33945754 on Build GRCh38
rs894305282 Chr.5: 33945580-33945580 on Build GRCh38
rs781030499 Chr.5: 33945728-33945728 on Build GRCh38
rs370957300 Chr.5: 33945595-33945596 on Build GRCh38
rs976313843 Chr.5: 33945672-33945672 on Build GRCh38
rs879768345 Chr.5: 33945696-33945697 on Build GRCh38
rs968627769 Chr.5: 33945531-33945531 on Build GRCh38
rs1024189468 Chr.5: 33945593-33945593 on Build GRCh38
rs556519292 Chr.5: 33945610-33945610 on Build GRCh38
rs977084992 Chr.5: 33945626-33945626 on Build GRCh38
rs1045278692 Chr.5: 33945893-33945893 on Build GRCh38
rs987502770 Chr.5: 33945789-33945789 on Build GRCh38
rs891584749 Chr.5: 33945680-33945680 on Build GRCh38
rs187081018 Chr.5: 33945823-33945823 on Build GRCh38
rs558594148 Chr.5: 33945655-33945655 on Build GRCh38
rs34262723 Chr.5: 33945695-33945695 on Build GRCh38
rs940846134 Chr.5: 33945614-33945614 on Build GRCh38
rs1009957538 Chr.5: 33945911-33945911 on Build GRCh38
rs907980087 Chr.5: 33945613-33945613 on Build GRCh38
rs386687047 Chr.5: 33945696-33945697 on Build GRCh38
rs62350869 Chr.5: 33945540-33945540 on Build GRCh38
rs996068978 Chr.5: 33945705-33945705 on Build GRCh38
rs397947003 Chr.5: 33945677-33945678 on Build GRCh38
rs572329672 Chr.5: 33945656-33945656 on Build GRCh38
rs942161146 Chr.5: 33945732-33945732 on Build GRCh38
rs989920888 Chr.5: 33945899-33945899 on Build GRCh38
rs138114400 Chr.5: 33945668-33945668 on Build GRCh38
rs1031855717 Chr.5: 33945628-33945628 on Build GRCh38
rs143719822 Chr.5: 33945810-33945810 on Build GRCh38
rs920750045 Chr.5: 33945824-33945824 on Build GRCh38
rs568302947 Chr.5: 33945566-33945566 on Build GRCh38
rs1017276081 Chr.5: 33945905-33945905 on Build GRCh38
rs1043214684 Chr.5: 33945916-33945916 on Build GRCh38
rs535871299 Chr.5: 33945764-33945764 on Build GRCh38
rs910751553 Chr.5: 33945662-33945662 on Build GRCh38
rs568625467 Chr.5: 33945588-33945588 on Build GRCh38
rs976199180 Chr.5: 33945935-33945935 on Build GRCh38
rs974663375 Chr.5: 33945552-33945552 on Build GRCh38
rs532110475 Chr.5: 33945765-33945765 on Build GRCh38
rs185451833 Chr.5: 33945659-33945659 on Build GRCh38
rs920763991 Chr.5: 33945634-33945634 on Build GRCh38
rs993459 Chr.5: 33945697-33945697 on Build GRCh38
rs1016806770 Chr.5: 33945557-33945557 on Build GRCh38
rs368685594 Chr.5: 33945697-33945698 on Build GRCh38
rs928254701 Chr.5: 33945894-33945894 on Build GRCh38
rs554569811 Chr.5: 33945529-33945529 on Build GRCh38
rs1047747579 Chr.5: 33945725-33945725 on Build GRCh38
rs754422726 Chr.5: 33945973-33945973 on Build GRCh38
rs145802190 Chr.5: 33945898-33945898 on Build GRCh38
rs964863736 Chr.5: 33945922-33945922 on Build GRCh38
rs35895021 Chr.5: 33945543-33945543 on Build GRCh38
rs904337346 Chr.5: 33945936-33945936 on Build GRCh38
rs571535119 Chr.5: 33945543-33945544 on Build GRCh38
rs576158042 Chr.5: 33945636-33945636 on Build GRCh38
rs116232277 Chr.5: 33945817-33945817 on Build GRCh38
rs375127539 Chr.5: 33945542-33945543 on Build GRCh38
rs894315119 Chr.5: 33945777-33945777 on Build GRCh38
rs990290248 Chr.5: 33945660-33945660 on Build GRCh38
rs921897844 Chr.5: 33945552-33945553 on Build GRCh38
rs59234689 Chr.5: 33945676-33945677 on Build GRCh38
rs557882709 Chr.5: 33945834-33945834 on Build GRCh38
rs548358132 Chr.5: 33945891-33945891 on Build GRCh38
rs35405 Chr.5: 33945653-33945653 on Build GRCh38
rs559442432 Chr.5: 33945814-33945814 on Build GRCh38
rs1002524409 Chr.5: 33945661-33945661 on Build GRCh38
rs968968303 Chr.5: 33945700-33945700 on Build GRCh38

View Full Product Details