Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35061296-35061809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRLR
Entrez Gene ID
5618
Gene Name
prolactin receptor
Gene Aliases
HPRL, MFAB, hPRLrI
UniGene
Hs.368587
Gene Chromosome Location
Chr.5: 35048295-35230589 on Build GRCh38
Transcripts
NM_000949.6 NM_001204314.2 NM_001204316.1 NM_001204317.1 NM_001204318.1 NR_037910.1 XM_006714484.2 XM_011514068.2 XM_011514069.2 XM_017009645.1 XM_017009646.1

SNP Detail

SNP ID Chromosome Location
rs917519861 Chr.5: 35061378-35061378 on Build GRCh38
rs893828128 Chr.5: 35061427-35061427 on Build GRCh38
rs1025492821 Chr.5: 35061730-35061730 on Build GRCh38
rs749829577 Chr.5: 35061593-35061593 on Build GRCh38
rs10213953 Chr.5: 35061657-35061657 on Build GRCh38
rs901108191 Chr.5: 35061677-35061677 on Build GRCh38
rs550656403 Chr.5: 35061520-35061520 on Build GRCh38
rs78799003 Chr.5: 35061371-35061371 on Build GRCh38
rs373009060 Chr.5: 35061731-35061731 on Build GRCh38
rs997979848 Chr.5: 35061708-35061708 on Build GRCh38
rs916915584 Chr.5: 35061761-35061761 on Build GRCh38
rs910114024 Chr.5: 35061736-35061736 on Build GRCh38
rs777219167 Chr.5: 35061525-35061525 on Build GRCh38
rs1018366550 Chr.5: 35061596-35061596 on Build GRCh38
rs936079465 Chr.5: 35061529-35061529 on Build GRCh38
rs889096057 Chr.5: 35061631-35061631 on Build GRCh38
rs561664826 Chr.5: 35061327-35061327 on Build GRCh38
rs951535920 Chr.5: 35061716-35061716 on Build GRCh38
rs149171682 Chr.5: 35061648-35061648 on Build GRCh38
rs387032 Chr.5: 35061629-35061629 on Build GRCh38
rs958438941 Chr.5: 35061350-35061350 on Build GRCh38
rs965878747 Chr.5: 35061624-35061624 on Build GRCh38
rs977558916 Chr.5: 35061461-35061461 on Build GRCh38
rs1039999121 Chr.5: 35061661-35061661 on Build GRCh38
rs878947937 Chr.5: 35061573-35061573 on Build GRCh38
rs528313344 Chr.5: 35061753-35061753 on Build GRCh38
rs991529243 Chr.5: 35061754-35061754 on Build GRCh38
rs1007261481 Chr.5: 35061585-35061585 on Build GRCh38
rs774712951 Chr.5: 35061656-35061656 on Build GRCh38
rs886977397 Chr.5: 35061742-35061742 on Build GRCh38
rs552755456 Chr.5: 35061655-35061655 on Build GRCh38
rs566178337 Chr.5: 35061732-35061732 on Build GRCh38
rs776051432 Chr.5: 35061555-35061556 on Build GRCh38
rs943413728 Chr.5: 35061652-35061652 on Build GRCh38
rs192466439 Chr.5: 35061783-35061783 on Build GRCh38
rs1054363188 Chr.5: 35061411-35061411 on Build GRCh38

View Full Product Details