Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39125856-39126401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYB
Entrez Gene ID
2533
Gene Name
FYN binding protein
Gene Aliases
ADAP, PRO0823, SLAP-130, SLAP130
UniGene
Hs.370503
Gene Chromosome Location
Chr.5: 39105252-39274523 on Build GRCh38
Transcripts
NM_001243093.1 NM_001465.4 NM_199335.3 XM_006714464.2 XM_006714465.2 XM_006714466.2 XM_011514008.2 XM_011514009.1 XM_011514010.1 XM_011514011.2 XM_011514012.2 XM_017009316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4533656 c.1972G>A p.G658R Chr.5: 39126071-39126071 on Build GRCh38
5494852 c.1965G>A p.M655I Chr.5: 39126078-39126078 on Build GRCh38
1437496 c.1924C>T p.Q642* Chr.5: 39126119-39126119 on Build GRCh38
6562493 c.2026G>T p.D676Y Chr.5: 39126017-39126017 on Build GRCh38
5175021 c.2045+9C>T p.? Chr.5: 39125989-39125989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374877960 Chr.5: 39126366-39126366 on Build GRCh38
rs780854929 Chr.5: 39125966-39125966 on Build GRCh38
rs1025317037 Chr.5: 39126193-39126193 on Build GRCh38
rs754677315 Chr.5: 39126116-39126116 on Build GRCh38
rs747059357 Chr.5: 39126147-39126147 on Build GRCh38
rs12520149 Chr.5: 39126034-39126034 on Build GRCh38
rs572038588 Chr.5: 39126350-39126350 on Build GRCh38
rs772233717 Chr.5: 39126048-39126048 on Build GRCh38
rs771276757 Chr.5: 39126076-39126076 on Build GRCh38
rs964111198 Chr.5: 39126230-39126230 on Build GRCh38
rs398084079 Chr.5: 39126346-39126346 on Build GRCh38
rs75710164 Chr.5: 39126102-39126102 on Build GRCh38
rs574397015 Chr.5: 39126231-39126231 on Build GRCh38
rs368018464 Chr.5: 39126098-39126098 on Build GRCh38
rs769987835 Chr.5: 39126166-39126166 on Build GRCh38
rs570806148 Chr.5: 39126167-39126167 on Build GRCh38
rs915363716 Chr.5: 39126284-39126284 on Build GRCh38
rs565491305 Chr.5: 39126249-39126249 on Build GRCh38
rs1012102541 Chr.5: 39126118-39126118 on Build GRCh38
rs955063285 Chr.5: 39126200-39126200 on Build GRCh38
rs773585873 Chr.5: 39126063-39126063 on Build GRCh38
rs764578625 Chr.5: 39126182-39126182 on Build GRCh38
rs972410479 Chr.5: 39125893-39125893 on Build GRCh38
rs766334953 Chr.5: 39126143-39126143 on Build GRCh38
rs199924788 Chr.5: 39126043-39126043 on Build GRCh38
rs755830124 Chr.5: 39126108-39126108 on Build GRCh38
rs749031297 Chr.5: 39126033-39126033 on Build GRCh38
rs35310393 Chr.5: 39126100-39126100 on Build GRCh38
rs763507672 Chr.5: 39126176-39126176 on Build GRCh38
rs946974436 Chr.5: 39126285-39126285 on Build GRCh38
rs900460593 Chr.5: 39126301-39126301 on Build GRCh38
rs755916583 Chr.5: 39125981-39125981 on Build GRCh38
rs199923708 Chr.5: 39126121-39126121 on Build GRCh38
rs756071144 Chr.5: 39125908-39125908 on Build GRCh38
rs962927025 Chr.5: 39125891-39125891 on Build GRCh38
rs372366850 Chr.5: 39126149-39126149 on Build GRCh38
rs374823451 Chr.5: 39126079-39126079 on Build GRCh38
rs747966460 Chr.5: 39126128-39126128 on Build GRCh38
rs76502274 Chr.5: 39126339-39126339 on Build GRCh38
rs556456798 Chr.5: 39126225-39126225 on Build GRCh38
rs986452157 Chr.5: 39126224-39126224 on Build GRCh38
rs765797489 Chr.5: 39126086-39126086 on Build GRCh38
rs779881721 Chr.5: 39125986-39125986 on Build GRCh38
rs779648525 Chr.5: 39126109-39126109 on Build GRCh38
rs11956450 Chr.5: 39126234-39126234 on Build GRCh38
rs975297864 Chr.5: 39125997-39125997 on Build GRCh38
rs761138444 Chr.5: 39126106-39126106 on Build GRCh38
rs975014845 Chr.5: 39125974-39125974 on Build GRCh38
rs748280176 Chr.5: 39126045-39126045 on Build GRCh38
rs538141091 Chr.5: 39126190-39126190 on Build GRCh38
rs147362795 Chr.5: 39125943-39125943 on Build GRCh38
rs968169834 Chr.5: 39126003-39126003 on Build GRCh38
rs759977448 Chr.5: 39126153-39126153 on Build GRCh38
rs369724916 Chr.5: 39126327-39126327 on Build GRCh38
rs1009289640 Chr.5: 39125887-39125887 on Build GRCh38
rs968317031 Chr.5: 39126328-39126328 on Build GRCh38
rs148564273 Chr.5: 39126288-39126288 on Build GRCh38
rs771331961 Chr.5: 39126148-39126148 on Build GRCh38
rs749975321 Chr.5: 39126184-39126184 on Build GRCh38
rs901323671 Chr.5: 39126071-39126071 on Build GRCh38
rs1025285612 Chr.5: 39125914-39125914 on Build GRCh38
rs374715818 Chr.5: 39125977-39125977 on Build GRCh38
rs778067764 Chr.5: 39126141-39126141 on Build GRCh38
rs890977161 Chr.5: 39126275-39126275 on Build GRCh38
rs371551176 Chr.5: 39126073-39126073 on Build GRCh38
rs552390836 Chr.5: 39126115-39126115 on Build GRCh38
rs955115519 Chr.5: 39125926-39125926 on Build GRCh38
rs776079266 Chr.5: 39126175-39126175 on Build GRCh38
rs978354652 Chr.5: 39126292-39126292 on Build GRCh38
rs895206552 Chr.5: 39126105-39126105 on Build GRCh38
rs910954897 Chr.5: 39125957-39125957 on Build GRCh38
rs778690148 Chr.5: 39126044-39126044 on Build GRCh38
rs1038249094 Chr.5: 39126130-39126130 on Build GRCh38
rs1008955180 Chr.5: 39126283-39126283 on Build GRCh38
rs915515261 Chr.5: 39126009-39126009 on Build GRCh38
rs759798372 Chr.5: 39126081-39126081 on Build GRCh38
rs924185823 Chr.5: 39126295-39126295 on Build GRCh38
rs756900217 Chr.5: 39126271-39126271 on Build GRCh38
rs759899642 Chr.5: 39126062-39126062 on Build GRCh38
rs144818935 Chr.5: 39126103-39126103 on Build GRCh38

View Full Product Details