Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 45259694-45260231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN1
Entrez Gene ID
348980
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Gene Aliases
BCNG-1, BCNG1, EIEE24, HAC-2
UniGene
Hs.353176
Gene Chromosome Location
Chr.5: 45254950-45696118 on Build GRCh38
Transcripts
NM_021072.3

SNP Detail

SNP ID Chromosome Location
rs966846458 Chr.5: 45260164-45260164 on Build GRCh38
rs114819805 Chr.5: 45259728-45259728 on Build GRCh38
rs952070281 Chr.5: 45260081-45260081 on Build GRCh38
rs768880058 Chr.5: 45259800-45259800 on Build GRCh38
rs944869621 Chr.5: 45259722-45259722 on Build GRCh38
rs997574309 Chr.5: 45260149-45260149 on Build GRCh38
rs1036867845 Chr.5: 45260035-45260035 on Build GRCh38
rs529205348 Chr.5: 45259816-45259816 on Build GRCh38
rs558969244 Chr.5: 45259912-45259912 on Build GRCh38
rs981588066 Chr.5: 45260061-45260061 on Build GRCh38
rs966631982 Chr.5: 45259990-45259990 on Build GRCh38
rs527418258 Chr.5: 45259815-45259815 on Build GRCh38
rs929530637 Chr.5: 45259875-45259875 on Build GRCh38
rs950256279 Chr.5: 45259806-45259806 on Build GRCh38
rs1000693953 Chr.5: 45260190-45260190 on Build GRCh38
rs113074291 Chr.5: 45260199-45260199 on Build GRCh38
rs990496179 Chr.5: 45260047-45260047 on Build GRCh38
rs1020820255 Chr.5: 45260178-45260178 on Build GRCh38
rs1038996892 Chr.5: 45260125-45260125 on Build GRCh38
rs937644804 Chr.5: 45259974-45259974 on Build GRCh38
rs536590848 Chr.5: 45260161-45260161 on Build GRCh38
rs917659156 Chr.5: 45260048-45260048 on Build GRCh38
rs181723853 Chr.5: 45259740-45259740 on Build GRCh38
rs1027618829 Chr.5: 45259881-45259881 on Build GRCh38
rs959329710 Chr.5: 45260023-45260023 on Build GRCh38
rs531382029 Chr.5: 45260053-45260053 on Build GRCh38
rs190444734 Chr.5: 45259964-45259964 on Build GRCh38
rs1049807306 Chr.5: 45260095-45260095 on Build GRCh38
rs924960848 Chr.5: 45259967-45259967 on Build GRCh38
rs979102136 Chr.5: 45259758-45259758 on Build GRCh38
rs901937214 Chr.5: 45260142-45260142 on Build GRCh38
rs374823518 Chr.5: 45259968-45259968 on Build GRCh38
rs902373539 Chr.5: 45260184-45260184 on Build GRCh38
rs903488849 Chr.5: 45259774-45259774 on Build GRCh38
rs868681848 Chr.5: 45260120-45260120 on Build GRCh38
rs1021138396 Chr.5: 45260134-45260134 on Build GRCh38
rs1055308580 Chr.5: 45259794-45259794 on Build GRCh38
rs919401163 Chr.5: 45260064-45260064 on Build GRCh38
rs924913240 Chr.5: 45259768-45259768 on Build GRCh38
rs952017842 Chr.5: 45259901-45259901 on Build GRCh38
rs1007501698 Chr.5: 45260088-45260088 on Build GRCh38
rs878860444 Chr.5: 45260070-45260070 on Build GRCh38
rs760671797 Chr.5: 45260110-45260110 on Build GRCh38
rs550928674 Chr.5: 45260102-45260102 on Build GRCh38
rs979737671 Chr.5: 45259995-45259995 on Build GRCh38
rs937795836 Chr.5: 45259776-45259776 on Build GRCh38
rs763326661 Chr.5: 45259738-45259738 on Build GRCh38
rs1013874505 Chr.5: 45259999-45259999 on Build GRCh38
rs539002540 Chr.5: 45260208-45260208 on Build GRCh38
rs1007545520 Chr.5: 45259963-45259963 on Build GRCh38
rs966579860 Chr.5: 45259747-45259747 on Build GRCh38
rs914018620 Chr.5: 45259955-45259955 on Build GRCh38
rs553658379 Chr.5: 45260193-45260193 on Build GRCh38
rs898088896 Chr.5: 45259821-45259821 on Build GRCh38
rs187242233 Chr.5: 45260195-45260195 on Build GRCh38
rs917664728 Chr.5: 45259803-45259803 on Build GRCh38
rs997354663 Chr.5: 45259971-45259971 on Build GRCh38
rs368283110 Chr.5: 45259984-45259984 on Build GRCh38
rs549247903 Chr.5: 45259880-45259880 on Build GRCh38
rs116151715 Chr.5: 45260122-45260122 on Build GRCh38
rs1024842626 Chr.5: 45259836-45259839 on Build GRCh38
rs1040550277 Chr.5: 45259725-45259725 on Build GRCh38
rs901925811 Chr.5: 45259966-45259966 on Build GRCh38
rs186118083 Chr.5: 45259929-45259929 on Build GRCh38
rs976274855 Chr.5: 45259940-45259940 on Build GRCh38
rs949104559 Chr.5: 45260059-45260059 on Build GRCh38
rs929457141 Chr.5: 45260091-45260091 on Build GRCh38
rs966645500 Chr.5: 45259931-45259931 on Build GRCh38
rs74886762 Chr.5: 45259905-45259905 on Build GRCh38
rs181985490 Chr.5: 45259997-45259997 on Build GRCh38
rs543222928 Chr.5: 45259729-45259729 on Build GRCh38
rs1055026965 Chr.5: 45259812-45259812 on Build GRCh38

View Full Product Details