Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60752089-60752589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

SNP Detail

SNP ID Chromosome Location
rs702564 Chr.5: 60752147-60752147 on Build GRCh38
rs969126358 Chr.5: 60752251-60752251 on Build GRCh38
rs752494401 Chr.5: 60752486-60752486 on Build GRCh38
rs554816397 Chr.5: 60752133-60752133 on Build GRCh38
rs1045334276 Chr.5: 60752438-60752438 on Build GRCh38
rs763757476 Chr.5: 60752434-60752438 on Build GRCh38
rs941588401 Chr.5: 60752436-60752436 on Build GRCh38
rs959662529 Chr.5: 60752139-60752139 on Build GRCh38
rs1040847839 Chr.5: 60752175-60752175 on Build GRCh38
rs751037563 Chr.5: 60752485-60752485 on Build GRCh38
rs543166438 Chr.5: 60752146-60752146 on Build GRCh38
rs942561670 Chr.5: 60752281-60752281 on Build GRCh38
rs1009970526 Chr.5: 60752488-60752488 on Build GRCh38
rs755865518 Chr.5: 60752510-60752510 on Build GRCh38
rs981893493 Chr.5: 60752258-60752258 on Build GRCh38
rs1023724657 Chr.5: 60752233-60752233 on Build GRCh38
rs1047084292 Chr.5: 60752371-60752371 on Build GRCh38
rs977207337 Chr.5: 60752294-60752294 on Build GRCh38
rs189471181 Chr.5: 60752136-60752136 on Build GRCh38
rs952753502 Chr.5: 60752215-60752215 on Build GRCh38
rs16878354 Chr.5: 60752201-60752201 on Build GRCh38
rs756689938 Chr.5: 60752557-60752557 on Build GRCh38
rs930081699 Chr.5: 60752355-60752355 on Build GRCh38
rs757403993 Chr.5: 60752275-60752275 on Build GRCh38
rs989396490 Chr.5: 60752397-60752397 on Build GRCh38
rs180924470 Chr.5: 60752153-60752153 on Build GRCh38
rs1035412549 Chr.5: 60752117-60752117 on Build GRCh38
rs751392422 Chr.5: 60752559-60752559 on Build GRCh38
rs1003719253 Chr.5: 60752470-60752470 on Build GRCh38
rs913449173 Chr.5: 60752398-60752398 on Build GRCh38
rs1019943256 Chr.5: 60752197-60752197 on Build GRCh38
rs559509009 Chr.5: 60752270-60752270 on Build GRCh38
rs960219741 Chr.5: 60752382-60752382 on Build GRCh38
rs868169576 Chr.5: 60752461-60752461 on Build GRCh38
rs145164561 Chr.5: 60752539-60752539 on Build GRCh38
rs751521827 Chr.5: 60752227-60752227 on Build GRCh38
rs928777224 Chr.5: 60752367-60752367 on Build GRCh38
rs562358304 Chr.5: 60752451-60752451 on Build GRCh38
rs759459965 Chr.5: 60752352-60752352 on Build GRCh38
rs1043456703 Chr.5: 60752142-60752142 on Build GRCh38
rs994026529 Chr.5: 60752195-60752195 on Build GRCh38
rs534997805 Chr.5: 60752312-60752312 on Build GRCh38
rs947378337 Chr.5: 60752128-60752128 on Build GRCh38
rs189679004 Chr.5: 60752428-60752428 on Build GRCh38
rs895561709 Chr.5: 60752487-60752487 on Build GRCh38
rs978726689 Chr.5: 60752232-60752232 on Build GRCh38
rs947626984 Chr.5: 60752418-60752418 on Build GRCh38
rs184922158 Chr.5: 60752334-60752334 on Build GRCh38
rs901041661 Chr.5: 60752182-60752182 on Build GRCh38
rs942774380 Chr.5: 60752484-60752484 on Build GRCh38

View Full Product Details