Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65635531-65636043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC13
Entrez Gene ID
80006
Gene Name
trafficking protein particle complex 13
Gene Aliases
C5orf44
UniGene
Hs.591760
Gene Chromosome Location
Chr.5: 65624731-65666136 on Build GRCh38
Transcripts
NM_001093755.1 NM_001093756.1 NM_001243737.1 NM_024941.3 XM_011543652.2 XM_017009890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581488 c.164G>A p.G55D Chr.5: 65635992-65635992 on Build GRCh38
5403712 c.123C>T p.L41L Chr.5: 65635951-65635951 on Build GRCh38
3346220 c.121C>T p.L41F Chr.5: 65635949-65635949 on Build GRCh38
4778685 c.152C>T p.S51L Chr.5: 65635980-65635980 on Build GRCh38
6581487 c.164G>A p.G55D Chr.5: 65635992-65635992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532781086 Chr.5: 65635656-65635656 on Build GRCh38
rs781065064 Chr.5: 65635994-65635994 on Build GRCh38
rs1035795170 Chr.5: 65635921-65635921 on Build GRCh38
rs891833307 Chr.5: 65635925-65635925 on Build GRCh38
rs1003255639 Chr.5: 65635907-65635907 on Build GRCh38
rs778759773 Chr.5: 65635686-65635686 on Build GRCh38
rs376135247 Chr.5: 65635699-65635699 on Build GRCh38
rs112555567 Chr.5: 65635687-65635687 on Build GRCh38
rs905848405 Chr.5: 65635793-65635793 on Build GRCh38
rs774044740 Chr.5: 65635937-65635937 on Build GRCh38
rs914594424 Chr.5: 65636008-65636008 on Build GRCh38
rs566730994 Chr.5: 65635601-65635601 on Build GRCh38
rs5868411 Chr.5: 65635777-65635778 on Build GRCh38
rs766306704 Chr.5: 65635977-65635977 on Build GRCh38
rs760795952 Chr.5: 65635614-65635614 on Build GRCh38
rs1034968296 Chr.5: 65635720-65635720 on Build GRCh38
rs199882916 Chr.5: 65635938-65635938 on Build GRCh38
rs753984454 Chr.5: 65635982-65635982 on Build GRCh38
rs772331225 Chr.5: 65635825-65635825 on Build GRCh38
rs1015035402 Chr.5: 65635862-65635862 on Build GRCh38
rs888374386 Chr.5: 65635730-65635730 on Build GRCh38
rs779273593 Chr.5: 65635984-65635984 on Build GRCh38
rs570299029 Chr.5: 65635962-65635962 on Build GRCh38
rs947946322 Chr.5: 65635768-65635768 on Build GRCh38
rs552688617 Chr.5: 65635657-65635657 on Build GRCh38
rs922147307 Chr.5: 65635958-65635958 on Build GRCh38
rs373703606 Chr.5: 65635991-65635991 on Build GRCh38
rs763748530 Chr.5: 65635908-65635908 on Build GRCh38
rs1044437264 Chr.5: 65635786-65635786 on Build GRCh38
rs757696278 Chr.5: 65635963-65635963 on Build GRCh38
rs568404516 Chr.5: 65635874-65635874 on Build GRCh38
rs550491488 Chr.5: 65635957-65635957 on Build GRCh38
rs943809223 Chr.5: 65635585-65635585 on Build GRCh38
rs748357016 Chr.5: 65635770-65635770 on Build GRCh38
rs1003447365 Chr.5: 65635718-65635718 on Build GRCh38
rs370490160 Chr.5: 65635985-65635985 on Build GRCh38
rs376180020 Chr.5: 65636015-65636015 on Build GRCh38
rs745615721 Chr.5: 65635905-65635905 on Build GRCh38
rs983247185 Chr.5: 65635845-65635845 on Build GRCh38
rs547052616 Chr.5: 65635576-65635576 on Build GRCh38
rs37343 Chr.5: 65635686-65635686 on Build GRCh38
rs755171533 Chr.5: 65635983-65635983 on Build GRCh38
rs1042590807 Chr.5: 65635690-65635690 on Build GRCh38
rs552345419 Chr.5: 65635729-65635729 on Build GRCh38
rs932021623 Chr.5: 65635679-65635679 on Build GRCh38
rs548523343 Chr.5: 65635742-65635742 on Build GRCh38
rs745751305 Chr.5: 65635999-65635999 on Build GRCh38
rs946163463 Chr.5: 65635687-65635687 on Build GRCh38
rs34009243 Chr.5: 65635870-65635871 on Build GRCh38
rs750870890 Chr.5: 65635960-65635960 on Build GRCh38
rs115830247 Chr.5: 65635717-65635717 on Build GRCh38

View Full Product Details