Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69094250-69094759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A5
Entrez Gene ID
64924
Gene Name
solute carrier family 30 member 5
Gene Aliases
ZNT5, ZNTL1, ZTL1, ZnT-5
UniGene
Hs.631975
Gene Chromosome Location
Chr.5: 69093949-69131072 on Build GRCh38
Transcripts
NM_001251969.1 NM_022902.4 NM_024055.4 XM_005248569.3 XM_006714672.3 XM_017009749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6529530 c.79G>C p.A27P Chr.5: 69094334-69094334 on Build GRCh38
4159955 c.23A>G p.D8G Chr.5: 69094278-69094278 on Build GRCh38
4159956 c.23A>G p.D8G Chr.5: 69094278-69094278 on Build GRCh38
6529529 c.79G>C p.A27P Chr.5: 69094334-69094334 on Build GRCh38
6390272 c.56T>A p.L19H Chr.5: 69094311-69094311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918437730 Chr.5: 69094574-69094574 on Build GRCh38
rs337259 Chr.5: 69094457-69094457 on Build GRCh38
rs1034678441 Chr.5: 69094535-69094535 on Build GRCh38
rs999675726 Chr.5: 69094280-69094280 on Build GRCh38
rs777592236 Chr.5: 69094707-69094707 on Build GRCh38
rs918302018 Chr.5: 69094293-69094294 on Build GRCh38
rs1050725512 Chr.5: 69094337-69094337 on Build GRCh38
rs896223583 Chr.5: 69094558-69094558 on Build GRCh38
rs896254742 Chr.5: 69094307-69094307 on Build GRCh38
rs1044016697 Chr.5: 69094506-69094506 on Build GRCh38
rs769532677 Chr.5: 69094568-69094568 on Build GRCh38
rs1001399451 Chr.5: 69094446-69094446 on Build GRCh38
rs986752354 Chr.5: 69094468-69094468 on Build GRCh38
rs1030152346 Chr.5: 69094656-69094656 on Build GRCh38
rs536999216 Chr.5: 69094377-69094377 on Build GRCh38
rs558596180 Chr.5: 69094498-69094498 on Build GRCh38
rs1052617380 Chr.5: 69094551-69094551 on Build GRCh38
rs942357414 Chr.5: 69094596-69094596 on Build GRCh38
rs934092132 Chr.5: 69094466-69094466 on Build GRCh38
rs969691677 Chr.5: 69094344-69094344 on Build GRCh38
rs756525450 Chr.5: 69094371-69094371 on Build GRCh38
rs929720122 Chr.5: 69094314-69094314 on Build GRCh38
rs904142404 Chr.5: 69094433-69094433 on Build GRCh38
rs377352065 Chr.5: 69094400-69094400 on Build GRCh38
rs190983637 Chr.5: 69094513-69094513 on Build GRCh38
rs892618618 Chr.5: 69094711-69094711 on Build GRCh38
rs375339220 Chr.5: 69094492-69094492 on Build GRCh38
rs1006305467 Chr.5: 69094583-69094583 on Build GRCh38
rs1052623378 Chr.5: 69094454-69094454 on Build GRCh38
rs912349237 Chr.5: 69094589-69094589 on Build GRCh38
rs971872756 Chr.5: 69094447-69094447 on Build GRCh38
rs528270005 Chr.5: 69094690-69094690 on Build GRCh38
rs574746815 Chr.5: 69094606-69094606 on Build GRCh38
rs993264124 Chr.5: 69094647-69094647 on Build GRCh38
rs1030723706 Chr.5: 69094286-69094286 on Build GRCh38
rs1044749650 Chr.5: 69094577-69094577 on Build GRCh38
rs925535882 Chr.5: 69094511-69094511 on Build GRCh38
rs188179381 Chr.5: 69094600-69094600 on Build GRCh38
rs1040351745 Chr.5: 69094427-69094427 on Build GRCh38
rs561044575 Chr.5: 69094672-69094672 on Build GRCh38
rs337260 Chr.5: 69094376-69094376 on Build GRCh38
rs1013364740 Chr.5: 69094564-69094564 on Build GRCh38
rs1023334514 Chr.5: 69094338-69094338 on Build GRCh38
rs867904934 Chr.5: 69094710-69094710 on Build GRCh38
rs554966035 Chr.5: 69094439-69094439 on Build GRCh38
rs1013227757 Chr.5: 69094325-69094325 on Build GRCh38
rs193162200 Chr.5: 69094609-69094609 on Build GRCh38
rs964045988 Chr.5: 69094732-69094732 on Build GRCh38
rs183550756 Chr.5: 69094578-69094578 on Build GRCh38
rs866951918 Chr.5: 69094597-69094597 on Build GRCh38
rs770225872 Chr.5: 69094294-69094296 on Build GRCh38
rs955440591 Chr.5: 69094668-69094668 on Build GRCh38
rs962660942 Chr.5: 69094563-69094563 on Build GRCh38
rs534138269 Chr.5: 69094488-69094488 on Build GRCh38
rs559048933 Chr.5: 69094585-69094585 on Build GRCh38

View Full Product Details