Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69104649-69105195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A5
Entrez Gene ID
64924
Gene Name
solute carrier family 30 member 5
Gene Aliases
ZNT5, ZNTL1, ZTL1, ZnT-5
UniGene
Hs.631975
Gene Chromosome Location
Chr.5: 69093949-69131072 on Build GRCh38
Transcripts
NM_001251969.1 NM_022902.4 NM_024055.4 XM_005248569.3 XM_006714672.3 XM_017009749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104364 c.352C>G p.P118A Chr.5: 69104709-69104709 on Build GRCh38
738332 c.359G>T p.R120M Chr.5: 69104716-69104716 on Build GRCh38
1069660 c.324G>T p.L108F Chr.5: 69104681-69104681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576928097 Chr.5: 69104797-69104797 on Build GRCh38
rs1004903770 Chr.5: 69105095-69105095 on Build GRCh38
rs985020744 Chr.5: 69105069-69105069 on Build GRCh38
rs922133880 Chr.5: 69105139-69105139 on Build GRCh38
rs575789051 Chr.5: 69104772-69104774 on Build GRCh38
rs1050407339 Chr.5: 69104801-69104801 on Build GRCh38
rs555815707 Chr.5: 69104983-69104983 on Build GRCh38
rs1010791304 Chr.5: 69104844-69104844 on Build GRCh38
rs78986159 Chr.5: 69104756-69104756 on Build GRCh38
rs1015574245 Chr.5: 69105118-69105118 on Build GRCh38
rs953527896 Chr.5: 69105157-69105157 on Build GRCh38
rs952114289 Chr.5: 69105068-69105068 on Build GRCh38
rs582068 Chr.5: 69104805-69104805 on Build GRCh38
rs74430495 Chr.5: 69104837-69104837 on Build GRCh38
rs893100800 Chr.5: 69105154-69105154 on Build GRCh38
rs893154503 Chr.5: 69104810-69104810 on Build GRCh38
rs940860264 Chr.5: 69105094-69105094 on Build GRCh38
rs1034755078 Chr.5: 69104992-69104992 on Build GRCh38
rs558823742 Chr.5: 69104794-69104794 on Build GRCh38
rs760662573 Chr.5: 69104701-69104702 on Build GRCh38
rs753799410 Chr.5: 69104750-69104750 on Build GRCh38
rs541414471 Chr.5: 69105149-69105149 on Build GRCh38
rs768041272 Chr.5: 69104731-69104731 on Build GRCh38
rs916221333 Chr.5: 69105040-69105040 on Build GRCh38
rs533798758 Chr.5: 69104773-69104773 on Build GRCh38
rs762050936 Chr.5: 69104715-69104715 on Build GRCh38
rs80069924 Chr.5: 69104738-69104738 on Build GRCh38
rs966568722 Chr.5: 69104911-69104911 on Build GRCh38
rs951809749 Chr.5: 69105049-69105049 on Build GRCh38
rs149336330 Chr.5: 69104768-69104770 on Build GRCh38
rs547854172 Chr.5: 69104742-69104742 on Build GRCh38
rs16898499 Chr.5: 69105138-69105138 on Build GRCh38
rs753262052 Chr.5: 69104982-69104982 on Build GRCh38
rs766994157 Chr.5: 69104744-69104744 on Build GRCh38
rs1022638241 Chr.5: 69104853-69104853 on Build GRCh38
rs990587103 Chr.5: 69105031-69105031 on Build GRCh38
rs774119476 Chr.5: 69104697-69104697 on Build GRCh38
rs895234212 Chr.5: 69104831-69104831 on Build GRCh38
rs1055590033 Chr.5: 69104795-69104795 on Build GRCh38
rs759044650 Chr.5: 69105160-69105160 on Build GRCh38
rs908158062 Chr.5: 69105073-69105075 on Build GRCh38
rs112391245 Chr.5: 69105044-69105044 on Build GRCh38
rs1002651906 Chr.5: 69104780-69104780 on Build GRCh38
rs745904319 Chr.5: 69105101-69105102 on Build GRCh38
rs960468360 Chr.5: 69105030-69105030 on Build GRCh38
rs966312068 Chr.5: 69105119-69105119 on Build GRCh38

View Full Product Details