Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69167828-69168338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1
Entrez Gene ID
891
Gene Name
cyclin B1
Gene Aliases
CCNB
UniGene
Hs.23960
Gene Chromosome Location
Chr.5: 69167010-69178245 on Build GRCh38
Transcripts
NM_031966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828348 c.22-1G>A p.? Chr.5: 69167907-69167907 on Build GRCh38
1739185 c.36T>G p.N12K Chr.5: 69167922-69167922 on Build GRCh38
4452423 c.184A>T p.M62L Chr.5: 69168070-69168070 on Build GRCh38
4957988 c.174C>T p.A58A Chr.5: 69168060-69168060 on Build GRCh38
1069677 c.245C>A p.P82H Chr.5: 69168225-69168225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773343741 Chr.5: 69168176-69168176 on Build GRCh38
rs780787172 Chr.5: 69168130-69168130 on Build GRCh38
rs1002946303 Chr.5: 69167997-69167997 on Build GRCh38
rs373480903 Chr.5: 69167857-69167857 on Build GRCh38
rs865916737 Chr.5: 69167854-69167854 on Build GRCh38
rs145759905 Chr.5: 69168072-69168072 on Build GRCh38
rs778022575 Chr.5: 69167875-69167875 on Build GRCh38
rs552306325 Chr.5: 69168090-69168090 on Build GRCh38
rs530270969 Chr.5: 69167852-69167852 on Build GRCh38
rs765963344 Chr.5: 69167979-69167979 on Build GRCh38
rs954971463 Chr.5: 69168242-69168242 on Build GRCh38
rs770501273 Chr.5: 69167939-69167939 on Build GRCh38
rs766962560 Chr.5: 69168153-69168156 on Build GRCh38
rs764207072 Chr.5: 69167962-69167962 on Build GRCh38
rs775282499 Chr.5: 69167898-69167904 on Build GRCh38
rs370662203 Chr.5: 69168117-69168117 on Build GRCh38
rs752500659 Chr.5: 69168112-69168112 on Build GRCh38
rs865987149 Chr.5: 69167983-69167983 on Build GRCh38
rs769586600 Chr.5: 69168131-69168131 on Build GRCh38
rs777039051 Chr.5: 69167950-69167950 on Build GRCh38
rs758600094 Chr.5: 69167869-69167869 on Build GRCh38
rs1013381457 Chr.5: 69167868-69167868 on Build GRCh38
rs1034366112 Chr.5: 69168021-69168021 on Build GRCh38
rs757225235 Chr.5: 69168229-69168229 on Build GRCh38
rs933588052 Chr.5: 69167969-69167969 on Build GRCh38
rs748534626 Chr.5: 69168292-69168292 on Build GRCh38
rs758336269 Chr.5: 69168007-69168007 on Build GRCh38
rs749098045 Chr.5: 69167890-69167890 on Build GRCh38
rs768849718 Chr.5: 69168288-69168288 on Build GRCh38
rs759192061 Chr.5: 69167858-69167858 on Build GRCh38
rs936308097 Chr.5: 69168215-69168215 on Build GRCh38
rs779376687 Chr.5: 69168242-69168243 on Build GRCh38
rs752181166 Chr.5: 69168153-69168154 on Build GRCh38
rs745557563 Chr.5: 69168053-69168053 on Build GRCh38
rs747630808 Chr.5: 69167917-69167917 on Build GRCh38
rs374129207 Chr.5: 69168203-69168203 on Build GRCh38
rs1000750363 Chr.5: 69167878-69167878 on Build GRCh38
rs377039637 Chr.5: 69167910-69167910 on Build GRCh38
rs757429226 Chr.5: 69167889-69167889 on Build GRCh38
rs773265958 Chr.5: 69168298-69168298 on Build GRCh38
rs899494711 Chr.5: 69167871-69167871 on Build GRCh38
rs768630510 Chr.5: 69167905-69167905 on Build GRCh38
rs769258814 Chr.5: 69168148-69168148 on Build GRCh38
rs752484310 Chr.5: 69168196-69168196 on Build GRCh38
rs768857099 Chr.5: 69168277-69168278 on Build GRCh38
rs762338775 Chr.5: 69167963-69167963 on Build GRCh38
rs776004787 Chr.5: 69168067-69168067 on Build GRCh38
rs752795547 Chr.5: 69167863-69167863 on Build GRCh38
rs943817844 Chr.5: 69168170-69168170 on Build GRCh38
rs763498172 Chr.5: 69168070-69168070 on Build GRCh38
rs765598533 Chr.5: 69168195-69168195 on Build GRCh38
rs527515881 Chr.5: 69168278-69168289 on Build GRCh38
rs755827930 Chr.5: 69168258-69168258 on Build GRCh38
rs776092366 Chr.5: 69167957-69167957 on Build GRCh38
rs780419001 Chr.5: 69168256-69168267 on Build GRCh38
rs560413531 Chr.5: 69168032-69168032 on Build GRCh38
rs760811100 Chr.5: 69168299-69168299 on Build GRCh38
rs767702393 Chr.5: 69167973-69167973 on Build GRCh38
rs751415136 Chr.5: 69168225-69168225 on Build GRCh38
rs904514899 Chr.5: 69168083-69168083 on Build GRCh38
rs1043201171 Chr.5: 69168075-69168075 on Build GRCh38
rs750559524 Chr.5: 69168087-69168087 on Build GRCh38
rs771109821 Chr.5: 69168302-69168302 on Build GRCh38
rs140927235 Chr.5: 69167919-69167919 on Build GRCh38
rs373876739 Chr.5: 69167881-69167881 on Build GRCh38
rs377662845 Chr.5: 69168183-69168183 on Build GRCh38
rs753152559 Chr.5: 69168042-69168042 on Build GRCh38
rs770587603 Chr.5: 69168044-69168044 on Build GRCh38
rs772739694 Chr.5: 69167922-69167922 on Build GRCh38
rs749741212 Chr.5: 69168136-69168136 on Build GRCh38
rs139043163 Chr.5: 69168114-69168114 on Build GRCh38
rs749645134 Chr.5: 69168286-69168286 on Build GRCh38
rs750829238 Chr.5: 69168247-69168252 on Build GRCh38
rs777205019 Chr.5: 69168014-69168014 on Build GRCh38
rs542396320 Chr.5: 69167867-69167867 on Build GRCh38
rs759860075 Chr.5: 69167953-69167953 on Build GRCh38
rs757825094 Chr.5: 69168160-69168161 on Build GRCh38
rs772003288 Chr.5: 69168162-69168162 on Build GRCh38
rs770244587 Chr.5: 69168066-69168066 on Build GRCh38
rs764944388 Chr.5: 69168105-69168105 on Build GRCh38
rs920947659 Chr.5: 69168208-69168208 on Build GRCh38
rs748035379 Chr.5: 69168002-69168002 on Build GRCh38
rs779896517 Chr.5: 69168272-69168272 on Build GRCh38
rs780512901 Chr.5: 69168235-69168235 on Build GRCh38
rs867133028 Chr.5: 69168270-69168270 on Build GRCh38
rs201559153 Chr.5: 69168132-69168132 on Build GRCh38
rs774488515 Chr.5: 69168071-69168071 on Build GRCh38
rs201224572 Chr.5: 69168115-69168115 on Build GRCh38
rs746606130 Chr.5: 69167929-69167929 on Build GRCh38
rs762399522 Chr.5: 69168161-69168161 on Build GRCh38
rs774765185 Chr.5: 69168150-69168150 on Build GRCh38
rs754814106 Chr.5: 69168000-69168000 on Build GRCh38
rs758742437 Chr.5: 69168250-69168267 on Build GRCh38
rs369506933 Chr.5: 69167966-69167966 on Build GRCh38
rs892418645 Chr.5: 69168308-69168308 on Build GRCh38
rs760827475 Chr.5: 69168178-69168178 on Build GRCh38
rs776196962 Chr.5: 69168052-69168052 on Build GRCh38
rs147065029 Chr.5: 69168289-69168289 on Build GRCh38
rs141602648 Chr.5: 69168200-69168200 on Build GRCh38
rs971913060 Chr.5: 69167987-69167987 on Build GRCh38
rs750014957 Chr.5: 69168240-69168240 on Build GRCh38
rs750148681 Chr.5: 69167956-69167956 on Build GRCh38
rs146181241 Chr.5: 69167851-69167851 on Build GRCh38

View Full Product Details