Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69259897-69260433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK7
Entrez Gene ID
1022
Gene Name
cyclin dependent kinase 7
Gene Aliases
CAK, CAK1, CDKN7, HCAK, MO15, STK1, p39MO15
UniGene
Hs.184298
Gene Chromosome Location
Chr.5: 69234795-69277430 on Build GRCh38
Transcripts
NM_001324069.1 NM_001324070.1 NM_001324071.1 NM_001324072.1 NM_001324074.1 NM_001324075.1 NM_001324077.1 NM_001324078.1 NM_001799.3 NR_136690.1 XM_011543094.2

SNP Detail

SNP ID Chromosome Location
rs567248192 Chr.5: 69260306-69260306 on Build GRCh38
rs1034352318 Chr.5: 69260391-69260391 on Build GRCh38
rs982511342 Chr.5: 69260219-69260219 on Build GRCh38
rs936591953 Chr.5: 69260312-69260312 on Build GRCh38
rs775887802 Chr.5: 69259941-69259941 on Build GRCh38
rs368299348 Chr.5: 69259947-69259947 on Build GRCh38
rs759201733 Chr.5: 69260088-69260088 on Build GRCh38
rs892279851 Chr.5: 69260321-69260321 on Build GRCh38
rs548745386 Chr.5: 69260119-69260119 on Build GRCh38
rs943994774 Chr.5: 69259975-69259975 on Build GRCh38
rs997805316 Chr.5: 69259948-69259948 on Build GRCh38
rs549092244 Chr.5: 69260218-69260218 on Build GRCh38
rs1040046638 Chr.5: 69260007-69260007 on Build GRCh38
rs545247410 Chr.5: 69260074-69260074 on Build GRCh38
rs755773421 Chr.5: 69260315-69260315 on Build GRCh38
rs774698219 Chr.5: 69259960-69259960 on Build GRCh38
rs563055858 Chr.5: 69260111-69260111 on Build GRCh38
rs1041796592 Chr.5: 69260342-69260342 on Build GRCh38
rs928029972 Chr.5: 69260076-69260076 on Build GRCh38
rs954283931 Chr.5: 69259980-69259980 on Build GRCh38
rs1003288340 Chr.5: 69260369-69260369 on Build GRCh38
rs1035468624 Chr.5: 69260222-69260222 on Build GRCh38
rs943717009 Chr.5: 69260274-69260274 on Build GRCh38
rs999674793 Chr.5: 69260371-69260374 on Build GRCh38
rs777246408 Chr.5: 69260366-69260367 on Build GRCh38
rs997838261 Chr.5: 69260309-69260309 on Build GRCh38
rs187250655 Chr.5: 69260077-69260077 on Build GRCh38
rs1001117484 Chr.5: 69260070-69260070 on Build GRCh38
rs966823304 Chr.5: 69260045-69260045 on Build GRCh38
rs755786113 Chr.5: 69260184-69260184 on Build GRCh38
rs143444863 Chr.5: 69260266-69260266 on Build GRCh38
rs974277494 Chr.5: 69260193-69260193 on Build GRCh38
rs1048225754 Chr.5: 69260207-69260207 on Build GRCh38
rs1035129616 Chr.5: 69260376-69260376 on Build GRCh38
rs1035022015 Chr.5: 69260132-69260132 on Build GRCh38
rs945187821 Chr.5: 69260326-69260326 on Build GRCh38
rs180962343 Chr.5: 69259930-69259930 on Build GRCh38
rs967317935 Chr.5: 69260371-69260371 on Build GRCh38
rs907249739 Chr.5: 69260365-69260365 on Build GRCh38
rs766350121 Chr.5: 69260075-69260075 on Build GRCh38
rs1027148782 Chr.5: 69260214-69260214 on Build GRCh38
rs909674480 Chr.5: 69260212-69260212 on Build GRCh38
rs762190330 Chr.5: 69259965-69259965 on Build GRCh38
rs113841347 Chr.5: 69260044-69260044 on Build GRCh38
rs1053576823 Chr.5: 69260313-69260313 on Build GRCh38
rs200813690 Chr.5: 69259954-69259954 on Build GRCh38
rs2130757 Chr.5: 69260127-69260127 on Build GRCh38
rs571472698 Chr.5: 69260299-69260299 on Build GRCh38
rs148835101 Chr.5: 69259974-69259974 on Build GRCh38
rs990806223 Chr.5: 69260088-69260089 on Build GRCh38
rs939862588 Chr.5: 69260235-69260235 on Build GRCh38
rs547473022 Chr.5: 69260379-69260381 on Build GRCh38
rs990837311 Chr.5: 69260405-69260405 on Build GRCh38
rs763667579 Chr.5: 69260204-69260204 on Build GRCh38
rs1049358361 Chr.5: 69260311-69260311 on Build GRCh38
rs377040324 Chr.5: 69260118-69260118 on Build GRCh38
rs1011089996 Chr.5: 69260032-69260032 on Build GRCh38
rs533218664 Chr.5: 69259984-69259984 on Build GRCh38

View Full Product Details