Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72861715-72862254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNPO1
Entrez Gene ID
3842
Gene Name
transportin 1
Gene Aliases
IPO2, KPNB2, MIP, MIP1, TRN
UniGene
Hs.482497
Gene Chromosome Location
Chr.5: 72816591-72914388 on Build GRCh38
Transcripts
NM_002270.3 NM_153188.2 XM_005248500.2 XM_005248501.1 XM_017009458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625595 c.372G>T p.Q124H Chr.5: 72861848-72861848 on Build GRCh38
1069885 c.424T>C p.Y142H Chr.5: 72861900-72861900 on Build GRCh38
7334518 c.346A>G p.T116A Chr.5: 72861822-72861822 on Build GRCh38
6264741 c.338T>C p.L113S Chr.5: 72861814-72861814 on Build GRCh38
3856209 c.364G>C p.E122Q Chr.5: 72861840-72861840 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751426106 Chr.5: 72861881-72861881 on Build GRCh38
rs768294052 Chr.5: 72861812-72861812 on Build GRCh38
rs375597055 Chr.5: 72862231-72862231 on Build GRCh38
rs1039402515 Chr.5: 72862106-72862106 on Build GRCh38
rs568018129 Chr.5: 72862172-72862172 on Build GRCh38
rs1032863103 Chr.5: 72862170-72862170 on Build GRCh38
rs376254935 Chr.5: 72861771-72861771 on Build GRCh38
rs772665753 Chr.5: 72861825-72861825 on Build GRCh38
rs188901104 Chr.5: 72862091-72862091 on Build GRCh38
rs774704464 Chr.5: 72861822-72861822 on Build GRCh38
rs763254014 Chr.5: 72861992-72861992 on Build GRCh38
rs144903373 Chr.5: 72861755-72861755 on Build GRCh38
rs759151077 Chr.5: 72861927-72861928 on Build GRCh38
rs772369529 Chr.5: 72861944-72861944 on Build GRCh38
rs766449131 Chr.5: 72861760-72861760 on Build GRCh38
rs759839019 Chr.5: 72861872-72861872 on Build GRCh38
rs941557448 Chr.5: 72861972-72861972 on Build GRCh38
rs901048090 Chr.5: 72861750-72861750 on Build GRCh38
rs534325427 Chr.5: 72861773-72861773 on Build GRCh38
rs572114534 Chr.5: 72861767-72861767 on Build GRCh38
rs185469979 Chr.5: 72861962-72861962 on Build GRCh38
rs531429109 Chr.5: 72862100-72862100 on Build GRCh38
rs377758025 Chr.5: 72861923-72861923 on Build GRCh38
rs774100674 Chr.5: 72861791-72861792 on Build GRCh38
rs564066697 Chr.5: 72862097-72862097 on Build GRCh38
rs761539949 Chr.5: 72862133-72862133 on Build GRCh38
rs1035197733 Chr.5: 72861867-72861867 on Build GRCh38
rs560758944 Chr.5: 72862011-72862011 on Build GRCh38
rs750377984 Chr.5: 72861908-72861908 on Build GRCh38
rs535655077 Chr.5: 72862165-72862165 on Build GRCh38
rs747765365 Chr.5: 72861942-72861942 on Build GRCh38
rs770690057 Chr.5: 72861779-72861779 on Build GRCh38
rs558815138 Chr.5: 72861775-72861775 on Build GRCh38
rs201796023 Chr.5: 72861792-72861792 on Build GRCh38
rs1032810961 Chr.5: 72861884-72861884 on Build GRCh38
rs544239717 Chr.5: 72861907-72861907 on Build GRCh38
rs770906071 Chr.5: 72862152-72862152 on Build GRCh38
rs200553704 Chr.5: 72861832-72861832 on Build GRCh38
rs915172415 Chr.5: 72862149-72862149 on Build GRCh38
rs140341824 Chr.5: 72862189-72862189 on Build GRCh38
rs749000344 Chr.5: 72861801-72861801 on Build GRCh38
rs373444861 Chr.5: 72861921-72861921 on Build GRCh38
rs929079112 Chr.5: 72862016-72862016 on Build GRCh38
rs778497187 Chr.5: 72861931-72861931 on Build GRCh38
rs780946157 Chr.5: 72861780-72861780 on Build GRCh38
rs372741080 Chr.5: 72861785-72861785 on Build GRCh38
rs898909630 Chr.5: 72861762-72861762 on Build GRCh38
rs182004425 Chr.5: 72861927-72861927 on Build GRCh38
rs963793880 Chr.5: 72862029-72862029 on Build GRCh38
rs765495171 Chr.5: 72861879-72861879 on Build GRCh38
rs757082032 Chr.5: 72861901-72861901 on Build GRCh38
rs1009243899 Chr.5: 72861821-72861821 on Build GRCh38
rs3213807 Chr.5: 72862055-72862055 on Build GRCh38
rs761220626 Chr.5: 72861834-72861834 on Build GRCh38
rs747533414 Chr.5: 72861953-72861953 on Build GRCh38
rs766425223 Chr.5: 72861838-72861838 on Build GRCh38
rs745335586 Chr.5: 72861751-72861751 on Build GRCh38
rs771398841 Chr.5: 72861959-72861959 on Build GRCh38
rs758143802 Chr.5: 72861936-72861936 on Build GRCh38
rs372984969 Chr.5: 72861770-72861770 on Build GRCh38
rs207466129 Chr.5: 72861753-72861753 on Build GRCh38
rs779595857 Chr.5: 72861794-72861794 on Build GRCh38
rs756066600 Chr.5: 72861919-72861919 on Build GRCh38
rs113568263 Chr.5: 72862209-72862209 on Build GRCh38
rs946711316 Chr.5: 72862160-72862160 on Build GRCh38
rs549802390 Chr.5: 72862113-72862113 on Build GRCh38
rs900409885 Chr.5: 72862107-72862107 on Build GRCh38
rs922475543 Chr.5: 72862082-72862082 on Build GRCh38
rs148233785 Chr.5: 72861949-72861949 on Build GRCh38
rs918718175 Chr.5: 72861994-72861994 on Build GRCh38
rs757277888 Chr.5: 72861777-72861777 on Build GRCh38
rs1024373226 Chr.5: 72862218-72862218 on Build GRCh38
rs370839088 Chr.5: 72862184-72862184 on Build GRCh38
rs762115795 Chr.5: 72861824-72861824 on Build GRCh38
rs776844205 Chr.5: 72861839-72861839 on Build GRCh38

View Full Product Details