Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73578645-73579190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP15
Entrez Gene ID
84135
Gene Name
UTP15, small subunit processome component
Gene Aliases
NET21
UniGene
Hs.406703
Gene Chromosome Location
Chr.5: 73565741-73583377 on Build GRCh38
Transcripts
NM_001284430.1 NM_001284431.1 NM_032175.3 XM_011543680.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193050 c.1160T>A p.I387K Chr.5: 73579030-73579030 on Build GRCh38
6859706 c.1227G>A p.A409A Chr.5: 73579097-73579097 on Build GRCh38
1069948 c.1226C>T p.A409V Chr.5: 73579096-73579096 on Build GRCh38
7232830 c.1212A>C p.G404G Chr.5: 73579082-73579082 on Build GRCh38
7348768 c.1115G>A p.R372Q Chr.5: 73578821-73578821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771002501 Chr.5: 73578704-73578706 on Build GRCh38
rs770890288 Chr.5: 73578892-73578892 on Build GRCh38
rs762217445 Chr.5: 73579076-73579076 on Build GRCh38
rs767983364 Chr.5: 73578991-73578991 on Build GRCh38
rs1046644908 Chr.5: 73579064-73579064 on Build GRCh38
rs919020796 Chr.5: 73578947-73578947 on Build GRCh38
rs764471134 Chr.5: 73578838-73578838 on Build GRCh38
rs575840740 Chr.5: 73579127-73579127 on Build GRCh38
rs146091678 Chr.5: 73579043-73579043 on Build GRCh38
rs949623747 Chr.5: 73579056-73579056 on Build GRCh38
rs746091377 Chr.5: 73578733-73578733 on Build GRCh38
rs986621528 Chr.5: 73578675-73578675 on Build GRCh38
rs769455349 Chr.5: 73579048-73579048 on Build GRCh38
rs773740815 Chr.5: 73579083-73579083 on Build GRCh38
rs781090516 Chr.5: 73579155-73579155 on Build GRCh38
rs189901442 Chr.5: 73578962-73578962 on Build GRCh38
rs146063136 Chr.5: 73578847-73578847 on Build GRCh38
rs779631776 Chr.5: 73579126-73579127 on Build GRCh38
rs779638552 Chr.5: 73579049-73579049 on Build GRCh38
rs949658737 Chr.5: 73578816-73578816 on Build GRCh38
rs375548127 Chr.5: 73579041-73579041 on Build GRCh38
rs343122 Chr.5: 73579082-73579082 on Build GRCh38
rs768087784 Chr.5: 73579128-73579129 on Build GRCh38
rs12652195 Chr.5: 73578976-73578976 on Build GRCh38
rs140195152 Chr.5: 73578820-73578820 on Build GRCh38
rs777561221 Chr.5: 73578882-73578882 on Build GRCh38
rs142301033 Chr.5: 73579040-73579040 on Build GRCh38
rs747910316 Chr.5: 73578751-73578751 on Build GRCh38
rs567464155 Chr.5: 73578700-73578700 on Build GRCh38
rs756209097 Chr.5: 73579000-73579000 on Build GRCh38
rs774719229 Chr.5: 73578988-73578988 on Build GRCh38
rs776891533 Chr.5: 73578952-73578952 on Build GRCh38
rs973270545 Chr.5: 73578933-73578933 on Build GRCh38
rs768340082 Chr.5: 73579072-73579072 on Build GRCh38
rs376105250 Chr.5: 73579107-73579107 on Build GRCh38
rs778757790 Chr.5: 73578975-73578975 on Build GRCh38
rs554252838 Chr.5: 73578925-73578925 on Build GRCh38
rs140164456 Chr.5: 73578869-73578869 on Build GRCh38
rs899271417 Chr.5: 73579161-73579161 on Build GRCh38
rs750486381 Chr.5: 73578862-73578862 on Build GRCh38
rs528682134 Chr.5: 73578768-73578768 on Build GRCh38
rs879082951 Chr.5: 73578871-73578871 on Build GRCh38
rs559859238 Chr.5: 73578737-73578737 on Build GRCh38
rs371751182 Chr.5: 73578837-73578837 on Build GRCh38
rs200550065 Chr.5: 73578974-73578974 on Build GRCh38
rs758732978 Chr.5: 73578885-73578886 on Build GRCh38
rs762626214 Chr.5: 73578978-73578978 on Build GRCh38
rs746818696 Chr.5: 73579024-73579024 on Build GRCh38
rs755174178 Chr.5: 73579021-73579021 on Build GRCh38
rs751356283 Chr.5: 73579117-73579117 on Build GRCh38
rs758045490 Chr.5: 73578980-73578980 on Build GRCh38
rs774742315 Chr.5: 73578882-73578897 on Build GRCh38
rs142119358 Chr.5: 73578759-73578759 on Build GRCh38
rs752805697 Chr.5: 73578881-73578883 on Build GRCh38
rs778262201 Chr.5: 73579155-73579156 on Build GRCh38
rs757301707 Chr.5: 73578729-73578729 on Build GRCh38
rs753943242 Chr.5: 73579101-73579101 on Build GRCh38
rs998042893 Chr.5: 73578685-73578685 on Build GRCh38
rs757017391 Chr.5: 73578897-73578898 on Build GRCh38
rs767504400 Chr.5: 73578852-73578852 on Build GRCh38
rs757712438 Chr.5: 73578851-73578851 on Build GRCh38
rs377331463 Chr.5: 73578830-73578830 on Build GRCh38
rs113620423 Chr.5: 73578984-73578984 on Build GRCh38
rs765465458 Chr.5: 73579108-73579108 on Build GRCh38
rs776224097 Chr.5: 73578812-73578812 on Build GRCh38
rs867664185 Chr.5: 73578776-73578776 on Build GRCh38
rs951414410 Chr.5: 73578911-73578911 on Build GRCh38
rs746925172 Chr.5: 73578789-73578789 on Build GRCh38
rs779678266 Chr.5: 73578757-73578757 on Build GRCh38
rs762283569 Chr.5: 73578989-73578989 on Build GRCh38
rs754067669 Chr.5: 73579017-73579017 on Build GRCh38
rs764190963 Chr.5: 73578893-73578893 on Build GRCh38
rs758159357 Chr.5: 73578879-73578879 on Build GRCh38
rs369328358 Chr.5: 73579054-73579054 on Build GRCh38
rs372847214 Chr.5: 73579074-73579074 on Build GRCh38
rs146995054 Chr.5: 73578821-73578821 on Build GRCh38
rs984489364 Chr.5: 73578966-73578966 on Build GRCh38
rs778137031 Chr.5: 73578889-73578890 on Build GRCh38
rs912395569 Chr.5: 73578678-73578678 on Build GRCh38
rs905324817 Chr.5: 73578823-73578823 on Build GRCh38
rs1053335176 Chr.5: 73579029-73579029 on Build GRCh38
rs769616892 Chr.5: 73578897-73578897 on Build GRCh38
rs1037764608 Chr.5: 73578839-73578839 on Build GRCh38
rs369979994 Chr.5: 73578994-73578994 on Build GRCh38
rs781288085 Chr.5: 73578731-73578731 on Build GRCh38
rs571082114 Chr.5: 73578782-73578782 on Build GRCh38
rs753374817 Chr.5: 73578896-73578896 on Build GRCh38
rs750112243 Chr.5: 73578977-73578977 on Build GRCh38
rs151157740 Chr.5: 73578811-73578811 on Build GRCh38
rs776035513 Chr.5: 73578834-73578834 on Build GRCh38
rs1037862402 Chr.5: 73579147-73579147 on Build GRCh38
rs766406593 Chr.5: 73579004-73579004 on Build GRCh38
rs778748859 Chr.5: 73578747-73578747 on Build GRCh38
rs766550475 Chr.5: 73578878-73578878 on Build GRCh38
rs34701243 Chr.5: 73578678-73578678 on Build GRCh38
rs761103496 Chr.5: 73579096-73579096 on Build GRCh38
rs375576396 Chr.5: 73578887-73578887 on Build GRCh38
rs185424089 Chr.5: 73578735-73578735 on Build GRCh38

View Full Product Details