Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74627318-74627825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENC1
Entrez Gene ID
8507
Gene Name
ectodermal-neural cortex 1
Gene Aliases
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
UniGene
Hs.104925
Gene Chromosome Location
Chr.5: 74627406-74641424 on Build GRCh38
Transcripts
NM_001256574.1 NM_001256575.1 NM_001256576.1 NM_003633.3 NR_046318.1 XM_011543696.2 XM_011543697.2

SNP Detail

SNP ID Chromosome Location
rs963664918 Chr.5: 74627589-74627589 on Build GRCh38
rs531714825 Chr.5: 74627403-74627403 on Build GRCh38
rs933454706 Chr.5: 74627356-74627356 on Build GRCh38
rs920508023 Chr.5: 74627513-74627513 on Build GRCh38
rs980966515 Chr.5: 74627586-74627586 on Build GRCh38
rs1041122313 Chr.5: 74627547-74627548 on Build GRCh38
rs919339586 Chr.5: 74627348-74627348 on Build GRCh38
rs1013968721 Chr.5: 74627561-74627561 on Build GRCh38
rs943214751 Chr.5: 74627467-74627467 on Build GRCh38
rs762052185 Chr.5: 74627525-74627525 on Build GRCh38
rs371470835 Chr.5: 74627653-74627653 on Build GRCh38
rs866009727 Chr.5: 74627546-74627546 on Build GRCh38
rs117574997 Chr.5: 74627587-74627587 on Build GRCh38
rs1035235139 Chr.5: 74627422-74627422 on Build GRCh38
rs565879219 Chr.5: 74627545-74627545 on Build GRCh38
rs750459341 Chr.5: 74627780-74627780 on Build GRCh38
rs895441053 Chr.5: 74627800-74627800 on Build GRCh38
rs972052253 Chr.5: 74627593-74627593 on Build GRCh38
rs969090188 Chr.5: 74627402-74627402 on Build GRCh38
rs1003729911 Chr.5: 74627721-74627723 on Build GRCh38
rs575090517 Chr.5: 74627761-74627761 on Build GRCh38
rs767637110 Chr.5: 74627671-74627671 on Build GRCh38
rs528693219 Chr.5: 74627759-74627759 on Build GRCh38
rs535701473 Chr.5: 74627517-74627517 on Build GRCh38
rs1044731574 Chr.5: 74627689-74627689 on Build GRCh38
rs76287955 Chr.5: 74627574-74627574 on Build GRCh38
rs758184863 Chr.5: 74627549-74627549 on Build GRCh38
rs1052971823 Chr.5: 74627788-74627788 on Build GRCh38
rs962223424 Chr.5: 74627764-74627764 on Build GRCh38
rs181040052 Chr.5: 74627726-74627726 on Build GRCh38
rs117300770 Chr.5: 74627582-74627582 on Build GRCh38
rs904445086 Chr.5: 74627763-74627763 on Build GRCh38
rs73113456 Chr.5: 74627675-74627675 on Build GRCh38
rs10677661 Chr.5: 74627469-74627470 on Build GRCh38
rs565466146 Chr.5: 74627421-74627421 on Build GRCh38
rs570784146 Chr.5: 74627376-74627376 on Build GRCh38
rs934610560 Chr.5: 74627765-74627765 on Build GRCh38
rs894030223 Chr.5: 74627667-74627667 on Build GRCh38
rs895546275 Chr.5: 74627560-74627560 on Build GRCh38
rs935512127 Chr.5: 74627629-74627629 on Build GRCh38
rs184988547 Chr.5: 74627426-74627426 on Build GRCh38
rs113669142 Chr.5: 74627435-74627435 on Build GRCh38
rs893516360 Chr.5: 74627357-74627357 on Build GRCh38
rs910640479 Chr.5: 74627672-74627672 on Build GRCh38
rs922785777 Chr.5: 74627605-74627605 on Build GRCh38
rs933320024 Chr.5: 74627645-74627645 on Build GRCh38
rs1010569227 Chr.5: 74627372-74627372 on Build GRCh38
rs961887650 Chr.5: 74627457-74627457 on Build GRCh38
rs1053152081 Chr.5: 74627636-74627636 on Build GRCh38
rs554050538 Chr.5: 74627529-74627529 on Build GRCh38
rs139423358 Chr.5: 74627516-74627516 on Build GRCh38
rs112035559 Chr.5: 74627456-74627457 on Build GRCh38
rs77357817 Chr.5: 74627468-74627468 on Build GRCh38
rs919231550 Chr.5: 74627604-74627604 on Build GRCh38
rs904959428 Chr.5: 74627694-74627694 on Build GRCh38
rs767006944 Chr.5: 74627472-74627472 on Build GRCh38

View Full Product Details