Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75118028-75118566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
UniGene
Hs.482533
Gene Chromosome Location
Chr.5: 75068297-75236897 on Build GRCh38
Transcripts
NM_001164443.1 XM_011543298.2 XM_011543299.1 XM_011543300.2 XM_011543301.2 XM_011543302.2 XM_017009317.1 XM_017009318.1 XM_017009319.1 XM_017009320.1 XM_017009321.1 XM_017009322.1

SNP Detail

SNP ID Chromosome Location
rs964060115 Chr.5: 75118132-75118132 on Build GRCh38
rs757192880 Chr.5: 75118275-75118275 on Build GRCh38
rs998673057 Chr.5: 75118502-75118502 on Build GRCh38
rs553974640 Chr.5: 75118315-75118315 on Build GRCh38
rs535984320 Chr.5: 75118533-75118533 on Build GRCh38
rs765115154 Chr.5: 75118128-75118128 on Build GRCh38
rs1030068041 Chr.5: 75118201-75118201 on Build GRCh38
rs976788650 Chr.5: 75118144-75118144 on Build GRCh38
rs745667870 Chr.5: 75118371-75118371 on Build GRCh38
rs946880842 Chr.5: 75118299-75118299 on Build GRCh38
rs1007352353 Chr.5: 75118462-75118462 on Build GRCh38
rs755207591 Chr.5: 75118346-75118346 on Build GRCh38
rs1049462910 Chr.5: 75118414-75118414 on Build GRCh38
rs146069313 Chr.5: 75118071-75118071 on Build GRCh38
rs921456137 Chr.5: 75118343-75118343 on Build GRCh38
rs926362467 Chr.5: 75118348-75118348 on Build GRCh38
rs751715157 Chr.5: 75118270-75118270 on Build GRCh38
rs1004590129 Chr.5: 75118112-75118112 on Build GRCh38
rs994558193 Chr.5: 75118090-75118090 on Build GRCh38
rs780195286 Chr.5: 75118382-75118382 on Build GRCh38
rs757822044 Chr.5: 75118247-75118247 on Build GRCh38
rs756864951 Chr.5: 75118281-75118281 on Build GRCh38
rs965686491 Chr.5: 75118176-75118176 on Build GRCh38
rs1018334473 Chr.5: 75118096-75118096 on Build GRCh38
rs780138799 Chr.5: 75118165-75118166 on Build GRCh38
rs764156226 Chr.5: 75118102-75118102 on Build GRCh38
rs756345381 Chr.5: 75118374-75118374 on Build GRCh38
rs1054992767 Chr.5: 75118053-75118053 on Build GRCh38
rs915369027 Chr.5: 75118380-75118380 on Build GRCh38
rs763863106 Chr.5: 75118260-75118260 on Build GRCh38
rs1027324930 Chr.5: 75118098-75118098 on Build GRCh38
rs939048320 Chr.5: 75118368-75118368 on Build GRCh38
rs752238868 Chr.5: 75118161-75118161 on Build GRCh38
rs988778377 Chr.5: 75118217-75118217 on Build GRCh38
rs781699301 Chr.5: 75118303-75118303 on Build GRCh38
rs542441894 Chr.5: 75118528-75118528 on Build GRCh38
rs749378930 Chr.5: 75118483-75118483 on Build GRCh38
rs56030809 Chr.5: 75118437-75118437 on Build GRCh38
rs929269170 Chr.5: 75118392-75118392 on Build GRCh38
rs1019018144 Chr.5: 75118137-75118137 on Build GRCh38
rs894992714 Chr.5: 75118469-75118469 on Build GRCh38
rs1477938 Chr.5: 75118492-75118492 on Build GRCh38
rs927934322 Chr.5: 75118403-75118403 on Build GRCh38
rs138837991 Chr.5: 75118387-75118387 on Build GRCh38
rs950388193 Chr.5: 75118103-75118103 on Build GRCh38
rs994592614 Chr.5: 75118481-75118481 on Build GRCh38
rs751491591 Chr.5: 75118171-75118171 on Build GRCh38
rs79758856 Chr.5: 75118106-75118106 on Build GRCh38
rs957015370 Chr.5: 75118216-75118216 on Build GRCh38
rs558329240 Chr.5: 75118251-75118251 on Build GRCh38
rs1055425793 Chr.5: 75118453-75118453 on Build GRCh38
rs886224473 Chr.5: 75118511-75118511 on Build GRCh38

View Full Product Details