Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 111473182-111473706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK4
Entrez Gene ID
814
Gene Name
calcium/calmodulin dependent protein kinase IV
Gene Aliases
CaMK IV, CaMK-GR, caMK
UniGene
Hs.438801
Gene Chromosome Location
Chr.5: 111223653-111498503 on Build GRCh38
Transcripts
NM_001323374.1 NM_001323375.1 NM_001323376.1 NM_001323377.1 NM_001744.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735100 c.629C>A p.P210H Chr.5: 111473314-111473314 on Build GRCh38
4397661 c.666G>A p.V222V Chr.5: 111473351-111473351 on Build GRCh38
6535658 c.677C>G p.S226C Chr.5: 111473362-111473362 on Build GRCh38
735099 c.648T>C p.C216C Chr.5: 111473333-111473333 on Build GRCh38
237111 c.626-6C>A p.? Chr.5: 111473305-111473305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764347562 Chr.5: 111473293-111473293 on Build GRCh38
rs773405128 Chr.5: 111473433-111473433 on Build GRCh38
rs577338956 Chr.5: 111473580-111473580 on Build GRCh38
rs554720728 Chr.5: 111473497-111473497 on Build GRCh38
rs1013689926 Chr.5: 111473560-111473560 on Build GRCh38
rs1050764929 Chr.5: 111473454-111473454 on Build GRCh38
rs1032338469 Chr.5: 111473605-111473605 on Build GRCh38
rs1046757291 Chr.5: 111473211-111473211 on Build GRCh38
rs542809622 Chr.5: 111473609-111473609 on Build GRCh38
rs958573714 Chr.5: 111473607-111473607 on Build GRCh38
rs565332997 Chr.5: 111473677-111473677 on Build GRCh38
rs143383553 Chr.5: 111473330-111473330 on Build GRCh38
rs1003684341 Chr.5: 111473575-111473575 on Build GRCh38
rs936927357 Chr.5: 111473339-111473339 on Build GRCh38
rs374868830 Chr.5: 111473404-111473404 on Build GRCh38
rs183639401 Chr.5: 111473551-111473551 on Build GRCh38
rs111328166 Chr.5: 111473678-111473678 on Build GRCh38
rs570144274 Chr.5: 111473524-111473524 on Build GRCh38
rs757208276 Chr.5: 111473264-111473264 on Build GRCh38
rs373842366 Chr.5: 111473298-111473298 on Build GRCh38
rs762031529 Chr.5: 111473417-111473417 on Build GRCh38
rs764248560 Chr.5: 111473651-111473651 on Build GRCh38
rs969349265 Chr.5: 111473255-111473255 on Build GRCh38
rs77496684 Chr.5: 111473676-111473676 on Build GRCh38
rs746856638 Chr.5: 111473397-111473397 on Build GRCh38
rs777357216 Chr.5: 111473385-111473385 on Build GRCh38
rs904962786 Chr.5: 111473553-111473553 on Build GRCh38
rs1005639270 Chr.5: 111473545-111473545 on Build GRCh38
rs992240753 Chr.5: 111473223-111473223 on Build GRCh38
rs577266216 Chr.5: 111473219-111473219 on Build GRCh38
rs143574278 Chr.5: 111473598-111473598 on Build GRCh38
rs757216723 Chr.5: 111473396-111473397 on Build GRCh38
rs761841062 Chr.5: 111473302-111473302 on Build GRCh38
rs746818554 Chr.5: 111473267-111473267 on Build GRCh38
rs1022708449 Chr.5: 111473671-111473671 on Build GRCh38
rs752713583 Chr.5: 111473508-111473508 on Build GRCh38
rs747837965 Chr.5: 111473364-111473364 on Build GRCh38
rs951476799 Chr.5: 111473210-111473210 on Build GRCh38
rs766646584 Chr.5: 111473322-111473322 on Build GRCh38
rs757797812 Chr.5: 111473378-111473378 on Build GRCh38
rs988373074 Chr.5: 111473663-111473663 on Build GRCh38
rs772987229 Chr.5: 111473260-111473260 on Build GRCh38
rs988750749 Chr.5: 111473377-111473377 on Build GRCh38
rs745855392 Chr.5: 111473411-111473412 on Build GRCh38
rs148870621 Chr.5: 111473493-111473493 on Build GRCh38
rs929935545 Chr.5: 111473495-111473495 on Build GRCh38
rs988533542 Chr.5: 111473336-111473336 on Build GRCh38
rs1024988573 Chr.5: 111473226-111473226 on Build GRCh38
rs756646007 Chr.5: 111473321-111473321 on Build GRCh38
rs187456406 Chr.5: 111473584-111473584 on Build GRCh38
rs761071617 Chr.5: 111473437-111473437 on Build GRCh38
rs750834121 Chr.5: 111473313-111473313 on Build GRCh38
rs544684230 Chr.5: 111473332-111473332 on Build GRCh38
rs745701813 Chr.5: 111473406-111473406 on Build GRCh38
rs779025974 Chr.5: 111473461-111473462 on Build GRCh38
rs769607894 Chr.5: 111473410-111473410 on Build GRCh38
rs778752233 Chr.5: 111473355-111473355 on Build GRCh38
rs890803914 Chr.5: 111473457-111473457 on Build GRCh38
rs945800316 Chr.5: 111473450-111473450 on Build GRCh38
rs763159981 Chr.5: 111473279-111473279 on Build GRCh38
rs986263547 Chr.5: 111473445-111473445 on Build GRCh38
rs942495649 Chr.5: 111473460-111473460 on Build GRCh38
rs767426524 Chr.5: 111473425-111473425 on Build GRCh38
rs914073535 Chr.5: 111473424-111473424 on Build GRCh38
rs114521245 Chr.5: 111473581-111473581 on Build GRCh38
rs770969921 Chr.5: 111473399-111473399 on Build GRCh38
rs1009861350 Chr.5: 111473615-111473615 on Build GRCh38
rs547721747 Chr.5: 111473265-111473265 on Build GRCh38
rs754657855 Chr.5: 111473341-111473341 on Build GRCh38
rs888154153 Chr.5: 111473521-111473521 on Build GRCh38
rs565903423 Chr.5: 111473306-111473306 on Build GRCh38
rs886907099 Chr.5: 111473552-111473552 on Build GRCh38
rs995428297 Chr.5: 111473511-111473511 on Build GRCh38
rs112730331 Chr.5: 111473412-111473412 on Build GRCh38
rs942345175 Chr.5: 111473485-111473485 on Build GRCh38
rs1024215184 Chr.5: 111473228-111473231 on Build GRCh38
rs781206707 Chr.5: 111473401-111473402 on Build GRCh38
rs767657103 Chr.5: 111473308-111473308 on Build GRCh38
rs888176763 Chr.5: 111473215-111473215 on Build GRCh38
rs55684459 Chr.5: 111473550-111473550 on Build GRCh38

View Full Product Details