Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115602319-115602821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927100
Entrez Gene ID
101927100
Gene Name
uncharacterized LOC101927100
Gene Aliases
-
UniGene
Hs.745340
Gene Chromosome Location
Chr.5: 115602155-115620419 on Build GRCh38
Transcripts
NR_109874.1 NR_109875.1 NM_021649.7 NM_001164468.3 NM_001164469.3

Gene Symbol
TICAM2
Entrez Gene ID
353376
Gene Name
toll like receptor adaptor molecule 2
Gene Aliases
MyD88-4, TICAM-2, TIRAP3, TIRP, TRAM
UniGene
Hs.718838
Gene Chromosome Location
Chr.5: 115578496-115602479 on Build GRCh38
Transcripts
NR_109874.1 NR_109875.1 NM_021649.7 NM_001164468.3 NM_001164469.3

Gene Symbol
TMED7-TICAM2
Entrez Gene ID
100302736
Gene Name
TMED7-TICAM2 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 115578496-115626179 on Build GRCh38
Transcripts
NR_109874.1 NR_109875.1 NM_021649.7 NM_001164468.3 NM_001164469.3

SNP Detail

SNP ID Chromosome Location
rs555678493 Chr.5: 115602697-115602697 on Build GRCh38
rs771696827 Chr.5: 115602584-115602588 on Build GRCh38
rs772239600 Chr.5: 115602693-115602693 on Build GRCh38
rs923389749 Chr.5: 115602524-115602524 on Build GRCh38
rs947065809 Chr.5: 115602660-115602660 on Build GRCh38
rs73254595 Chr.5: 115602663-115602663 on Build GRCh38
rs916604455 Chr.5: 115602633-115602633 on Build GRCh38
rs980720141 Chr.5: 115602541-115602541 on Build GRCh38
rs1050744670 Chr.5: 115602754-115602754 on Build GRCh38
rs1028664165 Chr.5: 115602460-115602461 on Build GRCh38
rs991439916 Chr.5: 115602593-115602593 on Build GRCh38
rs71581203 Chr.5: 115602634-115602634 on Build GRCh38
rs550822281 Chr.5: 115602391-115602391 on Build GRCh38
rs1006237063 Chr.5: 115602794-115602794 on Build GRCh38
rs570942084 Chr.5: 115602396-115602396 on Build GRCh38
rs1047827764 Chr.5: 115602763-115602763 on Build GRCh38
rs950760425 Chr.5: 115602453-115602453 on Build GRCh38
rs748042775 Chr.5: 115602711-115602711 on Build GRCh38
rs915040561 Chr.5: 115602742-115602742 on Build GRCh38
rs886537004 Chr.5: 115602784-115602784 on Build GRCh38
rs988598344 Chr.5: 115602567-115602567 on Build GRCh38
rs539913496 Chr.5: 115602397-115602397 on Build GRCh38
rs537037210 Chr.5: 115602799-115602799 on Build GRCh38
rs547219925 Chr.5: 115602536-115602536 on Build GRCh38
rs1006163324 Chr.5: 115602478-115602478 on Build GRCh38
rs1009005240 Chr.5: 115602759-115602759 on Build GRCh38
rs912967273 Chr.5: 115602576-115602576 on Build GRCh38
rs890367764 Chr.5: 115602755-115602755 on Build GRCh38
rs1016187664 Chr.5: 115602515-115602515 on Build GRCh38
rs1053581955 Chr.5: 115602725-115602725 on Build GRCh38
rs67428503 Chr.5: 115602600-115602604 on Build GRCh38
rs970351503 Chr.5: 115602522-115602522 on Build GRCh38
rs552905710 Chr.5: 115602718-115602718 on Build GRCh38
rs187359775 Chr.5: 115602561-115602561 on Build GRCh38
rs961773897 Chr.5: 115602550-115602550 on Build GRCh38
rs935415162 Chr.5: 115602703-115602703 on Build GRCh38
rs1008663837 Chr.5: 115602534-115602534 on Build GRCh38
rs746072754 Chr.5: 115602766-115602766 on Build GRCh38
rs773460655 Chr.5: 115602585-115602588 on Build GRCh38
rs1035870381 Chr.5: 115602546-115602546 on Build GRCh38
rs952999916 Chr.5: 115602563-115602563 on Build GRCh38
rs1041328242 Chr.5: 115602719-115602719 on Build GRCh38
rs771183930 Chr.5: 115602583-115602588 on Build GRCh38
rs996487517 Chr.5: 115602400-115602400 on Build GRCh38
rs769857294 Chr.5: 115602762-115602762 on Build GRCh38
rs1017550791 Chr.5: 115602693-115602707 on Build GRCh38
rs1053531007 Chr.5: 115602379-115602379 on Build GRCh38
rs530930334 Chr.5: 115602376-115602376 on Build GRCh38
rs997620409 Chr.5: 115602352-115602352 on Build GRCh38
rs901042658 Chr.5: 115602748-115602748 on Build GRCh38
rs890076119 Chr.5: 115602530-115602530 on Build GRCh38
rs536197467 Chr.5: 115602650-115602650 on Build GRCh38

View Full Product Details