Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115625787-115626290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMED7
Entrez Gene ID
51014
Gene Name
transmembrane p24 trafficking protein 7
Gene Aliases
CGI-109, p24g3, p24gamma3, p27
UniGene
Hs.642817
Gene Chromosome Location
Chr.5: 115613208-115626179 on Build GRCh38
Transcripts
NM_181836.5 NM_001164468.3 NM_001164469.3

Gene Symbol
TMED7-TICAM2
Entrez Gene ID
100302736
Gene Name
TMED7-TICAM2 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 115578496-115626179 on Build GRCh38
Transcripts
NM_181836.5 NM_001164468.3 NM_001164469.3

SNP Detail

SNP ID Chromosome Location
rs912318700 Chr.5: 115626114-115626114 on Build GRCh38
rs1047269475 Chr.5: 115625962-115625962 on Build GRCh38
rs982732582 Chr.5: 115626252-115626252 on Build GRCh38
rs778025165 Chr.5: 115626085-115626085 on Build GRCh38
rs1049558833 Chr.5: 115625881-115625881 on Build GRCh38
rs929991834 Chr.5: 115626249-115626249 on Build GRCh38
rs1011375011 Chr.5: 115626256-115626256 on Build GRCh38
rs988014423 Chr.5: 115625916-115625916 on Build GRCh38
rs776421972 Chr.5: 115626161-115626161 on Build GRCh38
rs546856277 Chr.5: 115626211-115626211 on Build GRCh38
rs901407835 Chr.5: 115625804-115625804 on Build GRCh38
rs754199592 Chr.5: 115625935-115625935 on Build GRCh38
rs948751078 Chr.5: 115626160-115626160 on Build GRCh38
rs564230101 Chr.5: 115626162-115626162 on Build GRCh38
rs1011947595 Chr.5: 115626074-115626074 on Build GRCh38
rs1047320498 Chr.5: 115626117-115626117 on Build GRCh38
rs988065067 Chr.5: 115626010-115626010 on Build GRCh38
rs574038722 Chr.5: 115625933-115625933 on Build GRCh38
rs1046248983 Chr.5: 115626003-115626003 on Build GRCh38
rs969098139 Chr.5: 115625862-115625862 on Build GRCh38
rs1053130555 Chr.5: 115626201-115626201 on Build GRCh38
rs967944915 Chr.5: 115626207-115626207 on Build GRCh38
rs748304955 Chr.5: 115626119-115626119 on Build GRCh38
rs986454699 Chr.5: 115626109-115626109 on Build GRCh38
rs892976540 Chr.5: 115626209-115626209 on Build GRCh38
rs1028672524 Chr.5: 115626260-115626260 on Build GRCh38
rs953735792 Chr.5: 115625951-115625951 on Build GRCh38
rs748215915 Chr.5: 115625975-115625975 on Build GRCh38
rs944869002 Chr.5: 115625961-115625961 on Build GRCh38
rs952497982 Chr.5: 115625843-115625843 on Build GRCh38
rs901464612 Chr.5: 115625958-115625958 on Build GRCh38
rs183064070 Chr.5: 115625876-115625876 on Build GRCh38
rs922600008 Chr.5: 115626104-115626104 on Build GRCh38
rs866578561 Chr.5: 115626014-115626014 on Build GRCh38
rs1023300416 Chr.5: 115625861-115625861 on Build GRCh38
rs542073900 Chr.5: 115625878-115625878 on Build GRCh38
rs781115447 Chr.5: 115626061-115626061 on Build GRCh38
rs1035097378 Chr.5: 115625997-115625997 on Build GRCh38
rs916128905 Chr.5: 115626158-115626158 on Build GRCh38
rs977241678 Chr.5: 115626240-115626240 on Build GRCh38
rs1003235435 Chr.5: 115625991-115625991 on Build GRCh38
rs892141559 Chr.5: 115626157-115626157 on Build GRCh38
rs972462706 Chr.5: 115626132-115626132 on Build GRCh38
rs73260574 Chr.5: 115625923-115625923 on Build GRCh38
rs1049777991 Chr.5: 115626154-115626154 on Build GRCh38
rs1023394724 Chr.5: 115625986-115625986 on Build GRCh38
rs1045757232 Chr.5: 115626163-115626163 on Build GRCh38
rs907201357 Chr.5: 115626175-115626175 on Build GRCh38
rs770740142 Chr.5: 115626139-115626139 on Build GRCh38
rs894461460 Chr.5: 115625890-115625890 on Build GRCh38
rs986019633 Chr.5: 115626208-115626208 on Build GRCh38
rs2546472 Chr.5: 115625946-115625946 on Build GRCh38
rs967892524 Chr.5: 115626023-115626023 on Build GRCh38
rs756616313 Chr.5: 115625832-115625832 on Build GRCh38
rs890700522 Chr.5: 115626265-115626265 on Build GRCh38
rs2546473 Chr.5: 115625959-115625959 on Build GRCh38
rs1005265020 Chr.5: 115625844-115625844 on Build GRCh38
rs557156758 Chr.5: 115625943-115625943 on Build GRCh38
rs558245464 Chr.5: 115625867-115625867 on Build GRCh38
rs935685262 Chr.5: 115625809-115625809 on Build GRCh38
rs971638000 Chr.5: 115625928-115625928 on Build GRCh38
rs546067028 Chr.5: 115626020-115626020 on Build GRCh38
rs922687133 Chr.5: 115626243-115626243 on Build GRCh38
rs1043165708 Chr.5: 115625938-115625938 on Build GRCh38
rs890652641 Chr.5: 115625850-115625850 on Build GRCh38
rs934781495 Chr.5: 115626179-115626179 on Build GRCh38
rs956644976 Chr.5: 115626002-115626002 on Build GRCh38
rs998619370 Chr.5: 115625930-115625930 on Build GRCh38
rs774302793 Chr.5: 115626150-115626150 on Build GRCh38
rs888668421 Chr.5: 115626121-115626121 on Build GRCh38
rs907275123 Chr.5: 115626032-115626032 on Build GRCh38
rs115927880 Chr.5: 115625820-115625820 on Build GRCh38
rs1003697014 Chr.5: 115626144-115626144 on Build GRCh38
rs888614507 Chr.5: 115625968-115625968 on Build GRCh38
rs771071020 Chr.5: 115626131-115626131 on Build GRCh38
rs147760720 Chr.5: 115626252-115626253 on Build GRCh38
rs377211306 Chr.5: 115625807-115625807 on Build GRCh38
rs944822982 Chr.5: 115625860-115625860 on Build GRCh38
rs929939631 Chr.5: 115626116-115626116 on Build GRCh38
rs533012765 Chr.5: 115626169-115626169 on Build GRCh38
rs200389655 Chr.5: 115626159-115626159 on Build GRCh38
rs1049309000 Chr.5: 115625979-115625979 on Build GRCh38
rs956818577 Chr.5: 115625903-115625903 on Build GRCh38
rs747170684 Chr.5: 115625990-115625990 on Build GRCh38
rs965869556 Chr.5: 115626223-115626223 on Build GRCh38
rs894504412 Chr.5: 115625987-115625987 on Build GRCh38
rs755465288 Chr.5: 115625954-115625954 on Build GRCh38
rs989744276 Chr.5: 115625810-115625810 on Build GRCh38
rs1034741574 Chr.5: 115626151-115626151 on Build GRCh38
rs575164956 Chr.5: 115626069-115626069 on Build GRCh38
rs747225809 Chr.5: 115626128-115626128 on Build GRCh38
rs911273863 Chr.5: 115625940-115625940 on Build GRCh38
rs904981738 Chr.5: 115626142-115626142 on Build GRCh38
rs564236403 Chr.5: 115626055-115626055 on Build GRCh38
rs998879569 Chr.5: 115626040-115626040 on Build GRCh38
rs1025765324 Chr.5: 115625837-115625837 on Build GRCh38

View Full Product Details