Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 129026945-129027452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A6
Entrez Gene ID
28965
Gene Name
solute carrier family 27 member 6
Gene Aliases
ACSVL2, FACVL2, FATP6, VLCS-H1
UniGene
Hs.49765
Gene Chromosome Location
Chr.5: 128965127-129033642 on Build GRCh38
Transcripts
NM_001017372.2 NM_001317984.1 NM_014031.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400457 c.1388G>A p.G463E Chr.5: 129027265-129027265 on Build GRCh38
4155439 c.1454+10G>T p.? Chr.5: 129027341-129027341 on Build GRCh38
224914 c.1297C>T p.P433S Chr.5: 129027174-129027174 on Build GRCh38
6168761 c.1256G>T p.G419V Chr.5: 129027133-129027133 on Build GRCh38
5633373 c.1437T>C p.R479R Chr.5: 129027314-129027314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753668750 Chr.5: 129027150-129027150 on Build GRCh38
rs914202281 Chr.5: 129027363-129027363 on Build GRCh38
rs754916870 Chr.5: 129027157-129027157 on Build GRCh38
rs752614803 Chr.5: 129027309-129027309 on Build GRCh38
rs780799756 Chr.5: 129027321-129027321 on Build GRCh38
rs901584962 Chr.5: 129027408-129027408 on Build GRCh38
rs772919130 Chr.5: 129027240-129027240 on Build GRCh38
rs772208898 Chr.5: 129027254-129027254 on Build GRCh38
rs749635811 Chr.5: 129027324-129027324 on Build GRCh38
rs772689663 Chr.5: 129027341-129027341 on Build GRCh38
rs774344869 Chr.5: 129027131-129027131 on Build GRCh38
rs752172866 Chr.5: 129027164-129027164 on Build GRCh38
rs755327509 Chr.5: 129027329-129027329 on Build GRCh38
rs149591099 Chr.5: 129027237-129027237 on Build GRCh38
rs374974700 Chr.5: 129027138-129027138 on Build GRCh38
rs761031404 Chr.5: 129027262-129027262 on Build GRCh38
rs185013594 Chr.5: 129027409-129027409 on Build GRCh38
rs375113354 Chr.5: 129027381-129027381 on Build GRCh38
rs749633287 Chr.5: 129027215-129027215 on Build GRCh38
rs143634523 Chr.5: 129027077-129027077 on Build GRCh38
rs972852564 Chr.5: 129027279-129027279 on Build GRCh38
rs927553232 Chr.5: 129027334-129027334 on Build GRCh38
rs1034714403 Chr.5: 129027074-129027074 on Build GRCh38
rs776621170 Chr.5: 129027277-129027277 on Build GRCh38
rs567124528 Chr.5: 129027116-129027118 on Build GRCh38
rs777362377 Chr.5: 129027117-129027117 on Build GRCh38
rs35186587 Chr.5: 129027171-129027171 on Build GRCh38
rs776875423 Chr.5: 129027368-129027368 on Build GRCh38
rs766927880 Chr.5: 129027166-129027167 on Build GRCh38
rs201584963 Chr.5: 129027118-129027118 on Build GRCh38
rs971584442 Chr.5: 129027296-129027296 on Build GRCh38
rs945711353 Chr.5: 129027387-129027387 on Build GRCh38
rs766682242 Chr.5: 129027269-129027269 on Build GRCh38
rs748627713 Chr.5: 129027335-129027335 on Build GRCh38
rs892529206 Chr.5: 129027238-129027238 on Build GRCh38
rs1016902252 Chr.5: 129027194-129027194 on Build GRCh38
rs769576556 Chr.5: 129027376-129027376 on Build GRCh38
rs201176637 Chr.5: 129027199-129027199 on Build GRCh38
rs191411601 Chr.5: 129027091-129027091 on Build GRCh38
rs778340575 Chr.5: 129027103-129027103 on Build GRCh38
rs371613205 Chr.5: 129027119-129027119 on Build GRCh38
rs754525332 Chr.5: 129027102-129027102 on Build GRCh38
rs757861120 Chr.5: 129027312-129027312 on Build GRCh38
rs769090885 Chr.5: 129027216-129027216 on Build GRCh38
rs904398516 Chr.5: 129027053-129027053 on Build GRCh38
rs773522500 Chr.5: 129027143-129027143 on Build GRCh38
rs756942467 Chr.5: 129027320-129027320 on Build GRCh38
rs922869862 Chr.5: 129027196-129027196 on Build GRCh38
rs147208886 Chr.5: 129027125-129027125 on Build GRCh38
rs748205675 Chr.5: 129027250-129027250 on Build GRCh38
rs780166569 Chr.5: 129027090-129027090 on Build GRCh38
rs771660017 Chr.5: 129027114-129027114 on Build GRCh38
rs373485141 Chr.5: 129027205-129027205 on Build GRCh38
rs370861929 Chr.5: 129027403-129027403 on Build GRCh38
rs549901731 Chr.5: 129027148-129027148 on Build GRCh38
rs773577960 Chr.5: 129027257-129027257 on Build GRCh38
rs759330024 Chr.5: 129027289-129027289 on Build GRCh38
rs200215777 Chr.5: 129027242-129027242 on Build GRCh38
rs1002639481 Chr.5: 129027325-129027325 on Build GRCh38
rs1044084003 Chr.5: 129027293-129027293 on Build GRCh38
rs769192760 Chr.5: 129027129-129027129 on Build GRCh38
rs376654449 Chr.5: 129027249-129027249 on Build GRCh38
rs777408066 Chr.5: 129027173-129027173 on Build GRCh38
rs1022313063 Chr.5: 129027052-129027052 on Build GRCh38
rs1054535166 Chr.5: 129027360-129027360 on Build GRCh38
rs918667274 Chr.5: 129027291-129027291 on Build GRCh38
rs567400175 Chr.5: 129027042-129027042 on Build GRCh38
rs780251899 Chr.5: 129027232-129027232 on Build GRCh38
rs781274585 Chr.5: 129026973-129026973 on Build GRCh38
rs541437624 Chr.5: 129027313-129027313 on Build GRCh38
rs779289426 Chr.5: 129027332-129027332 on Build GRCh38
rs766213587 Chr.5: 129027149-129027149 on Build GRCh38
rs778044831 Chr.5: 129027348-129027348 on Build GRCh38
rs760350156 Chr.5: 129027313-129027314 on Build GRCh38
rs771303633 Chr.5: 129027365-129027365 on Build GRCh38
rs892581552 Chr.5: 129026992-129026992 on Build GRCh38
rs747067639 Chr.5: 129027356-129027356 on Build GRCh38
rs2304578 Chr.5: 129027137-129027137 on Build GRCh38
rs950194682 Chr.5: 129027046-129027046 on Build GRCh38
rs559128144 Chr.5: 129027417-129027417 on Build GRCh38
rs867556415 Chr.5: 129027282-129027282 on Build GRCh38
rs765120068 Chr.5: 129027306-129027306 on Build GRCh38
rs747669070 Chr.5: 129027107-129027107 on Build GRCh38
rs368988238 Chr.5: 129027182-129027182 on Build GRCh38
rs369872344 Chr.5: 129027370-129027370 on Build GRCh38
rs767665885 Chr.5: 129027140-129027140 on Build GRCh38
rs186948046 Chr.5: 129026982-129026982 on Build GRCh38
rs745891985 Chr.5: 129027080-129027080 on Build GRCh38
rs1001467122 Chr.5: 129027073-129027073 on Build GRCh38

View Full Product Details