Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131641621-131642165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
UniGene
Hs.591273
Gene Chromosome Location
Chr.5: 131641714-131797063 on Build GRCh38
Transcripts
NM_001008738.2 NM_133372.2

SNP Detail

SNP ID Chromosome Location
rs148192803 Chr.5: 131641865-131641865 on Build GRCh38
rs982591823 Chr.5: 131641690-131641690 on Build GRCh38
rs995008517 Chr.5: 131641674-131641674 on Build GRCh38
rs958024501 Chr.5: 131641844-131641844 on Build GRCh38
rs893646263 Chr.5: 131642103-131642103 on Build GRCh38
rs899391877 Chr.5: 131641670-131641670 on Build GRCh38
rs141219047 Chr.5: 131642092-131642092 on Build GRCh38
rs181510352 Chr.5: 131641962-131641962 on Build GRCh38
rs978642661 Chr.5: 131641784-131641784 on Build GRCh38
rs754795170 Chr.5: 131641775-131641775 on Build GRCh38
rs534113335 Chr.5: 131641798-131641798 on Build GRCh38
rs1051887589 Chr.5: 131641896-131641896 on Build GRCh38
rs780321637 Chr.5: 131641921-131641921 on Build GRCh38
rs965074585 Chr.5: 131641883-131641883 on Build GRCh38
rs943030577 Chr.5: 131641992-131642014 on Build GRCh38
rs895850806 Chr.5: 131641678-131641678 on Build GRCh38
rs1024900588 Chr.5: 131641686-131641686 on Build GRCh38
rs915845592 Chr.5: 131641760-131641760 on Build GRCh38
rs568185234 Chr.5: 131642125-131642126 on Build GRCh38
rs1035829562 Chr.5: 131641711-131641711 on Build GRCh38
rs947265575 Chr.5: 131641777-131641777 on Build GRCh38
rs899424566 Chr.5: 131641989-131641989 on Build GRCh38
rs1014276921 Chr.5: 131642016-131642016 on Build GRCh38
rs879114236 Chr.5: 131642000-131642000 on Build GRCh38
rs201208820 Chr.5: 131641807-131641807 on Build GRCh38
rs921533798 Chr.5: 131641900-131641900 on Build GRCh38
rs950939638 Chr.5: 131641680-131641680 on Build GRCh38
rs959760898 Chr.5: 131641726-131641726 on Build GRCh38
rs995120068 Chr.5: 131642082-131642082 on Build GRCh38
rs886632481 Chr.5: 131642134-131642134 on Build GRCh38
rs1032579896 Chr.5: 131642098-131642099 on Build GRCh38
rs895766450 Chr.5: 131642015-131642015 on Build GRCh38
rs1044443916 Chr.5: 131642079-131642079 on Build GRCh38
rs934472776 Chr.5: 131641894-131641894 on Build GRCh38
rs911935926 Chr.5: 131641937-131641937 on Build GRCh38
rs769032392 Chr.5: 131641763-131641763 on Build GRCh38
rs991083668 Chr.5: 131641739-131641739 on Build GRCh38
rs568342712 Chr.5: 131641675-131641675 on Build GRCh38
rs932935337 Chr.5: 131641901-131641901 on Build GRCh38
rs748695308 Chr.5: 131641950-131641950 on Build GRCh38
rs143048434 Chr.5: 131641812-131641812 on Build GRCh38

View Full Product Details