Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132758157-132758698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132750817-132778216 on Build GRCh38
Transcripts
NM_001098811.1 NM_001098812.1 NM_001098813.1 NM_001300798.1 NM_001300799.1 NM_015146.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1 XM_017009258.1 XM_017009259.1 XM_017009260.1 XM_017009261.1

SNP Detail

SNP ID Chromosome Location
rs756356897 Chr.5: 132758512-132758512 on Build GRCh38
rs747681296 Chr.5: 132758463-132758463 on Build GRCh38
rs1005592048 Chr.5: 132758489-132758489 on Build GRCh38
rs1036677688 Chr.5: 132758672-132758672 on Build GRCh38
rs1028598533 Chr.5: 132758244-132758244 on Build GRCh38
rs922724475 Chr.5: 132758639-132758639 on Build GRCh38
rs958269171 Chr.5: 132758364-132758364 on Build GRCh38
rs775788652 Chr.5: 132758476-132758476 on Build GRCh38
rs764642987 Chr.5: 132758488-132758488 on Build GRCh38
rs913916463 Chr.5: 132758609-132758609 on Build GRCh38
rs116236254 Chr.5: 132758664-132758664 on Build GRCh38
rs1015757858 Chr.5: 132758294-132758294 on Build GRCh38
rs775884538 Chr.5: 132758537-132758537 on Build GRCh38
rs141229787 Chr.5: 132758260-132758260 on Build GRCh38
rs754448107 Chr.5: 132758459-132758459 on Build GRCh38
rs932811532 Chr.5: 132758642-132758642 on Build GRCh38
rs1005252640 Chr.5: 132758290-132758290 on Build GRCh38
rs760259103 Chr.5: 132758475-132758475 on Build GRCh38
rs377670560 Chr.5: 132758496-132758496 on Build GRCh38
rs371815283 Chr.5: 132758522-132758522 on Build GRCh38
rs961171553 Chr.5: 132758203-132758203 on Build GRCh38
rs376081226 Chr.5: 132758443-132758443 on Build GRCh38
rs73787063 Chr.5: 132758411-132758411 on Build GRCh38
rs755941164 Chr.5: 132758295-132758295 on Build GRCh38
rs754805472 Chr.5: 132758598-132758598 on Build GRCh38
rs960956288 Chr.5: 132758471-132758471 on Build GRCh38
rs990966323 Chr.5: 132758257-132758257 on Build GRCh38
rs763413094 Chr.5: 132758477-132758477 on Build GRCh38
rs774927929 Chr.5: 132758575-132758575 on Build GRCh38
rs1030728705 Chr.5: 132758510-132758510 on Build GRCh38
rs34160017 Chr.5: 132758656-132758657 on Build GRCh38
rs771580046 Chr.5: 132758527-132758527 on Build GRCh38
rs566040915 Chr.5: 132758427-132758427 on Build GRCh38
rs967817884 Chr.5: 132758278-132758278 on Build GRCh38
rs778905137 Chr.5: 132758517-132758517 on Build GRCh38
rs761025294 Chr.5: 132758589-132758589 on Build GRCh38
rs42403 Chr.5: 132758302-132758302 on Build GRCh38
rs115784309 Chr.5: 132758643-132758643 on Build GRCh38
rs750491567 Chr.5: 132758584-132758584 on Build GRCh38
rs535469405 Chr.5: 132758392-132758392 on Build GRCh38
rs754926113 Chr.5: 132758516-132758516 on Build GRCh38
rs767730032 Chr.5: 132758583-132758583 on Build GRCh38
rs200179947 Chr.5: 132758593-132758593 on Build GRCh38
rs78253221 Chr.5: 132758313-132758313 on Build GRCh38
rs767083820 Chr.5: 132758487-132758488 on Build GRCh38
rs370098786 Chr.5: 132758608-132758608 on Build GRCh38
rs769113406 Chr.5: 132758574-132758574 on Build GRCh38
rs942530938 Chr.5: 132758373-132758373 on Build GRCh38
rs182822481 Chr.5: 132758528-132758528 on Build GRCh38
rs371784308 Chr.5: 132758413-132758413 on Build GRCh38
rs557556946 Chr.5: 132758497-132758497 on Build GRCh38
rs913947483 Chr.5: 132758380-132758380 on Build GRCh38
rs559262204 Chr.5: 132758634-132758634 on Build GRCh38
rs766692872 Chr.5: 132758592-132758592 on Build GRCh38
rs370894893 Chr.5: 132758501-132758501 on Build GRCh38
rs17716311 Chr.5: 132758513-132758513 on Build GRCh38
rs528142897 Chr.5: 132758182-132758182 on Build GRCh38
rs11538383 Chr.5: 132758457-132758457 on Build GRCh38
rs548018527 Chr.5: 132758252-132758252 on Build GRCh38
rs761303590 Chr.5: 132758480-132758480 on Build GRCh38
rs942624064 Chr.5: 132758552-132758552 on Build GRCh38
rs930755969 Chr.5: 132758597-132758597 on Build GRCh38
rs374823905 Chr.5: 132758502-132758502 on Build GRCh38
rs1035686307 Chr.5: 132758470-132758470 on Build GRCh38
rs979164372 Chr.5: 132758507-132758507 on Build GRCh38
rs202034103 Chr.5: 132758543-132758543 on Build GRCh38
rs570064454 Chr.5: 132758337-132758337 on Build GRCh38
rs1014064452 Chr.5: 132758298-132758298 on Build GRCh38
rs746692668 Chr.5: 132758531-132758531 on Build GRCh38
rs1050423061 Chr.5: 132758452-132758452 on Build GRCh38
rs940705801 Chr.5: 132758622-132758622 on Build GRCh38
rs980406122 Chr.5: 132758335-132758335 on Build GRCh38
rs534762956 Chr.5: 132758432-132758432 on Build GRCh38
rs989534540 Chr.5: 132758369-132758369 on Build GRCh38
rs997067538 Chr.5: 132758242-132758242 on Build GRCh38
rs373829311 Chr.5: 132758547-132758547 on Build GRCh38
rs1024086655 Chr.5: 132758504-132758504 on Build GRCh38
rs57510773 Chr.5: 132758665-132758665 on Build GRCh38
rs772852321 Chr.5: 132758468-132758468 on Build GRCh38
rs912458032 Chr.5: 132758350-132758350 on Build GRCh38
rs771688591 Chr.5: 132758466-132758466 on Build GRCh38
rs778510246 Chr.5: 132758601-132758601 on Build GRCh38
rs932850103 Chr.5: 132758451-132758451 on Build GRCh38
rs770543424 Chr.5: 132758533-132758533 on Build GRCh38
rs2286012 Chr.5: 132758605-132758605 on Build GRCh38
rs1012483496 Chr.5: 132758641-132758641 on Build GRCh38
rs542261704 Chr.5: 132758633-132758633 on Build GRCh38

View Full Product Details