Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134564352-134564865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JADE2
Entrez Gene ID
23338
Gene Name
jade family PHD finger 2
Gene Aliases
JADE-2, PHF15
UniGene
Hs.732155
Gene Chromosome Location
Chr.5: 134524154-134583230 on Build GRCh38
Transcripts
NM_001289984.1 NM_001289985.1 NM_001308143.1 NM_015288.5 XM_005271945.1 XM_005271946.1 XM_005271948.4 XM_011543291.2 XM_017009282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1433178 c.955G>T p.G319C Chr.5: 134564596-134564596 on Build GRCh38
6346861 c.1017+7G>T p.? Chr.5: 134564617-134564617 on Build GRCh38
5884101 c.921G>T p.W307C Chr.5: 134564562-134564562 on Build GRCh38
5175842 c.1003G>T p.G335C Chr.5: 134564596-134564596 on Build GRCh38
6346860 c.969+7G>T p.? Chr.5: 134564617-134564617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370730516 Chr.5: 134564541-134564541 on Build GRCh38
rs778566596 Chr.5: 134564631-134564631 on Build GRCh38
rs187609910 Chr.5: 134564762-134564762 on Build GRCh38
rs750381388 Chr.5: 134564566-134564566 on Build GRCh38
rs778443943 Chr.5: 134564481-134564481 on Build GRCh38
rs770195087 Chr.5: 134564513-134564513 on Build GRCh38
rs763319586 Chr.5: 134564540-134564540 on Build GRCh38
rs898553851 Chr.5: 134564735-134564735 on Build GRCh38
rs768140777 Chr.5: 134564479-134564479 on Build GRCh38
rs908531537 Chr.5: 134564645-134564645 on Build GRCh38
rs528717535 Chr.5: 134564733-134564733 on Build GRCh38
rs780528985 Chr.5: 134564796-134564796 on Build GRCh38
rs372899480 Chr.5: 134564595-134564595 on Build GRCh38
rs987008863 Chr.5: 134564666-134564666 on Build GRCh38
rs329311 Chr.5: 134564421-134564421 on Build GRCh38
rs754664917 Chr.5: 134564624-134564624 on Build GRCh38
rs13179411 Chr.5: 134564823-134564823 on Build GRCh38
rs867123350 Chr.5: 134564578-134564578 on Build GRCh38
rs34077769 Chr.5: 134564734-134564734 on Build GRCh38
rs762010172 Chr.5: 134564552-134564552 on Build GRCh38
rs377573373 Chr.5: 134564443-134564443 on Build GRCh38
rs771385739 Chr.5: 134564500-134564500 on Build GRCh38
rs115546030 Chr.5: 134564462-134564462 on Build GRCh38
rs779958194 Chr.5: 134564592-134564592 on Build GRCh38
rs759759388 Chr.5: 134564503-134564503 on Build GRCh38
rs558599535 Chr.5: 134564420-134564420 on Build GRCh38
rs767301717 Chr.5: 134564553-134564553 on Build GRCh38
rs750874089 Chr.5: 134564761-134564761 on Build GRCh38
rs200513701 Chr.5: 134564546-134564546 on Build GRCh38
rs1038497098 Chr.5: 134564374-134564374 on Build GRCh38
rs930406169 Chr.5: 134564414-134564414 on Build GRCh38
rs867392774 Chr.5: 134564597-134564597 on Build GRCh38
rs200129793 Chr.5: 134564475-134564475 on Build GRCh38
rs564149779 Chr.5: 134564694-134564694 on Build GRCh38
rs964298644 Chr.5: 134564736-134564736 on Build GRCh38
rs369875016 Chr.5: 134564446-134564446 on Build GRCh38
rs940030991 Chr.5: 134564716-134564716 on Build GRCh38
rs34150573 Chr.5: 134564502-134564502 on Build GRCh38
rs756247089 Chr.5: 134564480-134564480 on Build GRCh38
rs967788128 Chr.5: 134564570-134564570 on Build GRCh38
rs1003456344 Chr.5: 134564819-134564819 on Build GRCh38
rs749036785 Chr.5: 134564478-134564478 on Build GRCh38
rs747481560 Chr.5: 134564487-134564487 on Build GRCh38
rs537012559 Chr.5: 134564384-134564384 on Build GRCh38
rs769236118 Chr.5: 134564820-134564820 on Build GRCh38
rs987837869 Chr.5: 134564548-134564548 on Build GRCh38
rs755857800 Chr.5: 134564576-134564576 on Build GRCh38
rs540910642 Chr.5: 134564743-134564743 on Build GRCh38
rs202020260 Chr.5: 134564538-134564538 on Build GRCh38
rs114626647 Chr.5: 134564778-134564778 on Build GRCh38
rs1038443059 Chr.5: 134564726-134564726 on Build GRCh38
rs530628745 Chr.5: 134564841-134564841 on Build GRCh38
rs912697917 Chr.5: 134564697-134564697 on Build GRCh38
rs755952120 Chr.5: 134564464-134564464 on Build GRCh38
rs201054669 Chr.5: 134564614-134564614 on Build GRCh38
rs329312 Chr.5: 134564808-134564808 on Build GRCh38
rs1048825723 Chr.5: 134564418-134564418 on Build GRCh38

View Full Product Details