Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138149415-138149917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_011543109.1 XM_011543110.1 XM_017008969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850366 c.3130C>T p.Q1044* Chr.5: 138149788-138149788 on Build GRCh38
4924490 c.3125A>G p.E1042G Chr.5: 138149793-138149793 on Build GRCh38
6044531 c.3124delG p.E1042fs*52 Chr.5: 138149794-138149794 on Build GRCh38
1433314 c.3202G>A p.E1068K Chr.5: 138149716-138149716 on Build GRCh38
3409755 c.3124G>A p.E1042K Chr.5: 138149794-138149794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776051190 Chr.5: 138149746-138149747 on Build GRCh38
rs143885755 Chr.5: 138149795-138149795 on Build GRCh38
rs915984771 Chr.5: 138149500-138149500 on Build GRCh38
rs532514561 Chr.5: 138149507-138149507 on Build GRCh38
rs780743956 Chr.5: 138149782-138149782 on Build GRCh38
rs774489025 Chr.5: 138149617-138149617 on Build GRCh38
rs765007562 Chr.5: 138149752-138149752 on Build GRCh38
rs530417796 Chr.5: 138149659-138149659 on Build GRCh38
rs770971232 Chr.5: 138149837-138149837 on Build GRCh38
rs376169246 Chr.5: 138149508-138149511 on Build GRCh38
rs913841441 Chr.5: 138149554-138149554 on Build GRCh38
rs749310368 Chr.5: 138149588-138149588 on Build GRCh38
rs747812776 Chr.5: 138149673-138149673 on Build GRCh38
rs374228012 Chr.5: 138149737-138149737 on Build GRCh38
rs375271644 Chr.5: 138149634-138149634 on Build GRCh38
rs750461020 Chr.5: 138149633-138149633 on Build GRCh38
rs749828660 Chr.5: 138149787-138149787 on Build GRCh38
rs764491570 Chr.5: 138149614-138149614 on Build GRCh38
rs529795012 Chr.5: 138149741-138149741 on Build GRCh38
rs761644095 Chr.5: 138149879-138149879 on Build GRCh38
rs768962262 Chr.5: 138149716-138149716 on Build GRCh38
rs928667246 Chr.5: 138149521-138149521 on Build GRCh38
rs1008717641 Chr.5: 138149670-138149670 on Build GRCh38
rs746459201 Chr.5: 138149664-138149665 on Build GRCh38
rs989247378 Chr.5: 138149850-138149850 on Build GRCh38
rs377759893 Chr.5: 138149593-138149593 on Build GRCh38
rs775954506 Chr.5: 138149818-138149818 on Build GRCh38
rs553820952 Chr.5: 138149517-138149517 on Build GRCh38
rs181564357 Chr.5: 138149668-138149668 on Build GRCh38
rs992308427 Chr.5: 138149577-138149577 on Build GRCh38
rs772457392 Chr.5: 138149825-138149825 on Build GRCh38
rs377665713 Chr.5: 138149758-138149758 on Build GRCh38
rs746551438 Chr.5: 138149772-138149772 on Build GRCh38
rs770304569 Chr.5: 138149689-138149689 on Build GRCh38
rs217270 Chr.5: 138149603-138149603 on Build GRCh38
rs777266119 Chr.5: 138149678-138149678 on Build GRCh38
rs754928893 Chr.5: 138149749-138149749 on Build GRCh38
rs370985949 Chr.5: 138149764-138149764 on Build GRCh38
rs777352837 Chr.5: 138149768-138149768 on Build GRCh38
rs773610764 Chr.5: 138149827-138149827 on Build GRCh38
rs563790076 Chr.5: 138149452-138149452 on Build GRCh38
rs775199069 Chr.5: 138149820-138149821 on Build GRCh38
rs775818154 Chr.5: 138149690-138149690 on Build GRCh38
rs774597154 Chr.5: 138149805-138149805 on Build GRCh38
rs974248897 Chr.5: 138149454-138149454 on Build GRCh38
rs767776917 Chr.5: 138149629-138149629 on Build GRCh38
rs577335626 Chr.5: 138149657-138149657 on Build GRCh38
rs946633064 Chr.5: 138149887-138149887 on Build GRCh38
rs202180335 Chr.5: 138149718-138149718 on Build GRCh38
rs763767847 Chr.5: 138149695-138149695 on Build GRCh38
rs769500874 Chr.5: 138149798-138149799 on Build GRCh38
rs760955123 Chr.5: 138149831-138149831 on Build GRCh38
rs535296955 Chr.5: 138149731-138149731 on Build GRCh38
rs774669505 Chr.5: 138149650-138149650 on Build GRCh38
rs758056191 Chr.5: 138149761-138149761 on Build GRCh38
rs112587095 Chr.5: 138149801-138149801 on Build GRCh38
rs749783105 Chr.5: 138149694-138149694 on Build GRCh38
rs554539584 Chr.5: 138149591-138149591 on Build GRCh38
rs767791251 Chr.5: 138149739-138149739 on Build GRCh38
rs1050334314 Chr.5: 138149649-138149649 on Build GRCh38
rs1033872792 Chr.5: 138149800-138149800 on Build GRCh38
rs149886352 Chr.5: 138149684-138149684 on Build GRCh38
rs1004323197 Chr.5: 138149651-138149651 on Build GRCh38
rs770440392 Chr.5: 138149670-138149671 on Build GRCh38
rs536375055 Chr.5: 138149548-138149548 on Build GRCh38
rs981022481 Chr.5: 138149527-138149527 on Build GRCh38
rs761950545 Chr.5: 138149732-138149732 on Build GRCh38
rs762085070 Chr.5: 138149621-138149621 on Build GRCh38
rs763175654 Chr.5: 138149604-138149604 on Build GRCh38
rs565944249 Chr.5: 138149644-138149644 on Build GRCh38
rs762750209 Chr.5: 138149822-138149823 on Build GRCh38
rs756811572 Chr.5: 138149778-138149778 on Build GRCh38
rs770216399 Chr.5: 138149595-138149595 on Build GRCh38
rs758041863 Chr.5: 138149676-138149676 on Build GRCh38
rs760799641 Chr.5: 138149740-138149740 on Build GRCh38
rs745518844 Chr.5: 138149796-138149797 on Build GRCh38
rs548354400 Chr.5: 138149833-138149833 on Build GRCh38
rs188381330 Chr.5: 138149542-138149542 on Build GRCh38

View Full Product Details