Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138873217-138873714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA1
Entrez Gene ID
1495
Gene Name
catenin alpha 1
Gene Aliases
CAP102, MDPT2
UniGene
Hs.445981
Gene Chromosome Location
Chr.5: 138753386-138935034 on Build GRCh38
Transcripts
NM_001290307.2 NM_001290309.2 NM_001290310.2 NM_001323982.1 NM_001323983.1 NM_001323984.1 NM_001323985.1 NM_001323986.1 NM_001903.4 NM_015564.2

Gene Symbol
LRRTM2
Entrez Gene ID
26045
Gene Name
leucine rich repeat transmembrane neuronal 2
Gene Aliases
-
UniGene
Hs.656653
Gene Chromosome Location
Chr.5: 138869390-138875368 on Build GRCh38
Transcripts
NM_001290307.2 NM_001290309.2 NM_001290310.2 NM_001323982.1 NM_001323983.1 NM_001323984.1 NM_001323985.1 NM_001323986.1 NM_001903.4 NM_015564.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061446 c.1326C>A p.F442L Chr.5: 138873235-138873235 on Build GRCh38
5816226 c.1118C>G p.T373S Chr.5: 138873443-138873443 on Build GRCh38
265345 c.989A>T p.Q330L Chr.5: 138873572-138873572 on Build GRCh38
1061450 c.1051C>A p.L351I Chr.5: 138873510-138873510 on Build GRCh38
3850482 c.981C>T p.G327G Chr.5: 138873580-138873580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745925455 Chr.5: 138873287-138873287 on Build GRCh38
rs775908680 Chr.5: 138873296-138873296 on Build GRCh38
rs770344936 Chr.5: 138873428-138873428 on Build GRCh38
rs762694769 Chr.5: 138873675-138873675 on Build GRCh38
rs770258740 Chr.5: 138873295-138873295 on Build GRCh38
rs556094863 Chr.5: 138873547-138873547 on Build GRCh38
rs1038326021 Chr.5: 138873566-138873566 on Build GRCh38
rs140121702 Chr.5: 138873347-138873347 on Build GRCh38
rs747557733 Chr.5: 138873268-138873268 on Build GRCh38
rs370700641 Chr.5: 138873388-138873388 on Build GRCh38
rs35835966 Chr.5: 138873283-138873284 on Build GRCh38
rs763689485 Chr.5: 138873582-138873582 on Build GRCh38
rs1046943475 Chr.5: 138873496-138873496 on Build GRCh38
rs768943323 Chr.5: 138873445-138873445 on Build GRCh38
rs772127640 Chr.5: 138873510-138873510 on Build GRCh38
rs149877566 Chr.5: 138873439-138873439 on Build GRCh38
rs774501018 Chr.5: 138873340-138873340 on Build GRCh38
rs754512980 Chr.5: 138873250-138873250 on Build GRCh38
rs1009693950 Chr.5: 138873625-138873625 on Build GRCh38
rs759214345 Chr.5: 138873548-138873548 on Build GRCh38
rs766251073 Chr.5: 138873541-138873541 on Build GRCh38
rs747045823 Chr.5: 138873622-138873622 on Build GRCh38
rs886918682 Chr.5: 138873538-138873538 on Build GRCh38
rs768694121 Chr.5: 138873339-138873339 on Build GRCh38
rs752266723 Chr.5: 138873569-138873569 on Build GRCh38
rs377375104 Chr.5: 138873364-138873364 on Build GRCh38
rs773201026 Chr.5: 138873531-138873531 on Build GRCh38
rs989713620 Chr.5: 138873253-138873253 on Build GRCh38
rs763781492 Chr.5: 138873682-138873682 on Build GRCh38
rs757948391 Chr.5: 138873418-138873418 on Build GRCh38
rs772215795 Chr.5: 138873613-138873613 on Build GRCh38
rs1000140136 Chr.5: 138873394-138873394 on Build GRCh38
rs753516147 Chr.5: 138873243-138873243 on Build GRCh38
rs200085138 Chr.5: 138873351-138873351 on Build GRCh38
rs774394145 Chr.5: 138873446-138873446 on Build GRCh38
rs1003597731 Chr.5: 138873267-138873267 on Build GRCh38
rs755265206 Chr.5: 138873608-138873608 on Build GRCh38
rs370649836 Chr.5: 138873612-138873612 on Build GRCh38
rs763227727 Chr.5: 138873337-138873337 on Build GRCh38
rs761550008 Chr.5: 138873257-138873259 on Build GRCh38
rs780375690 Chr.5: 138873606-138873606 on Build GRCh38
rs201021876 Chr.5: 138873655-138873655 on Build GRCh38
rs753515291 Chr.5: 138873390-138873390 on Build GRCh38
rs892431036 Chr.5: 138873286-138873286 on Build GRCh38
rs555273836 Chr.5: 138873391-138873391 on Build GRCh38
rs911650562 Chr.5: 138873444-138873444 on Build GRCh38
rs202002034 Chr.5: 138873611-138873611 on Build GRCh38
rs778383547 Chr.5: 138873397-138873397 on Build GRCh38
rs764983388 Chr.5: 138873557-138873557 on Build GRCh38
rs746360538 Chr.5: 138873425-138873425 on Build GRCh38
rs781776155 Chr.5: 138873423-138873423 on Build GRCh38
rs537690931 Chr.5: 138873539-138873539 on Build GRCh38
rs752285650 Chr.5: 138873413-138873413 on Build GRCh38
rs756785995 Chr.5: 138873688-138873688 on Build GRCh38
rs545994986 Chr.5: 138873656-138873656 on Build GRCh38
rs373602374 Chr.5: 138873607-138873607 on Build GRCh38
rs1045330892 Chr.5: 138873586-138873586 on Build GRCh38
rs751135821 Chr.5: 138873684-138873684 on Build GRCh38
rs201004428 Chr.5: 138873543-138873543 on Build GRCh38
rs879168467 Chr.5: 138873661-138873661 on Build GRCh38
rs778157485 Chr.5: 138873432-138873432 on Build GRCh38
rs776404173 Chr.5: 138873646-138873646 on Build GRCh38
rs961805292 Chr.5: 138873449-138873449 on Build GRCh38
rs761931404 Chr.5: 138873452-138873452 on Build GRCh38
rs758036554 Chr.5: 138873579-138873579 on Build GRCh38
rs778494250 Chr.5: 138873251-138873251 on Build GRCh38
rs145134809 Chr.5: 138873592-138873592 on Build GRCh38
rs750339801 Chr.5: 138873357-138873357 on Build GRCh38
rs775275972 Chr.5: 138873674-138873674 on Build GRCh38
rs781751777 Chr.5: 138873277-138873277 on Build GRCh38
rs930880454 Chr.5: 138873583-138873583 on Build GRCh38
rs369875630 Chr.5: 138873270-138873270 on Build GRCh38
rs770935138 Chr.5: 138873632-138873632 on Build GRCh38

View Full Product Details