Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139680659-139681149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC5
Entrez Gene ID
51523
Gene Name
CXXC finger protein 5
Gene Aliases
CF5, HSPC195, RINF, WID
UniGene
Hs.189119
Gene Chromosome Location
Chr.5: 139647299-139683885 on Build GRCh38
Transcripts
NM_001317199.1 NM_001317200.1 NM_001317201.1 NM_001317202.1 NM_001317203.1 NM_001317204.1 NM_001317205.1 NM_001317206.1 NM_001317207.1 NM_001317208.1 NM_001317209.1 NM_001317210.1 NM_001317211.1 NM_016463.8 XM_017009573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674291 c.542C>A p.P181Q Chr.5: 139681065-139681065 on Build GRCh38
5684195 c.465A>T p.A155A Chr.5: 139680988-139680988 on Build GRCh38
5737346 c.424G>A p.A142T Chr.5: 139680947-139680947 on Build GRCh38
3850532 c.586G>A p.V196M Chr.5: 139681109-139681109 on Build GRCh38
1433490 c.163delC p.E57fs*40 Chr.5: 139680686-139680686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763233207 Chr.5: 139680802-139680802 on Build GRCh38
rs759082953 Chr.5: 139681066-139681066 on Build GRCh38
rs143634097 Chr.5: 139680799-139680800 on Build GRCh38
rs751918874 Chr.5: 139680805-139680805 on Build GRCh38
rs759885869 Chr.5: 139680892-139680892 on Build GRCh38
rs777041360 Chr.5: 139681120-139681120 on Build GRCh38
rs775320507 Chr.5: 139680696-139680696 on Build GRCh38
rs746631038 Chr.5: 139680745-139680745 on Build GRCh38
rs751188530 Chr.5: 139680969-139680969 on Build GRCh38
rs753344527 Chr.5: 139680747-139680747 on Build GRCh38
rs769024620 Chr.5: 139681119-139681119 on Build GRCh38
rs761890663 Chr.5: 139680778-139680778 on Build GRCh38
rs775697390 Chr.5: 139681017-139681017 on Build GRCh38
rs356445 Chr.5: 139680901-139680901 on Build GRCh38
rs763354364 Chr.5: 139680711-139680711 on Build GRCh38
rs745505391 Chr.5: 139680757-139680757 on Build GRCh38
rs773400335 Chr.5: 139680886-139680886 on Build GRCh38
rs749977255 Chr.5: 139680824-139680824 on Build GRCh38
rs1051486370 Chr.5: 139681065-139681065 on Build GRCh38
rs867525029 Chr.5: 139681028-139681028 on Build GRCh38
rs376404815 Chr.5: 139680844-139680844 on Build GRCh38
rs768589732 Chr.5: 139680769-139680769 on Build GRCh38
rs746008707 Chr.5: 139681101-139681101 on Build GRCh38
rs752129196 Chr.5: 139680681-139680681 on Build GRCh38
rs764721156 Chr.5: 139680818-139680818 on Build GRCh38
rs762395670 Chr.5: 139681044-139681044 on Build GRCh38
rs776467777 Chr.5: 139680771-139680771 on Build GRCh38
rs369445056 Chr.5: 139680709-139680709 on Build GRCh38
rs370610029 Chr.5: 139681076-139681076 on Build GRCh38
rs199966118 Chr.5: 139680817-139680817 on Build GRCh38
rs760602682 Chr.5: 139680698-139680698 on Build GRCh38
rs748025698 Chr.5: 139680867-139680867 on Build GRCh38
rs771866037 Chr.5: 139680692-139680692 on Build GRCh38
rs199510687 Chr.5: 139680992-139680992 on Build GRCh38
rs914458547 Chr.5: 139680985-139680985 on Build GRCh38
rs772136553 Chr.5: 139681109-139681109 on Build GRCh38
rs758129709 Chr.5: 139680736-139680736 on Build GRCh38
rs545131681 Chr.5: 139681043-139681043 on Build GRCh38
rs779703887 Chr.5: 139680836-139680836 on Build GRCh38
rs375704655 Chr.5: 139680756-139680756 on Build GRCh38
rs780946656 Chr.5: 139680780-139680780 on Build GRCh38
rs572740082 Chr.5: 139680733-139680733 on Build GRCh38
rs113958458 Chr.5: 139680972-139680972 on Build GRCh38
rs768000715 Chr.5: 139680811-139680811 on Build GRCh38
rs777304436 Chr.5: 139680687-139680687 on Build GRCh38
rs753812369 Chr.5: 139681086-139681086 on Build GRCh38
rs999128990 Chr.5: 139681072-139681072 on Build GRCh38
rs746071745 Chr.5: 139681009-139681009 on Build GRCh38
rs955495477 Chr.5: 139680774-139680774 on Build GRCh38
rs376349808 Chr.5: 139680882-139680882 on Build GRCh38
rs373316102 Chr.5: 139680843-139680843 on Build GRCh38
rs767217574 Chr.5: 139681075-139681075 on Build GRCh38
rs528724719 Chr.5: 139681091-139681091 on Build GRCh38
rs778826979 Chr.5: 139681099-139681099 on Build GRCh38
rs768518328 Chr.5: 139680699-139680699 on Build GRCh38
rs555267983 Chr.5: 139680898-139680898 on Build GRCh38
rs761821894 Chr.5: 139680702-139680702 on Build GRCh38
rs771233999 Chr.5: 139680889-139680889 on Build GRCh38
rs769780482 Chr.5: 139680781-139680781 on Build GRCh38
rs202150540 Chr.5: 139681117-139681117 on Build GRCh38
rs773486136 Chr.5: 139680800-139680800 on Build GRCh38
rs754738616 Chr.5: 139680852-139680852 on Build GRCh38
rs903445219 Chr.5: 139680791-139680791 on Build GRCh38
rs771777955 Chr.5: 139680760-139680760 on Build GRCh38
rs772377109 Chr.5: 139681015-139681015 on Build GRCh38
rs755975384 Chr.5: 139681081-139681081 on Build GRCh38
rs748020882 Chr.5: 139680981-139680981 on Build GRCh38
rs766754071 Chr.5: 139680803-139680803 on Build GRCh38
rs201905705 Chr.5: 139680690-139680690 on Build GRCh38
rs544140536 Chr.5: 139680940-139680940 on Build GRCh38
rs780355584 Chr.5: 139681110-139681110 on Build GRCh38
rs999081562 Chr.5: 139680799-139680799 on Build GRCh38
rs780643200 Chr.5: 139680679-139680679 on Build GRCh38
rs764545803 Chr.5: 139681023-139681023 on Build GRCh38
rs746856074 Chr.5: 139680767-139680767 on Build GRCh38
rs750683322 Chr.5: 139680710-139680710 on Build GRCh38
rs756724252 Chr.5: 139680754-139680754 on Build GRCh38
rs755954309 Chr.5: 139680991-139680991 on Build GRCh38
rs751152936 Chr.5: 139681058-139681058 on Build GRCh38
rs766774149 Chr.5: 139680738-139680738 on Build GRCh38
rs773682660 Chr.5: 139681127-139681127 on Build GRCh38
rs867685359 Chr.5: 139680849-139680849 on Build GRCh38
rs749887237 Chr.5: 139680936-139680936 on Build GRCh38
rs868498701 Chr.5: 139680896-139680896 on Build GRCh38
rs981537253 Chr.5: 139680897-139680897 on Build GRCh38
rs777402542 Chr.5: 139680936-139680938 on Build GRCh38
rs749310451 Chr.5: 139680999-139680999 on Build GRCh38
rs200642331 Chr.5: 139680853-139680853 on Build GRCh38
rs769839621 Chr.5: 139680870-139680870 on Build GRCh38
rs755602658 Chr.5: 139680805-139680813 on Build GRCh38
rs774716253 Chr.5: 139680891-139680891 on Build GRCh38
rs748794317 Chr.5: 139680689-139680689 on Build GRCh38
rs754299477 Chr.5: 139680800-139680802 on Build GRCh38
rs369903909 Chr.5: 139680763-139680763 on Build GRCh38
rs779733703 Chr.5: 139680743-139680743 on Build GRCh38
rs771147937 Chr.5: 139681005-139681005 on Build GRCh38
rs778460948 Chr.5: 139680691-139680691 on Build GRCh38
rs777792508 Chr.5: 139681082-139681082 on Build GRCh38
rs754719545 Chr.5: 139680971-139680971 on Build GRCh38
rs183852682 Chr.5: 139681046-139681046 on Build GRCh38
rs371426324 Chr.5: 139680721-139680721 on Build GRCh38
rs749398982 Chr.5: 139680888-139680888 on Build GRCh38
rs756380231 Chr.5: 139680685-139680686 on Build GRCh38
rs761055117 Chr.5: 139680902-139680902 on Build GRCh38
rs780847835 Chr.5: 139680979-139680979 on Build GRCh38
rs145177099 Chr.5: 139680905-139680906 on Build GRCh38
rs373625100 Chr.5: 139681006-139681006 on Build GRCh38
rs759845813 Chr.5: 139680806-139680806 on Build GRCh38
rs758025785 Chr.5: 139680828-139680828 on Build GRCh38
rs1045910763 Chr.5: 139681060-139681060 on Build GRCh38
rs755606525 Chr.5: 139680685-139680685 on Build GRCh38
rs909915231 Chr.5: 139680995-139680995 on Build GRCh38
rs757935649 Chr.5: 139680939-139680939 on Build GRCh38
rs776750218 Chr.5: 139680700-139680700 on Build GRCh38
rs1052970340 Chr.5: 139680938-139680938 on Build GRCh38
rs371784240 Chr.5: 139681021-139681021 on Build GRCh38
rs767759718 Chr.5: 139680900-139680900 on Build GRCh38
rs764635929 Chr.5: 139680905-139680905 on Build GRCh38
rs373740936 Chr.5: 139681045-139681045 on Build GRCh38

View Full Product Details