Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140642234-140642741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO6
Entrez Gene ID
55374
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
PRO1580
UniGene
Hs.534333
Gene Chromosome Location
Chr.5: 140596529-140647411 on Build GRCh38
Transcripts
NM_001300980.1 NM_001300982.1 NM_018502.4 XM_005268477.1 XM_011537663.2 XM_011537665.2 XM_011537668.2 XM_017009617.1 XM_017009618.1 XM_017009619.1 XM_017009620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925230 c.688C>T p.P230S Chr.5: 140642670-140642670 on Build GRCh38
1486341 c.655C>T p.L219L Chr.5: 140642637-140642637 on Build GRCh38
3715096 c.562C>A p.L188M Chr.5: 140642378-140642378 on Build GRCh38
1433645 c.584C>T p.A195V Chr.5: 140642400-140642400 on Build GRCh38
5796259 c.664G>A p.E222K Chr.5: 140642646-140642646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147139789 Chr.5: 140642304-140642304 on Build GRCh38
rs367652614 Chr.5: 140642672-140642672 on Build GRCh38
rs758448668 Chr.5: 140642346-140642346 on Build GRCh38
rs150745857 Chr.5: 140642708-140642708 on Build GRCh38
rs779467512 Chr.5: 140642412-140642412 on Build GRCh38
rs375279046 Chr.5: 140642535-140642535 on Build GRCh38
rs981147017 Chr.5: 140642547-140642547 on Build GRCh38
rs778345722 Chr.5: 140642560-140642560 on Build GRCh38
rs373610346 Chr.5: 140642661-140642661 on Build GRCh38
rs756530494 Chr.5: 140642556-140642556 on Build GRCh38
rs1001231519 Chr.5: 140642716-140642716 on Build GRCh38
rs776294510 Chr.5: 140642429-140642429 on Build GRCh38
rs997560802 Chr.5: 140642293-140642293 on Build GRCh38
rs995145495 Chr.5: 140642274-140642274 on Build GRCh38
rs769591678 Chr.5: 140642457-140642457 on Build GRCh38
rs774101521 Chr.5: 140642711-140642711 on Build GRCh38
rs370959058 Chr.5: 140642458-140642458 on Build GRCh38
rs768249065 Chr.5: 140642426-140642426 on Build GRCh38
rs749204166 Chr.5: 140642322-140642322 on Build GRCh38
rs765117515 Chr.5: 140642328-140642328 on Build GRCh38
rs760307908 Chr.5: 140642297-140642297 on Build GRCh38
rs754101301 Chr.5: 140642657-140642657 on Build GRCh38
rs979327039 Chr.5: 140642420-140642420 on Build GRCh38
rs772921575 Chr.5: 140642616-140642616 on Build GRCh38
rs1033956464 Chr.5: 140642354-140642354 on Build GRCh38
rs890938688 Chr.5: 140642262-140642262 on Build GRCh38
rs761528487 Chr.5: 140642452-140642452 on Build GRCh38
rs561672362 Chr.5: 140642469-140642469 on Build GRCh38
rs568835693 Chr.5: 140642540-140642540 on Build GRCh38
rs750363969 Chr.5: 140642338-140642338 on Build GRCh38
rs770728144 Chr.5: 140642629-140642629 on Build GRCh38
rs377678275 Chr.5: 140642318-140642318 on Build GRCh38
rs778363987 Chr.5: 140642376-140642376 on Build GRCh38
rs759187139 Chr.5: 140642706-140642720 on Build GRCh38
rs750621013 Chr.5: 140642263-140642263 on Build GRCh38
rs969784108 Chr.5: 140642327-140642327 on Build GRCh38
rs764528027 Chr.5: 140642654-140642654 on Build GRCh38
rs772747901 Chr.5: 140642288-140642288 on Build GRCh38
rs201116062 Chr.5: 140642627-140642627 on Build GRCh38
rs762866201 Chr.5: 140642460-140642460 on Build GRCh38
rs12153256 Chr.5: 140642273-140642273 on Build GRCh38
rs777507939 Chr.5: 140642691-140642691 on Build GRCh38
rs780748958 Chr.5: 140642680-140642680 on Build GRCh38
rs761618339 Chr.5: 140642326-140642326 on Build GRCh38
rs1049670160 Chr.5: 140642256-140642256 on Build GRCh38
rs767760700 Chr.5: 140642641-140642641 on Build GRCh38
rs754335549 Chr.5: 140642577-140642577 on Build GRCh38
rs745333896 Chr.5: 140642267-140642267 on Build GRCh38
rs1049617550 Chr.5: 140642558-140642558 on Build GRCh38
rs746462026 Chr.5: 140642588-140642588 on Build GRCh38
rs774380896 Chr.5: 140642634-140642634 on Build GRCh38
rs759624265 Chr.5: 140642637-140642637 on Build GRCh38
rs746551806 Chr.5: 140642353-140642353 on Build GRCh38
rs199968038 Chr.5: 140642592-140642592 on Build GRCh38
rs138690094 Chr.5: 140642486-140642486 on Build GRCh38
rs749000767 Chr.5: 140642697-140642697 on Build GRCh38
rs779495860 Chr.5: 140642587-140642587 on Build GRCh38
rs766429366 Chr.5: 140642347-140642347 on Build GRCh38
rs775191769 Chr.5: 140642286-140642286 on Build GRCh38
rs756581512 Chr.5: 140642367-140642367 on Build GRCh38
rs781676399 Chr.5: 140642360-140642360 on Build GRCh38
rs757671520 Chr.5: 140642659-140642659 on Build GRCh38
rs752961041 Chr.5: 140642644-140642644 on Build GRCh38
rs761014726 Chr.5: 140642652-140642652 on Build GRCh38
rs925270868 Chr.5: 140642530-140642530 on Build GRCh38
rs202097501 Chr.5: 140642615-140642615 on Build GRCh38
rs749815324 Chr.5: 140642390-140642390 on Build GRCh38
rs766452856 Chr.5: 140642465-140642465 on Build GRCh38
rs539451281 Chr.5: 140642269-140642269 on Build GRCh38
rs750952346 Chr.5: 140642671-140642671 on Build GRCh38
rs770443795 Chr.5: 140642448-140642451 on Build GRCh38
rs1036830467 Chr.5: 140642620-140642620 on Build GRCh38
rs534948938 Chr.5: 140642683-140642683 on Build GRCh38
rs757750491 Chr.5: 140642582-140642582 on Build GRCh38
rs747857244 Chr.5: 140642612-140642612 on Build GRCh38
rs752975405 Chr.5: 140642551-140642551 on Build GRCh38
rs979575302 Chr.5: 140642507-140642507 on Build GRCh38
rs960853239 Chr.5: 140642406-140642406 on Build GRCh38
rs755195789 Chr.5: 140642353-140642353 on Build GRCh38
rs142729300 Chr.5: 140642682-140642682 on Build GRCh38
rs528303205 Chr.5: 140642348-140642348 on Build GRCh38
rs545656201 Chr.5: 140642303-140642303 on Build GRCh38
rs188646559 Chr.5: 140642423-140642423 on Build GRCh38
rs771603330 Chr.5: 140642405-140642405 on Build GRCh38
rs776362647 Chr.5: 140642547-140642553 on Build GRCh38

View Full Product Details