Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140697432-140697941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HARS2
Entrez Gene ID
23438
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
Gene Aliases
HARSL, HARSR, HO3, PRLTS2
UniGene
Hs.432560
Gene Chromosome Location
Chr.5: 140691426-140699318 on Build GRCh38
Transcripts
NM_001278731.1 NM_001278732.1 NM_012208.3 XM_011537619.1 XM_011537620.1 XM_017009287.1 XM_017009288.1 XM_017009289.1 XM_017009290.1 XM_017009291.1 XM_017009292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1433660 c.1267C>T p.Q423* Chr.5: 140697638-140697638 on Build GRCh38
4928696 c.1217G>A p.R406Q Chr.5: 140697588-140697588 on Build GRCh38
6169041 c.1300G>T p.D434Y Chr.5: 140697671-140697671 on Build GRCh38
5995891 c.1218G>A p.R406R Chr.5: 140697589-140697589 on Build GRCh38
3850709 c.1258A>G p.N420D Chr.5: 140697629-140697629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748682790 Chr.5: 140697913-140697913 on Build GRCh38
rs765844783 Chr.5: 140697563-140697563 on Build GRCh38
rs893327008 Chr.5: 140697742-140697742 on Build GRCh38
rs754642868 Chr.5: 140697565-140697565 on Build GRCh38
rs753760217 Chr.5: 140697610-140697610 on Build GRCh38
rs752472621 Chr.5: 140697588-140697588 on Build GRCh38
rs755985809 Chr.5: 140697595-140697595 on Build GRCh38
rs753803186 Chr.5: 140697705-140697705 on Build GRCh38
rs182774418 Chr.5: 140697846-140697846 on Build GRCh38
rs202077661 Chr.5: 140697652-140697652 on Build GRCh38
rs767221029 Chr.5: 140697690-140697690 on Build GRCh38
rs777141443 Chr.5: 140697545-140697545 on Build GRCh38
rs374485989 Chr.5: 140697917-140697917 on Build GRCh38
rs747268587 Chr.5: 140697736-140697736 on Build GRCh38
rs746245647 Chr.5: 140697520-140697521 on Build GRCh38
rs528162410 Chr.5: 140697724-140697724 on Build GRCh38
rs546295202 Chr.5: 140697764-140697764 on Build GRCh38
rs575506078 Chr.5: 140697647-140697647 on Build GRCh38
rs972522327 Chr.5: 140697805-140697805 on Build GRCh38
rs1011954903 Chr.5: 140697837-140697837 on Build GRCh38
rs772217829 Chr.5: 140697638-140697638 on Build GRCh38
rs764531628 Chr.5: 140697536-140697536 on Build GRCh38
rs762273590 Chr.5: 140697653-140697653 on Build GRCh38
rs751200040 Chr.5: 140697564-140697564 on Build GRCh38
rs770460592 Chr.5: 140697661-140697661 on Build GRCh38
rs1056435678 Chr.5: 140697880-140697880 on Build GRCh38
rs746101027 Chr.5: 140697635-140697635 on Build GRCh38
rs764031393 Chr.5: 140697698-140697698 on Build GRCh38
rs757211523 Chr.5: 140697712-140697712 on Build GRCh38
rs894783121 Chr.5: 140697832-140697832 on Build GRCh38
rs920700420 Chr.5: 140697825-140697825 on Build GRCh38
rs994002854 Chr.5: 140697870-140697870 on Build GRCh38
rs201542363 Chr.5: 140697721-140697721 on Build GRCh38
rs760515856 Chr.5: 140697696-140697696 on Build GRCh38
rs756560727 Chr.5: 140697593-140697594 on Build GRCh38
rs530332900 Chr.5: 140697831-140697831 on Build GRCh38
rs752429573 Chr.5: 140697691-140697691 on Build GRCh38
rs975119434 Chr.5: 140697674-140697674 on Build GRCh38
rs768983450 Chr.5: 140697895-140697895 on Build GRCh38
rs772702914 Chr.5: 140697506-140697506 on Build GRCh38
rs987716492 Chr.5: 140697770-140697770 on Build GRCh38
rs1034840778 Chr.5: 140697598-140697598 on Build GRCh38
rs73273318 Chr.5: 140697828-140697828 on Build GRCh38
rs767415065 Chr.5: 140697587-140697587 on Build GRCh38
rs199678463 Chr.5: 140697697-140697697 on Build GRCh38
rs962481103 Chr.5: 140697474-140697474 on Build GRCh38
rs962532143 Chr.5: 140697863-140697863 on Build GRCh38
rs968085072 Chr.5: 140697672-140697672 on Build GRCh38
rs138090816 Chr.5: 140697625-140697625 on Build GRCh38
rs759090412 Chr.5: 140697688-140697688 on Build GRCh38
rs773799131 Chr.5: 140697683-140697683 on Build GRCh38
rs777813199 Chr.5: 140697605-140697605 on Build GRCh38
rs747329757 Chr.5: 140697645-140697645 on Build GRCh38
rs35430230 Chr.5: 140697455-140697456 on Build GRCh38
rs918483134 Chr.5: 140697586-140697586 on Build GRCh38
rs563477361 Chr.5: 140697644-140697644 on Build GRCh38
rs934014791 Chr.5: 140697593-140697593 on Build GRCh38
rs757339668 Chr.5: 140697616-140697616 on Build GRCh38
rs907430886 Chr.5: 140697826-140697826 on Build GRCh38
rs569958538 Chr.5: 140697873-140697873 on Build GRCh38
rs770131133 Chr.5: 140697914-140697914 on Build GRCh38
rs879568016 Chr.5: 140697798-140697798 on Build GRCh38
rs781464398 Chr.5: 140697911-140697911 on Build GRCh38
rs912058160 Chr.5: 140697795-140697795 on Build GRCh38
rs750416601 Chr.5: 140697713-140697713 on Build GRCh38
rs775902829 Chr.5: 140697521-140697521 on Build GRCh38
rs772473801 Chr.5: 140697520-140697520 on Build GRCh38
rs574659187 Chr.5: 140697495-140697495 on Build GRCh38
rs761036011 Chr.5: 140697523-140697523 on Build GRCh38
rs953616948 Chr.5: 140697750-140697750 on Build GRCh38
rs1042389330 Chr.5: 140697751-140697751 on Build GRCh38
rs762481131 Chr.5: 140697547-140697547 on Build GRCh38

View Full Product Details