Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140978866-140979377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA10
Entrez Gene ID
56139
Gene Name
protocadherin alpha 10
Gene Aliases
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140855883-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA11
Entrez Gene ID
56138
Gene Name
protocadherin alpha 11
Gene Aliases
CNR7, CNRN7, CNRS7, CRNR7, PCDH-ALPHA11
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140867513-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA12
Entrez Gene ID
56137
Gene Name
protocadherin alpha 12
Gene Aliases
PCDH-ALPHA12
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140875302-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA13
Entrez Gene ID
56136
Gene Name
protocadherin alpha 13
Gene Aliases
CNR5, CNRN5, CNRS5, CRNR5, PCDH-ALPHA13
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140882269-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHAC1
Entrez Gene ID
56135
Gene Name
protocadherin alpha subfamily C, 1
Gene Aliases
PCDH-ALPHA-C1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140926717-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Gene Symbol
PCDHAC2
Entrez Gene ID
56134
Gene Name
protocadherin alpha subfamily C, 2
Gene Aliases
PCDH-ALPHA-C2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140966162-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4762061 c.2389C>T p.R797* Chr.5: 140978952-140978952 on Build GRCh38
6740100 c.2428C>A p.L810M Chr.5: 140978988-140978988 on Build GRCh38
3918943 c.2430G>A p.L810L Chr.5: 140978990-140978990 on Build GRCh38
1062434 c.2437G>T p.A813S Chr.5: 140978994-140978994 on Build GRCh38
6740099 c.2431C>A p.L811M Chr.5: 140978988-140978988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986002369 Chr.5: 140979213-140979213 on Build GRCh38
rs781944777 Chr.5: 140978930-140978930 on Build GRCh38
rs781913955 Chr.5: 140978953-140978953 on Build GRCh38
rs782163702 Chr.5: 140979042-140979042 on Build GRCh38
rs782808442 Chr.5: 140979329-140979329 on Build GRCh38
rs2240296 Chr.5: 140979078-140979078 on Build GRCh38
rs180999093 Chr.5: 140979295-140979295 on Build GRCh38
rs374803810 Chr.5: 140979061-140979061 on Build GRCh38
rs782287157 Chr.5: 140978934-140978934 on Build GRCh38
rs1019875909 Chr.5: 140979079-140979079 on Build GRCh38
rs782577579 Chr.5: 140978981-140978981 on Build GRCh38
rs149397164 Chr.5: 140979006-140979006 on Build GRCh38
rs782325875 Chr.5: 140979026-140979026 on Build GRCh38
rs782153299 Chr.5: 140979316-140979316 on Build GRCh38
rs782073950 Chr.5: 140978950-140978950 on Build GRCh38
rs782420685 Chr.5: 140978941-140978941 on Build GRCh38
rs375756933 Chr.5: 140979047-140979047 on Build GRCh38
rs954222917 Chr.5: 140979197-140979197 on Build GRCh38
rs918582309 Chr.5: 140979261-140979261 on Build GRCh38
rs1035458883 Chr.5: 140979025-140979025 on Build GRCh38
rs782774245 Chr.5: 140978971-140978971 on Build GRCh38
rs1048294748 Chr.5: 140979303-140979303 on Build GRCh38
rs781815387 Chr.5: 140978983-140978983 on Build GRCh38
rs782532288 Chr.5: 140978905-140978905 on Build GRCh38
rs563728648 Chr.5: 140979083-140979083 on Build GRCh38
rs782109224 Chr.5: 140978967-140978967 on Build GRCh38
rs147892853 Chr.5: 140979173-140979173 on Build GRCh38
rs782686641 Chr.5: 140978989-140978989 on Build GRCh38
rs1037042492 Chr.5: 140979248-140979248 on Build GRCh38
rs569091265 Chr.5: 140978922-140978922 on Build GRCh38
rs781894146 Chr.5: 140978925-140978925 on Build GRCh38
rs782082075 Chr.5: 140979054-140979054 on Build GRCh38
rs782380399 Chr.5: 140979125-140979125 on Build GRCh38
rs782016038 Chr.5: 140979027-140979027 on Build GRCh38
rs782321430 Chr.5: 140978909-140978909 on Build GRCh38
rs782371496 Chr.5: 140979043-140979043 on Build GRCh38
rs113796939 Chr.5: 140979350-140979350 on Build GRCh38
rs782169362 Chr.5: 140979015-140979015 on Build GRCh38
rs141879545 Chr.5: 140978973-140978973 on Build GRCh38
rs201501759 Chr.5: 140978957-140978957 on Build GRCh38
rs552719327 Chr.5: 140979220-140979220 on Build GRCh38
rs141662665 Chr.5: 140979275-140979275 on Build GRCh38
rs560855761 Chr.5: 140978960-140978960 on Build GRCh38
rs781997267 Chr.5: 140978942-140978942 on Build GRCh38
rs766062040 Chr.5: 140979241-140979241 on Build GRCh38
rs138901709 Chr.5: 140978986-140978986 on Build GRCh38
rs150254638 Chr.5: 140978974-140978974 on Build GRCh38
rs782602741 Chr.5: 140978907-140978907 on Build GRCh38
rs782253140 Chr.5: 140978997-140978997 on Build GRCh38
rs782336746 Chr.5: 140979000-140979000 on Build GRCh38
rs979649515 Chr.5: 140979109-140979109 on Build GRCh38
rs374951627 Chr.5: 140978952-140978952 on Build GRCh38
rs944063929 Chr.5: 140979246-140979246 on Build GRCh38
rs782006168 Chr.5: 140979178-140979178 on Build GRCh38

View Full Product Details