Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141053722-141054232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101926905
Entrez Gene ID
101926905
Gene Name
uncharacterized LOC101926905
Gene Aliases
-
UniGene
Hs.570900
Gene Chromosome Location
Chr.5: 141046205-141078031 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Gene Symbol
PCDHB1
Entrez Gene ID
29930
Gene Name
protocadherin beta 1
Gene Aliases
PCDH-BETA1
UniGene
Hs.278950
Gene Chromosome Location
Chr.5: 141051376-141053962 on Build GRCh38
Transcripts
NR_105056.2 NM_013340.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6102444 c.2400T>A p.D800E Chr.5: 141053870-141053870 on Build GRCh38
1062526 c.2293A>G p.N765D Chr.5: 141053763-141053763 on Build GRCh38
3918978 c.2367G>A p.M789I Chr.5: 141053837-141053837 on Build GRCh38
7083740 c.2334C>A p.F778L Chr.5: 141053804-141053804 on Build GRCh38
4005899 c.2299G>C p.E767Q Chr.5: 141053769-141053769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73793568 Chr.5: 141054102-141054102 on Build GRCh38
rs782300615 Chr.5: 141053802-141053802 on Build GRCh38
rs141616383 Chr.5: 141053903-141053903 on Build GRCh38
rs782168171 Chr.5: 141053900-141053900 on Build GRCh38
rs375316519 Chr.5: 141053913-141053913 on Build GRCh38
rs781820809 Chr.5: 141053914-141053914 on Build GRCh38
rs782628676 Chr.5: 141053929-141053929 on Build GRCh38
rs782315063 Chr.5: 141053838-141053838 on Build GRCh38
rs146792189 Chr.5: 141054084-141054084 on Build GRCh38
rs781806837 Chr.5: 141053907-141053907 on Build GRCh38
rs969212338 Chr.5: 141054043-141054043 on Build GRCh38
rs782437168 Chr.5: 141053774-141053774 on Build GRCh38
rs1010815135 Chr.5: 141054008-141054008 on Build GRCh38
rs781992243 Chr.5: 141053750-141053750 on Build GRCh38
rs777182019 Chr.5: 141054098-141054098 on Build GRCh38
rs782255869 Chr.5: 141053814-141053814 on Build GRCh38
rs781971257 Chr.5: 141053845-141053845 on Build GRCh38
rs782173555 Chr.5: 141053805-141053805 on Build GRCh38
rs190946862 Chr.5: 141054137-141054137 on Build GRCh38
rs17119401 Chr.5: 141054186-141054186 on Build GRCh38
rs782294522 Chr.5: 141053797-141053797 on Build GRCh38
rs782138101 Chr.5: 141053834-141053834 on Build GRCh38
rs781850814 Chr.5: 141054140-141054140 on Build GRCh38
rs781949354 Chr.5: 141053811-141053811 on Build GRCh38
rs987375614 Chr.5: 141054119-141054119 on Build GRCh38
rs371501773 Chr.5: 141053787-141053787 on Build GRCh38
rs186538514 Chr.5: 141054014-141054014 on Build GRCh38
rs530357860 Chr.5: 141053851-141053851 on Build GRCh38
rs782354216 Chr.5: 141053810-141053810 on Build GRCh38
rs931910219 Chr.5: 141053779-141053779 on Build GRCh38
rs201121650 Chr.5: 141053788-141053788 on Build GRCh38
rs201993759 Chr.5: 141053822-141053822 on Build GRCh38
rs537422212 Chr.5: 141054050-141054050 on Build GRCh38
rs180731232 Chr.5: 141053775-141053775 on Build GRCh38
rs782797033 Chr.5: 141053759-141053759 on Build GRCh38
rs34502840 Chr.5: 141054020-141054021 on Build GRCh38
rs1036146087 Chr.5: 141053817-141053817 on Build GRCh38
rs782518378 Chr.5: 141053923-141053923 on Build GRCh38
rs782785019 Chr.5: 141053856-141053856 on Build GRCh38
rs913295117 Chr.5: 141053766-141053766 on Build GRCh38
rs960265608 Chr.5: 141054005-141054005 on Build GRCh38
rs770655645 Chr.5: 141053763-141053763 on Build GRCh38
rs141650927 Chr.5: 141053772-141053772 on Build GRCh38
rs781904410 Chr.5: 141053870-141053870 on Build GRCh38
rs1001805330 Chr.5: 141053934-141053934 on Build GRCh38
rs544976743 Chr.5: 141053842-141053842 on Build GRCh38
rs781855795 Chr.5: 141053878-141053878 on Build GRCh38
rs148366092 Chr.5: 141053753-141053753 on Build GRCh38
rs782231242 Chr.5: 141053976-141053976 on Build GRCh38
rs2233596 Chr.5: 141053971-141053971 on Build GRCh38
rs138500217 Chr.5: 141053819-141053819 on Build GRCh38
rs782401194 Chr.5: 141053946-141053946 on Build GRCh38
rs953358941 Chr.5: 141054106-141054106 on Build GRCh38
rs968784240 Chr.5: 141053767-141053767 on Build GRCh38
rs372163310 Chr.5: 141053855-141053855 on Build GRCh38
rs782291185 Chr.5: 141053945-141053945 on Build GRCh38
rs374317987 Chr.5: 141053757-141053757 on Build GRCh38
rs376341784 Chr.5: 141053776-141053776 on Build GRCh38

View Full Product Details