Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141320011-141320519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF7
Entrez Gene ID
6879
Gene Name
TATA-box binding protein associated factor 7
Gene Aliases
TAF2F, TAFII55
UniGene
Hs.438838
Gene Chromosome Location
Chr.5: 141318490-141320784 on Build GRCh38
Transcripts
NM_005642.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3851626 c.4A>T p.S2C Chr.5: 141320041-141320041 on Build GRCh38
1267383 c.7A>G p.K3E Chr.5: 141320038-141320038 on Build GRCh38
1062906 c.31G>A p.E11K Chr.5: 141320014-141320014 on Build GRCh38
5002255 c.19G>T p.D7Y Chr.5: 141320026-141320026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963453001 Chr.5: 141320247-141320248 on Build GRCh38
rs748648514 Chr.5: 141320417-141320417 on Build GRCh38
rs879732527 Chr.5: 141320168-141320168 on Build GRCh38
rs149745811 Chr.5: 141320192-141320192 on Build GRCh38
rs776940334 Chr.5: 141320063-141320063 on Build GRCh38
rs533293322 Chr.5: 141320450-141320450 on Build GRCh38
rs759944347 Chr.5: 141320096-141320096 on Build GRCh38
rs11547633 Chr.5: 141320273-141320273 on Build GRCh38
rs746788124 Chr.5: 141320398-141320398 on Build GRCh38
rs751891312 Chr.5: 141320089-141320089 on Build GRCh38
rs901989994 Chr.5: 141320340-141320340 on Build GRCh38
rs143483447 Chr.5: 141320314-141320314 on Build GRCh38
rs768974692 Chr.5: 141320059-141320059 on Build GRCh38
rs1024584658 Chr.5: 141320458-141320458 on Build GRCh38
rs1040972005 Chr.5: 141320334-141320334 on Build GRCh38
rs973890750 Chr.5: 141320257-141320257 on Build GRCh38
rs902940155 Chr.5: 141320147-141320147 on Build GRCh38
rs546888773 Chr.5: 141320459-141320459 on Build GRCh38
rs549796165 Chr.5: 141320331-141320331 on Build GRCh38
rs766978445 Chr.5: 141320081-141320081 on Build GRCh38
rs1016657160 Chr.5: 141320159-141320159 on Build GRCh38
rs762131387 Chr.5: 141320071-141320071 on Build GRCh38
rs950748731 Chr.5: 141320473-141320473 on Build GRCh38
rs146447648 Chr.5: 141320346-141320346 on Build GRCh38
rs745952353 Chr.5: 141320269-141320269 on Build GRCh38
rs1013120993 Chr.5: 141320373-141320373 on Build GRCh38
rs1047979259 Chr.5: 141320361-141320361 on Build GRCh38
rs894248932 Chr.5: 141320435-141320435 on Build GRCh38
rs991731504 Chr.5: 141320206-141320206 on Build GRCh38
rs1035839807 Chr.5: 141320172-141320172 on Build GRCh38
rs189264580 Chr.5: 141320482-141320482 on Build GRCh38
rs1012656834 Chr.5: 141320457-141320457 on Build GRCh38
rs145439224 Chr.5: 141320270-141320270 on Build GRCh38
rs1023371154 Chr.5: 141320223-141320224 on Build GRCh38
rs758671841 Chr.5: 141320412-141320412 on Build GRCh38
rs1011723291 Chr.5: 141320110-141320110 on Build GRCh38
rs779988443 Chr.5: 141320049-141320049 on Build GRCh38
rs998576685 Chr.5: 141320348-141320348 on Build GRCh38
rs912411269 Chr.5: 141320308-141320308 on Build GRCh38
rs763358109 Chr.5: 141320078-141320078 on Build GRCh38
rs773358259 Chr.5: 141320074-141320074 on Build GRCh38
rs896013808 Chr.5: 141320368-141320368 on Build GRCh38
rs899525832 Chr.5: 141320108-141320108 on Build GRCh38
rs758522665 Chr.5: 141320107-141320107 on Build GRCh38
rs746836986 Chr.5: 141320050-141320050 on Build GRCh38
rs148827922 Chr.5: 141320311-141320311 on Build GRCh38
rs924635403 Chr.5: 141320302-141320302 on Build GRCh38
rs17602721 Chr.5: 141320112-141320112 on Build GRCh38
rs910588397 Chr.5: 141320321-141320321 on Build GRCh38
rs765135260 Chr.5: 141320218-141320219 on Build GRCh38
rs530628337 Chr.5: 141320378-141320378 on Build GRCh38
rs11547632 Chr.5: 141320401-141320401 on Build GRCh38
rs920925821 Chr.5: 141320318-141320318 on Build GRCh38
rs540505752 Chr.5: 141320281-141320281 on Build GRCh38
rs958565993 Chr.5: 141320171-141320171 on Build GRCh38
rs550543212 Chr.5: 141320399-141320399 on Build GRCh38
rs769700302 Chr.5: 141320073-141320073 on Build GRCh38
rs561582522 Chr.5: 141320353-141320353 on Build GRCh38
rs772103968 Chr.5: 141320048-141320048 on Build GRCh38
rs994807376 Chr.5: 141320430-141320431 on Build GRCh38
rs770801383 Chr.5: 141320079-141320079 on Build GRCh38
rs745348532 Chr.5: 141320195-141320195 on Build GRCh38
rs1016606224 Chr.5: 141320429-141320429 on Build GRCh38
rs991769774 Chr.5: 141320175-141320175 on Build GRCh38

View Full Product Details