Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141392793-141393326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_014004.2 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619740 c.464C>A p.P155Q Chr.5: 141393277-141393277 on Build GRCh38
1063200 c.243C>T p.S81S Chr.5: 141393056-141393056 on Build GRCh38
1063198 c.94C>T p.R32C Chr.5: 141392907-141392907 on Build GRCh38
1567396 c.430G>A p.E144K Chr.5: 141393243-141393243 on Build GRCh38
5447627 c.117G>A p.T39T Chr.5: 141392930-141392930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000509941 Chr.5: 141392866-141392866 on Build GRCh38
rs888115124 Chr.5: 141393280-141393280 on Build GRCh38
rs568235859 Chr.5: 141392930-141392930 on Build GRCh38
rs756948310 Chr.5: 141393140-141393140 on Build GRCh38
rs747012880 Chr.5: 141393080-141393080 on Build GRCh38
rs759327251 Chr.5: 141392847-141392847 on Build GRCh38
rs768666493 Chr.5: 141393159-141393160 on Build GRCh38
rs754075095 Chr.5: 141393013-141393013 on Build GRCh38
rs758868753 Chr.5: 141393021-141393021 on Build GRCh38
rs780347240 Chr.5: 141392941-141392941 on Build GRCh38
rs980151183 Chr.5: 141393065-141393065 on Build GRCh38
rs573495684 Chr.5: 141392986-141392986 on Build GRCh38
rs755325492 Chr.5: 141393024-141393024 on Build GRCh38
rs376887343 Chr.5: 141393297-141393297 on Build GRCh38
rs368785983 Chr.5: 141393012-141393012 on Build GRCh38
rs148589854 Chr.5: 141393112-141393112 on Build GRCh38
rs746908655 Chr.5: 141393170-141393170 on Build GRCh38
rs372159245 Chr.5: 141393023-141393023 on Build GRCh38
rs950133126 Chr.5: 141393222-141393222 on Build GRCh38
rs759053140 Chr.5: 141393293-141393293 on Build GRCh38
rs113280752 Chr.5: 141393169-141393169 on Build GRCh38
rs776358777 Chr.5: 141392825-141392825 on Build GRCh38
rs868285753 Chr.5: 141393027-141393027 on Build GRCh38
rs771952632 Chr.5: 141393052-141393052 on Build GRCh38
rs373432896 Chr.5: 141393204-141393204 on Build GRCh38
rs953803165 Chr.5: 141392978-141392978 on Build GRCh38
rs774531655 Chr.5: 141393293-141393293 on Build GRCh38
rs770759647 Chr.5: 141393173-141393173 on Build GRCh38
rs891971324 Chr.5: 141393101-141393101 on Build GRCh38
rs151037104 Chr.5: 141393242-141393242 on Build GRCh38
rs537186931 Chr.5: 141393264-141393264 on Build GRCh38
rs777446564 Chr.5: 141393137-141393137 on Build GRCh38
rs753113857 Chr.5: 141393228-141393228 on Build GRCh38
rs1005323306 Chr.5: 141393081-141393081 on Build GRCh38
rs781611512 Chr.5: 141393093-141393093 on Build GRCh38
rs755308269 Chr.5: 141393134-141393134 on Build GRCh38
rs201895231 Chr.5: 141392844-141392844 on Build GRCh38
rs770357830 Chr.5: 141392977-141392977 on Build GRCh38
rs772870041 Chr.5: 141392964-141392964 on Build GRCh38
rs774279389 Chr.5: 141393206-141393206 on Build GRCh38
rs769377200 Chr.5: 141392956-141392956 on Build GRCh38
rs781226042 Chr.5: 141393272-141393272 on Build GRCh38
rs773579763 Chr.5: 141393276-141393276 on Build GRCh38
rs774288267 Chr.5: 141392993-141392993 on Build GRCh38
rs922844407 Chr.5: 141392833-141392834 on Build GRCh38
rs757363357 Chr.5: 141392870-141392870 on Build GRCh38
rs778464494 Chr.5: 141393047-141393047 on Build GRCh38
rs752035971 Chr.5: 141393130-141393130 on Build GRCh38
rs771057124 Chr.5: 141393285-141393285 on Build GRCh38
rs768038476 Chr.5: 141393299-141393299 on Build GRCh38
rs563145930 Chr.5: 141392985-141392985 on Build GRCh38
rs766823317 Chr.5: 141393127-141393127 on Build GRCh38
rs892304367 Chr.5: 141392976-141392976 on Build GRCh38
rs756361613 Chr.5: 141392855-141392855 on Build GRCh38
rs753058672 Chr.5: 141393139-141393139 on Build GRCh38
rs775192271 Chr.5: 141393001-141393001 on Build GRCh38
rs768471669 Chr.5: 141393171-141393171 on Build GRCh38
rs142751758 Chr.5: 141393120-141393120 on Build GRCh38
rs889451396 Chr.5: 141393167-141393167 on Build GRCh38
rs369051018 Chr.5: 141393196-141393196 on Build GRCh38
rs747812941 Chr.5: 141392830-141392830 on Build GRCh38
rs375085176 Chr.5: 141393179-141393179 on Build GRCh38
rs746317747 Chr.5: 141392907-141392907 on Build GRCh38
rs760159490 Chr.5: 141393217-141393217 on Build GRCh38
rs759529663 Chr.5: 141392995-141392995 on Build GRCh38
rs750632906 Chr.5: 141392833-141392833 on Build GRCh38
rs747367099 Chr.5: 141392949-141392949 on Build GRCh38
rs772046833 Chr.5: 141393000-141393000 on Build GRCh38
rs371909748 Chr.5: 141393141-141393141 on Build GRCh38
rs201229959 Chr.5: 141393255-141393255 on Build GRCh38
rs748877946 Chr.5: 141393038-141393038 on Build GRCh38
rs988866288 Chr.5: 141393203-141393203 on Build GRCh38
rs182052960 Chr.5: 141393006-141393006 on Build GRCh38
rs774547937 Chr.5: 141393232-141393233 on Build GRCh38
rs752526573 Chr.5: 141392850-141392850 on Build GRCh38
rs745941759 Chr.5: 141393200-141393200 on Build GRCh38
rs762881601 Chr.5: 141392998-141392999 on Build GRCh38
rs781030004 Chr.5: 141393004-141393004 on Build GRCh38
rs77141792 Chr.5: 141392843-141392843 on Build GRCh38
rs775449711 Chr.5: 141393074-141393074 on Build GRCh38
rs755014362 Chr.5: 141393265-141393265 on Build GRCh38
rs567943150 Chr.5: 141393261-141393261 on Build GRCh38
rs776552182 Chr.5: 141393103-141393103 on Build GRCh38
rs750579370 Chr.5: 141393114-141393114 on Build GRCh38
rs748824740 Chr.5: 141392968-141392968 on Build GRCh38
rs749579658 Chr.5: 141393277-141393277 on Build GRCh38
rs1057103671 Chr.5: 141393095-141393095 on Build GRCh38
rs777293240 Chr.5: 141393034-141393034 on Build GRCh38
rs762190466 Chr.5: 141393111-141393111 on Build GRCh38
rs758960557 Chr.5: 141392927-141392927 on Build GRCh38
rs726684 Chr.5: 141392860-141392860 on Build GRCh38
rs377394191 Chr.5: 141393241-141393241 on Build GRCh38
rs770162660 Chr.5: 141393194-141393194 on Build GRCh38
rs769641711 Chr.5: 141393275-141393275 on Build GRCh38
rs755462760 Chr.5: 141393015-141393015 on Build GRCh38
rs756421828 Chr.5: 141393233-141393233 on Build GRCh38
rs760420305 Chr.5: 141393088-141393088 on Build GRCh38
rs779595460 Chr.5: 141392880-141392880 on Build GRCh38
rs374832992 Chr.5: 141392848-141392848 on Build GRCh38
rs745405842 Chr.5: 141393144-141393144 on Build GRCh38
rs772636303 Chr.5: 141392837-141392837 on Build GRCh38
rs745354934 Chr.5: 141393048-141393048 on Build GRCh38
rs927273885 Chr.5: 141393071-141393071 on Build GRCh38
rs761990025 Chr.5: 141393014-141393014 on Build GRCh38
rs767983504 Chr.5: 141393225-141393225 on Build GRCh38
rs1028021318 Chr.5: 141393156-141393156 on Build GRCh38
rs770560068 Chr.5: 141393043-141393043 on Build GRCh38
rs771570808 Chr.5: 141393162-141393162 on Build GRCh38
rs533530561 Chr.5: 141393086-141393086 on Build GRCh38
rs747983531 Chr.5: 141393274-141393274 on Build GRCh38
rs764346882 Chr.5: 141392858-141392858 on Build GRCh38
rs768432261 Chr.5: 141393102-141393102 on Build GRCh38

View Full Product Details