Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 143282171-143282714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR3C1
Entrez Gene ID
2908
Gene Name
nuclear receptor subfamily 3 group C member 1
Gene Aliases
GCCR, GCR, GCRST, GR, GRL
UniGene
Hs.122926
Gene Chromosome Location
Chr.5: 143277931-143435512 on Build GRCh38
Transcripts
NM_000176.2 NM_001018074.1 NM_001018075.1 NM_001018076.1 NM_001018077.1 NM_001020825.1 NM_001024094.1 NM_001204258.1 NM_001204259.1 NM_001204260.1 NM_001204261.1 NM_001204262.1 NM_001204263.1 NM_001204264.1 XM_005268419.3 XM_005268420.4 XM_005268422.3 XM_005268423.3 XM_011537637.2 XM_017009397.1 XM_017009398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1063645 c.2174C>A p.S725Y Chr.5: 143282578-143282578 on Build GRCh38
3720203 c.2174T>C p.M725T Chr.5: 143282575-143282575 on Build GRCh38
5958482 c.2059G>T p.D687Y Chr.5: 143282690-143282690 on Build GRCh38
6529649 c.2083A>G p.K695E Chr.5: 143282666-143282666 on Build GRCh38
1063646 c.2053G>T p.E685* Chr.5: 143282699-143282699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72542755 Chr.5: 143282222-143282222 on Build GRCh38
rs758528337 Chr.5: 143282538-143282538 on Build GRCh38
rs948422515 Chr.5: 143282480-143282480 on Build GRCh38
rs1013575599 Chr.5: 143282525-143282525 on Build GRCh38
rs897055628 Chr.5: 143282439-143282439 on Build GRCh38
rs10482704 Chr.5: 143282198-143282198 on Build GRCh38
rs951475765 Chr.5: 143282217-143282217 on Build GRCh38
rs759445158 Chr.5: 143282608-143282608 on Build GRCh38
rs258750 Chr.5: 143282324-143282324 on Build GRCh38
rs72481853 Chr.5: 143282531-143282531 on Build GRCh38
rs769056375 Chr.5: 143282548-143282548 on Build GRCh38
rs72542756 Chr.5: 143282199-143282199 on Build GRCh38
rs1048220786 Chr.5: 143282471-143282471 on Build GRCh38
rs777651449 Chr.5: 143282564-143282564 on Build GRCh38
rs775051931 Chr.5: 143282616-143282616 on Build GRCh38
rs113037974 Chr.5: 143282346-143282346 on Build GRCh38
rs541779640 Chr.5: 143282442-143282442 on Build GRCh38
rs774364229 Chr.5: 143282607-143282607 on Build GRCh38
rs929915599 Chr.5: 143282468-143282468 on Build GRCh38
rs763752146 Chr.5: 143282673-143282673 on Build GRCh38
rs764474909 Chr.5: 143282684-143282684 on Build GRCh38
rs564350182 Chr.5: 143282477-143282477 on Build GRCh38
rs72542754 Chr.5: 143282225-143282225 on Build GRCh38
rs545931159 Chr.5: 143282583-143282583 on Build GRCh38
rs577805753 Chr.5: 143282482-143282482 on Build GRCh38
rs560600948 Chr.5: 143282500-143282500 on Build GRCh38
rs762197361 Chr.5: 143282690-143282690 on Build GRCh38
rs980483872 Chr.5: 143282464-143282464 on Build GRCh38
rs868455100 Chr.5: 143282470-143282470 on Build GRCh38
rs780036686 Chr.5: 143282542-143282542 on Build GRCh38
rs966985372 Chr.5: 143282558-143282558 on Build GRCh38
rs757704932 Chr.5: 143282529-143282529 on Build GRCh38
rs943659861 Chr.5: 143282432-143282432 on Build GRCh38
rs910990929 Chr.5: 143282391-143282391 on Build GRCh38
rs72481859 Chr.5: 143282221-143282221 on Build GRCh38
rs760310145 Chr.5: 143282642-143282642 on Build GRCh38
rs771583164 Chr.5: 143282609-143282609 on Build GRCh38
rs184014459 Chr.5: 143282326-143282326 on Build GRCh38
rs755709126 Chr.5: 143282550-143282551 on Build GRCh38
rs370379496 Chr.5: 143282204-143282204 on Build GRCh38
rs774334389 Chr.5: 143282395-143282395 on Build GRCh38
rs887171126 Chr.5: 143282560-143282560 on Build GRCh38
rs770730144 Chr.5: 143282598-143282598 on Build GRCh38
rs575017724 Chr.5: 143282420-143282420 on Build GRCh38
rs9324911 Chr.5: 143282277-143282277 on Build GRCh38
rs563383688 Chr.5: 143282289-143282289 on Build GRCh38
rs965782947 Chr.5: 143282287-143282287 on Build GRCh38
rs376438855 Chr.5: 143282543-143282543 on Build GRCh38
rs68012717 Chr.5: 143282688-143282688 on Build GRCh38
rs376392309 Chr.5: 143282249-143282249 on Build GRCh38
rs985176330 Chr.5: 143282380-143282380 on Build GRCh38
rs72481855 Chr.5: 143282485-143282485 on Build GRCh38
rs529320390 Chr.5: 143282640-143282640 on Build GRCh38
rs749929503 Chr.5: 143282408-143282408 on Build GRCh38
rs72481857 Chr.5: 143282323-143282323 on Build GRCh38
rs760965236 Chr.5: 143282483-143282483 on Build GRCh38
rs936827397 Chr.5: 143282367-143282367 on Build GRCh38
rs750431120 Chr.5: 143282534-143282534 on Build GRCh38
rs992587232 Chr.5: 143282268-143282268 on Build GRCh38
rs72481854 Chr.5: 143282487-143282487 on Build GRCh38
rs187197352 Chr.5: 143282365-143282365 on Build GRCh38
rs777408738 Chr.5: 143282257-143282258 on Build GRCh38
rs1033696086 Chr.5: 143282494-143282494 on Build GRCh38
rs763362243 Chr.5: 143282301-143282301 on Build GRCh38
rs375800105 Chr.5: 143282589-143282589 on Build GRCh38
rs975344259 Chr.5: 143282291-143282291 on Build GRCh38
rs1041131072 Chr.5: 143282436-143282436 on Build GRCh38
rs779481252 Chr.5: 143282530-143282530 on Build GRCh38

View Full Product Details