Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147370523-147370940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK32A
Entrez Gene ID
202374
Gene Name
serine/threonine kinase 32A
Gene Aliases
YANK1
UniGene
Hs.585069
Gene Chromosome Location
Chr.5: 147234963-147401996 on Build GRCh38
Transcripts
NM_001112724.1 NM_001287740.1 XM_011537576.1 XM_011537577.2 XM_011537579.2 XM_017009213.1 XM_017009214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1486472 c.720G>C p.T240T Chr.5: 147370713-147370713 on Build GRCh38
6577779 c.686C>T p.T229I Chr.5: 147370679-147370679 on Build GRCh38
3852475 c.777+2T>C p.? Chr.5: 147370772-147370772 on Build GRCh38
1288246 c.666G>A p.P222P Chr.5: 147370659-147370659 on Build GRCh38
5774763 c.661-1G>T p.? Chr.5: 147370653-147370653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006145974 Chr.5: 147370868-147370868 on Build GRCh38
rs202216975 Chr.5: 147370617-147370617 on Build GRCh38
rs958871190 Chr.5: 147370557-147370557 on Build GRCh38
rs766888423 Chr.5: 147370615-147370615 on Build GRCh38
rs768543830 Chr.5: 147370788-147370788 on Build GRCh38
rs377192467 Chr.5: 147370694-147370694 on Build GRCh38
rs532235744 Chr.5: 147370547-147370547 on Build GRCh38
rs961835846 Chr.5: 147370575-147370575 on Build GRCh38
rs778106501 Chr.5: 147370755-147370755 on Build GRCh38
rs776249458 Chr.5: 147370605-147370605 on Build GRCh38
rs570106666 Chr.5: 147370835-147370835 on Build GRCh38
rs763346990 Chr.5: 147370712-147370712 on Build GRCh38
rs368677802 Chr.5: 147370659-147370659 on Build GRCh38
rs750751349 Chr.5: 147370611-147370611 on Build GRCh38
rs746827293 Chr.5: 147370787-147370787 on Build GRCh38
rs79182349 Chr.5: 147370594-147370594 on Build GRCh38
rs1006093286 Chr.5: 147370564-147370564 on Build GRCh38
rs762571701 Chr.5: 147370801-147370801 on Build GRCh38
rs772913984 Chr.5: 147370797-147370797 on Build GRCh38
rs764877039 Chr.5: 147370608-147370608 on Build GRCh38
rs769657142 Chr.5: 147370697-147370698 on Build GRCh38
rs775488705 Chr.5: 147370669-147370669 on Build GRCh38
rs752775183 Chr.5: 147370729-147370729 on Build GRCh38
rs550361613 Chr.5: 147370829-147370829 on Build GRCh38
rs79366764 Chr.5: 147370597-147370597 on Build GRCh38
rs781481072 Chr.5: 147370618-147370618 on Build GRCh38
rs371450812 Chr.5: 147370565-147370565 on Build GRCh38
rs971826528 Chr.5: 147370577-147370577 on Build GRCh38
rs756362796 Chr.5: 147370629-147370629 on Build GRCh38
rs776056104 Chr.5: 147370685-147370685 on Build GRCh38
rs752095296 Chr.5: 147370714-147370714 on Build GRCh38
rs1056191877 Chr.5: 147370843-147370843 on Build GRCh38
rs1034296768 Chr.5: 147370553-147370553 on Build GRCh38
rs763447459 Chr.5: 147370785-147370785 on Build GRCh38
rs898536328 Chr.5: 147370883-147370883 on Build GRCh38
rs530297464 Chr.5: 147370766-147370766 on Build GRCh38
rs149829069 Chr.5: 147370567-147370567 on Build GRCh38
rs908363133 Chr.5: 147370836-147370836 on Build GRCh38
rs1028578940 Chr.5: 147370634-147370634 on Build GRCh38
rs758715555 Chr.5: 147370612-147370612 on Build GRCh38
rs987836251 Chr.5: 147370676-147370676 on Build GRCh38
rs776576808 Chr.5: 147370789-147370789 on Build GRCh38
rs187250629 Chr.5: 147370619-147370619 on Build GRCh38
rs778163507 Chr.5: 147370633-147370633 on Build GRCh38
rs761399471 Chr.5: 147370607-147370607 on Build GRCh38
rs375706377 Chr.5: 147370597-147370597 on Build GRCh38
rs201186688 Chr.5: 147370670-147370670 on Build GRCh38
rs752047732 Chr.5: 147370616-147370616 on Build GRCh38
rs755371722 Chr.5: 147370721-147370721 on Build GRCh38
rs745652013 Chr.5: 147370644-147370644 on Build GRCh38
rs762587520 Chr.5: 147370780-147370780 on Build GRCh38
rs939263634 Chr.5: 147370798-147370798 on Build GRCh38
rs1003273911 Chr.5: 147370546-147370546 on Build GRCh38
rs1056304317 Chr.5: 147370596-147370597 on Build GRCh38
rs1008428514 Chr.5: 147370658-147370658 on Build GRCh38
rs749535396 Chr.5: 147370757-147370757 on Build GRCh38
rs756309936 Chr.5: 147370745-147370745 on Build GRCh38
rs759702466 Chr.5: 147370854-147370854 on Build GRCh38
rs766720419 Chr.5: 147370713-147370713 on Build GRCh38
rs746204111 Chr.5: 147370607-147370607 on Build GRCh38
rs776831665 Chr.5: 147370810-147370810 on Build GRCh38
rs926595749 Chr.5: 147370763-147370763 on Build GRCh38
rs1041836239 Chr.5: 147370747-147370747 on Build GRCh38
rs769072429 Chr.5: 147370795-147370802 on Build GRCh38
rs939200258 Chr.5: 147370580-147370580 on Build GRCh38
rs199544939 Chr.5: 147370696-147370696 on Build GRCh38
rs779652326 Chr.5: 147370775-147370775 on Build GRCh38
rs769334942 Chr.5: 147370791-147370791 on Build GRCh38
rs768167953 Chr.5: 147370728-147370728 on Build GRCh38
rs996866583 Chr.5: 147370906-147370906 on Build GRCh38
rs559431844 Chr.5: 147370596-147370596 on Build GRCh38

View Full Product Details