Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147586615-147587144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP2
Entrez Gene ID
9832
Gene Name
janus kinase and microtubule interacting protein 2
Gene Aliases
JAMIP2, NECC1
UniGene
Hs.740461
Gene Chromosome Location
Chr.5: 147585439-147782848 on Build GRCh38
Transcripts
NM_001270934.1 NM_001270941.1 NM_001282282.1 NM_014790.4 XM_017010097.1 XM_017010098.1 NR_038902.1

Gene Symbol
JAKMIP2-AS1
Entrez Gene ID
153469
Gene Name
JAKMIP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.184323
Gene Chromosome Location
Chr.5: 147559994-147662009 on Build GRCh38
Transcripts
NM_001270934.1 NM_001270941.1 NM_001282282.1 NM_014790.4 XM_017010097.1 XM_017010098.1 NR_038902.1

SNP Detail

SNP ID Chromosome Location
rs185251051 Chr.5: 147586856-147586856 on Build GRCh38
rs555434884 Chr.5: 147587093-147587093 on Build GRCh38
rs4705178 Chr.5: 147586648-147586648 on Build GRCh38
rs1009127791 Chr.5: 147587114-147587114 on Build GRCh38
rs5872028 Chr.5: 147586759-147586759 on Build GRCh38
rs1030260286 Chr.5: 147586702-147586702 on Build GRCh38
rs1046701555 Chr.5: 147586897-147586897 on Build GRCh38
rs958229746 Chr.5: 147586841-147586841 on Build GRCh38
rs761389404 Chr.5: 147586717-147586717 on Build GRCh38
rs760790046 Chr.5: 147586674-147586674 on Build GRCh38
rs74947166 Chr.5: 147586931-147586931 on Build GRCh38
rs559309015 Chr.5: 147586806-147586806 on Build GRCh38
rs190705387 Chr.5: 147587084-147587084 on Build GRCh38
rs561201092 Chr.5: 147587001-147587001 on Build GRCh38
rs564433055 Chr.5: 147586842-147586842 on Build GRCh38
rs544916764 Chr.5: 147586826-147586826 on Build GRCh38
rs527611442 Chr.5: 147586793-147586793 on Build GRCh38
rs997364009 Chr.5: 147587014-147587014 on Build GRCh38
rs577840737 Chr.5: 147586807-147586807 on Build GRCh38
rs75544567 Chr.5: 147586775-147586775 on Build GRCh38
rs1022003058 Chr.5: 147586939-147586939 on Build GRCh38
rs1008012222 Chr.5: 147587099-147587099 on Build GRCh38
rs908824592 Chr.5: 147586743-147586743 on Build GRCh38
rs1038374242 Chr.5: 147586794-147586794 on Build GRCh38
rs997395128 Chr.5: 147586695-147586695 on Build GRCh38
rs939998610 Chr.5: 147586757-147586757 on Build GRCh38
rs949588081 Chr.5: 147586885-147586885 on Build GRCh38
rs887560431 Chr.5: 147586949-147586949 on Build GRCh38
rs1008744048 Chr.5: 147586900-147586900 on Build GRCh38
rs754300763 Chr.5: 147586760-147586760 on Build GRCh38
rs113483761 Chr.5: 147586952-147586952 on Build GRCh38
rs148895689 Chr.5: 147586983-147586983 on Build GRCh38
rs940318982 Chr.5: 147587018-147587018 on Build GRCh38
rs760071207 Chr.5: 147586642-147586642 on Build GRCh38
rs919888202 Chr.5: 147587058-147587058 on Build GRCh38
rs547396774 Chr.5: 147586798-147586798 on Build GRCh38
rs537032924 Chr.5: 147587089-147587089 on Build GRCh38
rs183335364 Chr.5: 147587102-147587102 on Build GRCh38
rs542660124 Chr.5: 147586860-147586860 on Build GRCh38
rs573693437 Chr.5: 147586721-147586721 on Build GRCh38
rs796909621 Chr.5: 147586776-147586776 on Build GRCh38
rs112915199 Chr.5: 147586958-147586958 on Build GRCh38
rs180856112 Chr.5: 147586737-147586737 on Build GRCh38
rs1019265273 Chr.5: 147586901-147586901 on Build GRCh38
rs763550372 Chr.5: 147586644-147586644 on Build GRCh38
rs77389992 Chr.5: 147587069-147587069 on Build GRCh38
rs1055544254 Chr.5: 147586942-147586942 on Build GRCh38
rs926740991 Chr.5: 147586719-147586719 on Build GRCh38
rs1030229207 Chr.5: 147587061-147587061 on Build GRCh38
rs754359190 Chr.5: 147586744-147586744 on Build GRCh38
rs747460787 Chr.5: 147586759-147586760 on Build GRCh38
rs1049802840 Chr.5: 147586865-147586865 on Build GRCh38
rs961895362 Chr.5: 147586694-147586694 on Build GRCh38
rs914974276 Chr.5: 147586643-147586643 on Build GRCh38
rs939556691 Chr.5: 147586728-147586728 on Build GRCh38
rs755417137 Chr.5: 147586973-147586973 on Build GRCh38
rs925387229 Chr.5: 147586828-147586828 on Build GRCh38
rs537104125 Chr.5: 147586638-147586638 on Build GRCh38
rs551307978 Chr.5: 147587070-147587070 on Build GRCh38
rs376621075 Chr.5: 147586925-147586926 on Build GRCh38
rs561424656 Chr.5: 147586866-147586866 on Build GRCh38
rs928268830 Chr.5: 147586914-147586914 on Build GRCh38
rs376724217 Chr.5: 147586915-147586915 on Build GRCh38
rs77307865 Chr.5: 147587115-147587115 on Build GRCh38
rs78029591 Chr.5: 147586774-147586776 on Build GRCh38
rs1038598074 Chr.5: 147587028-147587028 on Build GRCh38

View Full Product Details