Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149867817-149868312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6A
Entrez Gene ID
5145
Gene Name
phosphodiesterase 6A
Gene Aliases
CGPR-A, PDEA, RP43
UniGene
Hs.567314
Gene Chromosome Location
Chr.5: 149857956-149944793 on Build GRCh38
Transcripts
NM_000440.2 XM_011537650.2 XM_011537651.2 XM_011537652.1 XM_011537653.2 XM_011537654.2 XM_017009572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1261780 c.2180C>T p.P727L Chr.5: 149868114-149868114 on Build GRCh38
1130835 c.2151C>T p.T717T Chr.5: 149868143-149868143 on Build GRCh38
1064208 c.2177A>C p.K726T Chr.5: 149868117-149868117 on Build GRCh38
6170024 c.2181C>A p.P727P Chr.5: 149868113-149868113 on Build GRCh38
3827383 c.2145G>A p.M715I Chr.5: 149868149-149868149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111830959 Chr.5: 149868074-149868074 on Build GRCh38
rs542108387 Chr.5: 149867994-149867994 on Build GRCh38
rs568698935 Chr.5: 149868098-149868098 on Build GRCh38
rs138312324 Chr.5: 149868282-149868282 on Build GRCh38
rs548484716 Chr.5: 149868244-149868244 on Build GRCh38
rs562699961 Chr.5: 149867892-149867892 on Build GRCh38
rs371263362 Chr.5: 149867840-149867840 on Build GRCh38
rs964760051 Chr.5: 149867862-149867862 on Build GRCh38
rs751365781 Chr.5: 149868082-149868082 on Build GRCh38
rs368489306 Chr.5: 149868206-149868206 on Build GRCh38
rs899254396 Chr.5: 149867999-149867999 on Build GRCh38
rs556435007 Chr.5: 149867912-149867912 on Build GRCh38
rs80172488 Chr.5: 149868145-149868145 on Build GRCh38
rs747525716 Chr.5: 149868058-149868083 on Build GRCh38
rs768685072 Chr.5: 149868056-149868056 on Build GRCh38
rs200483225 Chr.5: 149868171-149868171 on Build GRCh38
rs73796302 Chr.5: 149868247-149868247 on Build GRCh38
rs191915544 Chr.5: 149868270-149868270 on Build GRCh38
rs938936209 Chr.5: 149867922-149867922 on Build GRCh38
rs375189945 Chr.5: 149868195-149868195 on Build GRCh38
rs148193179 Chr.5: 149868024-149868024 on Build GRCh38
rs570531243 Chr.5: 149868290-149868290 on Build GRCh38
rs762041831 Chr.5: 149868071-149868071 on Build GRCh38
rs780572921 Chr.5: 149868160-149868160 on Build GRCh38
rs72830272 Chr.5: 149867872-149867872 on Build GRCh38
rs1056801070 Chr.5: 149868274-149868274 on Build GRCh38
rs1035820136 Chr.5: 149868011-149868011 on Build GRCh38
rs777359709 Chr.5: 149868128-149868128 on Build GRCh38
rs6864267 Chr.5: 149868175-149868175 on Build GRCh38
rs531812937 Chr.5: 149867995-149867995 on Build GRCh38
rs915410340 Chr.5: 149868191-149868191 on Build GRCh38
rs760764085 Chr.5: 149868048-149868048 on Build GRCh38
rs775592389 Chr.5: 149867850-149867850 on Build GRCh38
rs574839790 Chr.5: 149867979-149867979 on Build GRCh38
rs977567092 Chr.5: 149867903-149867903 on Build GRCh38
rs755625234 Chr.5: 149868064-149868064 on Build GRCh38
rs890327644 Chr.5: 149868064-149868064 on Build GRCh38
rs993078912 Chr.5: 149868228-149868228 on Build GRCh38
rs144970068 Chr.5: 149868150-149868150 on Build GRCh38
rs150486819 Chr.5: 149868220-149868220 on Build GRCh38
rs1053961573 Chr.5: 149867859-149867859 on Build GRCh38
rs1005083015 Chr.5: 149868020-149868020 on Build GRCh38
rs774406349 Chr.5: 149867849-149867849 on Build GRCh38
rs1022012026 Chr.5: 149868153-149868153 on Build GRCh38
rs759432911 Chr.5: 149868091-149868091 on Build GRCh38
rs768740269 Chr.5: 149868182-149868182 on Build GRCh38
rs755688626 Chr.5: 149868124-149868124 on Build GRCh38
rs989225511 Chr.5: 149867870-149867870 on Build GRCh38
rs915903085 Chr.5: 149867952-149867952 on Build GRCh38
rs373679810 Chr.5: 149867843-149867843 on Build GRCh38
rs769927442 Chr.5: 149868276-149868276 on Build GRCh38
rs371526546 Chr.5: 149868255-149868255 on Build GRCh38
rs776762589 Chr.5: 149868185-149868185 on Build GRCh38
rs1037760781 Chr.5: 149867997-149867997 on Build GRCh38
rs977853064 Chr.5: 149868186-149868186 on Build GRCh38
rs545847713 Chr.5: 149868142-149868142 on Build GRCh38
rs1057513831 Chr.5: 149867951-149867951 on Build GRCh38
rs113551142 Chr.5: 149868165-149868165 on Build GRCh38
rs574549728 Chr.5: 149868158-149868158 on Build GRCh38
rs935475529 Chr.5: 149867856-149867856 on Build GRCh38
rs143980724 Chr.5: 149868112-149868112 on Build GRCh38
rs561785039 Chr.5: 149868087-149868087 on Build GRCh38
rs367817449 Chr.5: 149868069-149868069 on Build GRCh38
rs756926282 Chr.5: 149868143-149868143 on Build GRCh38
rs886206925 Chr.5: 149868018-149868018 on Build GRCh38
rs1002573309 Chr.5: 149868003-149868003 on Build GRCh38
rs993288623 Chr.5: 149868013-149868013 on Build GRCh38

View Full Product Details