Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150534515-150535034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDST1
Entrez Gene ID
3340
Gene Name
N-deacetylase and N-sulfotransferase 1
Gene Aliases
HSST, MRT46, NST1
UniGene
Hs.222055
Gene Chromosome Location
Chr.5: 150485783-150558211 on Build GRCh38
Transcripts
NM_001301063.1 NM_001543.4 XM_005268433.1 XM_005268434.2 XM_005268435.4 XM_005268436.3 XM_005268437.4 XM_005268438.1 XM_005268439.1 XM_005268441.1 XM_005268442.3 XM_006714782.1 XM_006714783.1 XM_011537638.2 XM_017009427.1 XM_017009428.1 XM_017009429.1 XM_017009430.1 XM_017009431.1 XM_017009432.1 XM_017009433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7379222 c.1208C>A p.S403Y Chr.5: 150534978-150534978 on Build GRCh38
6723620 c.1126C>G p.L376V Chr.5: 150534896-150534896 on Build GRCh38
1186878 c.1174A>C p.S392R Chr.5: 150534944-150534944 on Build GRCh38
3612547 c.1209C>T p.S403S Chr.5: 150534979-150534979 on Build GRCh38
3852793 c.1228G>A p.A410T Chr.5: 150534998-150534998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563481684 Chr.5: 150534695-150534695 on Build GRCh38
rs920466423 Chr.5: 150534989-150534989 on Build GRCh38
rs948586306 Chr.5: 150534641-150534641 on Build GRCh38
rs1021713897 Chr.5: 150534581-150534581 on Build GRCh38
rs2545341 Chr.5: 150534839-150534839 on Build GRCh38
rs377365968 Chr.5: 150534884-150534884 on Build GRCh38
rs1008996871 Chr.5: 150534711-150534711 on Build GRCh38
rs1021787560 Chr.5: 150534770-150534770 on Build GRCh38
rs767605033 Chr.5: 150534979-150534979 on Build GRCh38
rs967441526 Chr.5: 150534812-150534812 on Build GRCh38
rs757992007 Chr.5: 150534863-150534863 on Build GRCh38
rs996982564 Chr.5: 150534568-150534568 on Build GRCh38
rs1037543603 Chr.5: 150534650-150534650 on Build GRCh38
rs774589898 Chr.5: 150534822-150534822 on Build GRCh38
rs139282708 Chr.5: 150534883-150534883 on Build GRCh38
rs760644473 Chr.5: 150534824-150534824 on Build GRCh38
rs920422671 Chr.5: 150534651-150534651 on Build GRCh38
rs6890511 Chr.5: 150534664-150534664 on Build GRCh38
rs926250549 Chr.5: 150534816-150534816 on Build GRCh38
rs763138768 Chr.5: 150534819-150534819 on Build GRCh38
rs888341264 Chr.5: 150534767-150534767 on Build GRCh38
rs753869718 Chr.5: 150534838-150534838 on Build GRCh38
rs374549152 Chr.5: 150534988-150534988 on Build GRCh38
rs554692999 Chr.5: 150534562-150534562 on Build GRCh38
rs992805648 Chr.5: 150534622-150534622 on Build GRCh38
rs748699863 Chr.5: 150534923-150534923 on Build GRCh38
rs960124289 Chr.5: 150534830-150534830 on Build GRCh38
rs968833454 Chr.5: 150534604-150534604 on Build GRCh38
rs781640507 Chr.5: 150534919-150534919 on Build GRCh38
rs546918913 Chr.5: 150534780-150534780 on Build GRCh38
rs976833173 Chr.5: 150534804-150534804 on Build GRCh38
rs997015204 Chr.5: 150534763-150534763 on Build GRCh38
rs1022935509 Chr.5: 150534859-150534859 on Build GRCh38
rs536442073 Chr.5: 150534540-150534540 on Build GRCh38
rs373708646 Chr.5: 150534827-150534827 on Build GRCh38
rs577889106 Chr.5: 150534668-150534668 on Build GRCh38
rs895966334 Chr.5: 150534592-150534592 on Build GRCh38
rs763443185 Chr.5: 150534744-150534744 on Build GRCh38
rs144446775 Chr.5: 150534687-150534687 on Build GRCh38
rs960294962 Chr.5: 150534621-150534621 on Build GRCh38
rs540654216 Chr.5: 150534607-150534607 on Build GRCh38
rs750085442 Chr.5: 150534854-150534854 on Build GRCh38
rs6890812 Chr.5: 150534809-150534809 on Build GRCh38
rs560999923 Chr.5: 150534907-150534907 on Build GRCh38
rs1055872396 Chr.5: 150534585-150534585 on Build GRCh38
rs542839572 Chr.5: 150534820-150534820 on Build GRCh38
rs915845056 Chr.5: 150534623-150534623 on Build GRCh38
rs769031648 Chr.5: 150534790-150534790 on Build GRCh38
rs113234854 Chr.5: 150534813-150534813 on Build GRCh38
rs770319054 Chr.5: 150534852-150534852 on Build GRCh38
rs924664163 Chr.5: 150534613-150534613 on Build GRCh38
rs965171706 Chr.5: 150534798-150534798 on Build GRCh38
rs371020778 Chr.5: 150534980-150534980 on Build GRCh38
rs770178778 Chr.5: 150534940-150534940 on Build GRCh38
rs778062322 Chr.5: 150534946-150534946 on Build GRCh38
rs761839351 Chr.5: 150534849-150534849 on Build GRCh38
rs759660579 Chr.5: 150535006-150535006 on Build GRCh38
rs371096881 Chr.5: 150534851-150534851 on Build GRCh38
rs998973189 Chr.5: 150534796-150534796 on Build GRCh38
rs572977615 Chr.5: 150534582-150534582 on Build GRCh38
rs1038644990 Chr.5: 150534741-150534741 on Build GRCh38
rs1047919575 Chr.5: 150534669-150534669 on Build GRCh38

View Full Product Details