Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150777756-150778283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM3
Entrez Gene ID
85027
Gene Name
small integral membrane protein 3
Gene Aliases
C5orf62, MST150, NID67
UniGene
Hs.29444
Gene Chromosome Location
Chr.5: 150777946-150796736 on Build GRCh38
Transcripts
NM_032947.4

SNP Detail

SNP ID Chromosome Location
rs77067738 Chr.5: 150778082-150778082 on Build GRCh38
rs969929785 Chr.5: 150777835-150777835 on Build GRCh38
rs1047460538 Chr.5: 150777881-150777881 on Build GRCh38
rs1041029022 Chr.5: 150777949-150777949 on Build GRCh38
rs6873198 Chr.5: 150778138-150778138 on Build GRCh38
rs76036587 Chr.5: 150778081-150778081 on Build GRCh38
rs9324654 Chr.5: 150777862-150777862 on Build GRCh38
rs538983219 Chr.5: 150778105-150778105 on Build GRCh38
rs893930840 Chr.5: 150778016-150778016 on Build GRCh38
rs150407038 Chr.5: 150778077-150778077 on Build GRCh38
rs928297714 Chr.5: 150777847-150777847 on Build GRCh38
rs775810001 Chr.5: 150778089-150778089 on Build GRCh38
rs537256678 Chr.5: 150778152-150778152 on Build GRCh38
rs138887482 Chr.5: 150777824-150777824 on Build GRCh38
rs1032423273 Chr.5: 150778001-150778001 on Build GRCh38
rs1036046273 Chr.5: 150778088-150778088 on Build GRCh38
rs917575648 Chr.5: 150777804-150777804 on Build GRCh38
rs201868736 Chr.5: 150778052-150778052 on Build GRCh38
rs780549615 Chr.5: 150777858-150777858 on Build GRCh38
rs942606117 Chr.5: 150778174-150778174 on Build GRCh38
rs900682122 Chr.5: 150777950-150777950 on Build GRCh38
rs958945845 Chr.5: 150778067-150778067 on Build GRCh38
rs567455899 Chr.5: 150778066-150778066 on Build GRCh38
rs772684102 Chr.5: 150778165-150778165 on Build GRCh38
rs370906772 Chr.5: 150778124-150778124 on Build GRCh38
rs949139818 Chr.5: 150777807-150777807 on Build GRCh38
rs763032676 Chr.5: 150778106-150778106 on Build GRCh38
rs778288686 Chr.5: 150778082-150778082 on Build GRCh38
rs1041392146 Chr.5: 150778233-150778233 on Build GRCh38
rs911174221 Chr.5: 150777893-150777893 on Build GRCh38
rs533725279 Chr.5: 150777953-150777953 on Build GRCh38
rs9324653 Chr.5: 150777798-150777798 on Build GRCh38
rs901185189 Chr.5: 150777973-150777973 on Build GRCh38
rs182183487 Chr.5: 150778037-150778037 on Build GRCh38
rs151275929 Chr.5: 150777892-150777894 on Build GRCh38
rs1034640578 Chr.5: 150777782-150777782 on Build GRCh38
rs972468905 Chr.5: 150777814-150777814 on Build GRCh38
rs933511422 Chr.5: 150778245-150778245 on Build GRCh38
rs577363073 Chr.5: 150777974-150777974 on Build GRCh38
rs186453037 Chr.5: 150778071-150778071 on Build GRCh38
rs764039752 Chr.5: 150777797-150777797 on Build GRCh38
rs563963161 Chr.5: 150777987-150777987 on Build GRCh38
rs911066146 Chr.5: 150778159-150778159 on Build GRCh38
rs9324655 Chr.5: 150777924-150777924 on Build GRCh38
rs768991594 Chr.5: 150778110-150778110 on Build GRCh38
rs367829174 Chr.5: 150777881-150777881 on Build GRCh38
rs904690000 Chr.5: 150778023-150778023 on Build GRCh38
rs138706166 Chr.5: 150778128-150778128 on Build GRCh38
rs189758318 Chr.5: 150777951-150777951 on Build GRCh38
rs997754474 Chr.5: 150777956-150777956 on Build GRCh38
rs1039143371 Chr.5: 150777931-150777931 on Build GRCh38
rs1010315055 Chr.5: 150778020-150778020 on Build GRCh38
rs142896672 Chr.5: 150777841-150777841 on Build GRCh38
rs1041848525 Chr.5: 150778022-150778022 on Build GRCh38

View Full Product Details